Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44644606-44645098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF220
Entrez Gene ID
55182
Gene Name
ring finger protein 220
Gene Aliases
C1orf164
UniGene
Hs.456557
Gene Chromosome Location
Chr.1: 44404846-44651724 on Build GRCh38
Transcripts
NM_001319956.1 NM_001319957.1 NM_018150.3 XM_005270992.1 XM_005270993.1 XM_005270996.1 XM_011541698.1 XM_011541699.1 XM_011541700.1 XM_011541702.1 XM_017001623.1 XM_017001624.1 XM_017001625.1 XM_017001626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5993195 c.1133G>T p.G378V Chr.1: 44644704-44644704 on Build GRCh38
5231908 c.382G>C p.E128Q Chr.1: 44644727-44644727 on Build GRCh38
5231907 c.517G>C p.E173Q Chr.1: 44644727-44644727 on Build GRCh38
314855 c.1184A>T p.D395V Chr.1: 44644755-44644755 on Build GRCh38
5993191 c.1132G>T p.G378C Chr.1: 44644703-44644703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974030507 Chr.1: 44644971-44644971 on Build GRCh38
rs754536964 Chr.1: 44644718-44644718 on Build GRCh38
rs760604843 Chr.1: 44644682-44644682 on Build GRCh38
rs960528935 Chr.1: 44644628-44644628 on Build GRCh38
rs1044357721 Chr.1: 44644872-44644872 on Build GRCh38
rs766061302 Chr.1: 44644817-44644817 on Build GRCh38
rs1055625924 Chr.1: 44644659-44644659 on Build GRCh38
rs200720362 Chr.1: 44645071-44645071 on Build GRCh38
rs767475531 Chr.1: 44644664-44644664 on Build GRCh38
rs759844996 Chr.1: 44644662-44644662 on Build GRCh38
rs758117084 Chr.1: 44644701-44644701 on Build GRCh38
rs996467393 Chr.1: 44644868-44644868 on Build GRCh38
rs780201372 Chr.1: 44644846-44644846 on Build GRCh38
rs530667608 Chr.1: 44644944-44644944 on Build GRCh38
rs749593281 Chr.1: 44644657-44644657 on Build GRCh38
rs116503928 Chr.1: 44645070-44645070 on Build GRCh38
rs200316239 Chr.1: 44644728-44644728 on Build GRCh38
rs113047812 Chr.1: 44644918-44644918 on Build GRCh38
rs780877912 Chr.1: 44644829-44644829 on Build GRCh38
rs199591500 Chr.1: 44645046-44645046 on Build GRCh38
rs150037632 Chr.1: 44645011-44645011 on Build GRCh38
rs746583462 Chr.1: 44644711-44644711 on Build GRCh38
rs544545271 Chr.1: 44644890-44644890 on Build GRCh38
rs778362041 Chr.1: 44644720-44644720 on Build GRCh38
rs776512643 Chr.1: 44644790-44644790 on Build GRCh38
rs368455337 Chr.1: 44644658-44644658 on Build GRCh38
rs775608740 Chr.1: 44644667-44644667 on Build GRCh38
rs761851964 Chr.1: 44644801-44644801 on Build GRCh38
rs960475332 Chr.1: 44644891-44644891 on Build GRCh38
rs756940103 Chr.1: 44644813-44644813 on Build GRCh38
rs145131553 Chr.1: 44645018-44645018 on Build GRCh38
rs936951859 Chr.1: 44644629-44644629 on Build GRCh38
rs761640157 Chr.1: 44645013-44645013 on Build GRCh38
rs916389502 Chr.1: 44644636-44644636 on Build GRCh38
rs779146030 Chr.1: 44644841-44644841 on Build GRCh38
rs778546662 Chr.1: 44644813-44644814 on Build GRCh38
rs369982770 Chr.1: 44644949-44644949 on Build GRCh38
rs762748541 Chr.1: 44645032-44645032 on Build GRCh38
rs557912233 Chr.1: 44644811-44644811 on Build GRCh38
rs775698325 Chr.1: 44644735-44644735 on Build GRCh38
rs770481596 Chr.1: 44644647-44644647 on Build GRCh38
rs749739685 Chr.1: 44644833-44644833 on Build GRCh38
rs757981872 Chr.1: 44644814-44644814 on Build GRCh38
rs763613540 Chr.1: 44644654-44644654 on Build GRCh38
rs746181029 Chr.1: 44644843-44644843 on Build GRCh38
rs969613169 Chr.1: 44644967-44644967 on Build GRCh38
rs999106755 Chr.1: 44644888-44644888 on Build GRCh38
rs374079139 Chr.1: 44644825-44644825 on Build GRCh38
rs753445544 Chr.1: 44644656-44644663 on Build GRCh38
rs767125160 Chr.1: 44645076-44645076 on Build GRCh38
rs1052841126 Chr.1: 44644634-44644634 on Build GRCh38
rs201292571 Chr.1: 44644656-44644656 on Build GRCh38
rs753786574 Chr.1: 44644693-44644693 on Build GRCh38
rs750012378 Chr.1: 44644698-44644698 on Build GRCh38
rs535375215 Chr.1: 44644721-44644721 on Build GRCh38
rs764917122 Chr.1: 44644807-44644807 on Build GRCh38
rs768693048 Chr.1: 44644959-44644959 on Build GRCh38
rs555385582 Chr.1: 44644726-44644726 on Build GRCh38
rs772408858 Chr.1: 44644734-44644734 on Build GRCh38
rs41269055 Chr.1: 44644885-44644885 on Build GRCh38
rs150936501 Chr.1: 44644759-44644759 on Build GRCh38
rs772440016 Chr.1: 44644844-44644844 on Build GRCh38
rs751266675 Chr.1: 44644818-44644818 on Build GRCh38
rs867278984 Chr.1: 44644906-44644906 on Build GRCh38
rs773026242 Chr.1: 44645025-44645025 on Build GRCh38
rs913882159 Chr.1: 44644861-44644861 on Build GRCh38
rs779698056 Chr.1: 44644710-44644710 on Build GRCh38
rs147534605 Chr.1: 44644732-44644732 on Build GRCh38
rs200299898 Chr.1: 44644812-44644812 on Build GRCh38
rs757843181 Chr.1: 44644834-44644834 on Build GRCh38
rs756955984 Chr.1: 44644694-44644694 on Build GRCh38
rs763952963 Chr.1: 44644689-44644689 on Build GRCh38
rs764181984 Chr.1: 44644765-44644765 on Build GRCh38
rs991917083 Chr.1: 44644633-44644633 on Build GRCh38

View Full Product Details