Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44820379-44820878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTCH2
Entrez Gene ID
8643
Gene Name
patched 2
Gene Aliases
PTC2
UniGene
Hs.591497
Gene Chromosome Location
Chr.1: 44819844-44842944 on Build GRCh38
Transcripts
NM_001166292.1

SNP Detail

SNP ID Chromosome Location
rs899351780 Chr.1: 44820693-44820693 on Build GRCh38
rs895946007 Chr.1: 44820400-44820400 on Build GRCh38
rs181152082 Chr.1: 44820404-44820404 on Build GRCh38
rs1028945696 Chr.1: 44820590-44820590 on Build GRCh38
rs1031089818 Chr.1: 44820654-44820654 on Build GRCh38
rs369312707 Chr.1: 44820772-44820772 on Build GRCh38
rs1047482160 Chr.1: 44820709-44820709 on Build GRCh38
rs917513311 Chr.1: 44820667-44820667 on Build GRCh38
rs748510565 Chr.1: 44820703-44820703 on Build GRCh38
rs371688861 Chr.1: 44820658-44820658 on Build GRCh38
rs950404176 Chr.1: 44820506-44820506 on Build GRCh38
rs1018871281 Chr.1: 44820583-44820583 on Build GRCh38
rs769009116 Chr.1: 44820613-44820613 on Build GRCh38
rs944044146 Chr.1: 44820797-44820797 on Build GRCh38
rs189756125 Chr.1: 44820650-44820650 on Build GRCh38
rs931575913 Chr.1: 44820762-44820762 on Build GRCh38
rs994520787 Chr.1: 44820696-44820696 on Build GRCh38
rs545293102 Chr.1: 44820516-44820516 on Build GRCh38
rs759068550 Chr.1: 44820852-44820852 on Build GRCh38
rs773535130 Chr.1: 44820729-44820729 on Build GRCh38
rs1041189137 Chr.1: 44820799-44820799 on Build GRCh38
rs373328320 Chr.1: 44820438-44820438 on Build GRCh38
rs949019496 Chr.1: 44820672-44820672 on Build GRCh38
rs766379252 Chr.1: 44820403-44820403 on Build GRCh38
rs777079047 Chr.1: 44820688-44820688 on Build GRCh38
rs915093655 Chr.1: 44820648-44820648 on Build GRCh38
rs947482386 Chr.1: 44820649-44820649 on Build GRCh38
rs529863437 Chr.1: 44820748-44820748 on Build GRCh38
rs948216599 Chr.1: 44820408-44820408 on Build GRCh38
rs747435195 Chr.1: 44820661-44820661 on Build GRCh38
rs11573603 Chr.1: 44820657-44820657 on Build GRCh38
rs754821728 Chr.1: 44820407-44820407 on Build GRCh38
rs771943530 Chr.1: 44820439-44820439 on Build GRCh38
rs560944695 Chr.1: 44820664-44820664 on Build GRCh38
rs994573127 Chr.1: 44820495-44820495 on Build GRCh38
rs987318862 Chr.1: 44820543-44820543 on Build GRCh38
rs185397548 Chr.1: 44820457-44820457 on Build GRCh38
rs866542297 Chr.1: 44820476-44820476 on Build GRCh38
rs1025916137 Chr.1: 44820499-44820499 on Build GRCh38
rs752347446 Chr.1: 44820428-44820428 on Build GRCh38
rs12079597 Chr.1: 44820421-44820421 on Build GRCh38
rs867899024 Chr.1: 44820778-44820778 on Build GRCh38
rs558442200 Chr.1: 44820644-44820644 on Build GRCh38
rs1045329698 Chr.1: 44820416-44820416 on Build GRCh38
rs931342354 Chr.1: 44820453-44820456 on Build GRCh38
rs964861460 Chr.1: 44820605-44820605 on Build GRCh38
rs1019065622 Chr.1: 44820824-44820824 on Build GRCh38
rs901709546 Chr.1: 44820570-44820570 on Build GRCh38
rs1047268826 Chr.1: 44820764-44820764 on Build GRCh38
rs372863068 Chr.1: 44820783-44820783 on Build GRCh38
rs375003257 Chr.1: 44820713-44820713 on Build GRCh38
rs766317327 Chr.1: 44820751-44820751 on Build GRCh38
rs767173269 Chr.1: 44820417-44820417 on Build GRCh38
rs549549303 Chr.1: 44820636-44820636 on Build GRCh38
rs1008520356 Chr.1: 44820782-44820782 on Build GRCh38
rs776224345 Chr.1: 44820792-44820792 on Build GRCh38
rs964650763 Chr.1: 44820833-44820833 on Build GRCh38

View Full Product Details