Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16518561-16519070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf44
Entrez Gene ID
84167
Gene Name
chromosome 19 open reading frame 44
Gene Aliases
-
UniGene
Hs.631627
Gene Chromosome Location
Chr.19: 16496311-16521369 on Build GRCh38
Transcripts
NM_032207.3 XM_006722920.3 NM_006387.5

Gene Symbol
CHERP
Entrez Gene ID
10523
Gene Name
calcium homeostasis endoplasmic reticulum protein
Gene Aliases
DAN16, SCAF6, SRA1
UniGene
Hs.740364
Gene Chromosome Location
Chr.19: 16517889-16542452 on Build GRCh38
Transcripts
NM_032207.3 XM_006722920.3 NM_006387.5

SNP Detail

SNP ID Chromosome Location
rs969745640 Chr.19: 16518863-16518863 on Build GRCh38
rs553369429 Chr.19: 16518726-16518726 on Build GRCh38
rs1008350115 Chr.19: 16518995-16518995 on Build GRCh38
rs977363864 Chr.19: 16518871-16518871 on Build GRCh38
rs769729690 Chr.19: 16518661-16518661 on Build GRCh38
rs900749999 Chr.19: 16518955-16518955 on Build GRCh38
rs916509716 Chr.19: 16518851-16518851 on Build GRCh38
rs531994416 Chr.19: 16518913-16518913 on Build GRCh38
rs556984680 Chr.19: 16518650-16518650 on Build GRCh38
rs899582585 Chr.19: 16519012-16519012 on Build GRCh38
rs963367762 Chr.19: 16518839-16518839 on Build GRCh38
rs537190326 Chr.19: 16519042-16519042 on Build GRCh38
rs868618393 Chr.19: 16518675-16518675 on Build GRCh38
rs577160232 Chr.19: 16518679-16518679 on Build GRCh38
rs1014852044 Chr.19: 16518836-16518836 on Build GRCh38
rs528353360 Chr.19: 16518993-16518993 on Build GRCh38
rs769062797 Chr.19: 16518835-16518835 on Build GRCh38
rs369864628 Chr.19: 16518981-16518981 on Build GRCh38
rs973309922 Chr.19: 16518676-16518676 on Build GRCh38
rs924599470 Chr.19: 16518875-16518875 on Build GRCh38
rs181417043 Chr.19: 16518845-16518845 on Build GRCh38
rs893488028 Chr.19: 16518727-16518727 on Build GRCh38
rs555774684 Chr.19: 16518969-16518969 on Build GRCh38
rs547989327 Chr.19: 16519013-16519013 on Build GRCh38
rs1030421806 Chr.19: 16518611-16518611 on Build GRCh38
rs927728979 Chr.19: 16518701-16518701 on Build GRCh38
rs954991724 Chr.19: 16518619-16518619 on Build GRCh38
rs907906819 Chr.19: 16518648-16518648 on Build GRCh38
rs375535436 Chr.19: 16518918-16518918 on Build GRCh38
rs1039420266 Chr.19: 16518939-16518939 on Build GRCh38
rs996349439 Chr.19: 16518756-16518756 on Build GRCh38
rs935065947 Chr.19: 16518879-16518879 on Build GRCh38
rs1043329237 Chr.19: 16518743-16518743 on Build GRCh38
rs116593693 Chr.19: 16518898-16518898 on Build GRCh38
rs565858119 Chr.19: 16518992-16518992 on Build GRCh38
rs962401505 Chr.19: 16518631-16518632 on Build GRCh38
rs947916308 Chr.19: 16518695-16518695 on Build GRCh38
rs535821588 Chr.19: 16518760-16518760 on Build GRCh38
rs116799974 Chr.19: 16518738-16518738 on Build GRCh38
rs995134599 Chr.19: 16518786-16518786 on Build GRCh38
rs879649905 Chr.19: 16518718-16518718 on Build GRCh38
rs975339447 Chr.19: 16518670-16518670 on Build GRCh38
rs948883574 Chr.19: 16518940-16518940 on Build GRCh38
rs1021501765 Chr.19: 16518852-16518852 on Build GRCh38
rs546510592 Chr.19: 16518700-16518700 on Build GRCh38
rs1038280002 Chr.19: 16518957-16518957 on Build GRCh38
rs916393622 Chr.19: 16518684-16518684 on Build GRCh38
rs1025366202 Chr.19: 16518642-16518642 on Build GRCh38
rs1016950042 Chr.19: 16518623-16518623 on Build GRCh38
rs114767194 Chr.19: 16518870-16518870 on Build GRCh38
rs774219433 Chr.19: 16518914-16518914 on Build GRCh38
rs11539314 Chr.19: 16519033-16519033 on Build GRCh38
rs111525430 Chr.19: 16518806-16518806 on Build GRCh38
rs1019457421 Chr.19: 16518654-16518654 on Build GRCh38
rs73513183 Chr.19: 16518638-16518638 on Build GRCh38
rs761523635 Chr.19: 16519010-16519010 on Build GRCh38
rs886829154 Chr.19: 16518808-16518808 on Build GRCh38
rs981965694 Chr.19: 16518866-16518866 on Build GRCh38
rs1007965250 Chr.19: 16518811-16518811 on Build GRCh38
rs542460648 Chr.19: 16518751-16518751 on Build GRCh38
rs929559744 Chr.19: 16518980-16518980 on Build GRCh38
rs899424653 Chr.19: 16518769-16518769 on Build GRCh38
rs138776573 Chr.19: 16519044-16519044 on Build GRCh38
rs938705782 Chr.19: 16518878-16518878 on Build GRCh38
rs1019352952 Chr.19: 16519000-16519000 on Build GRCh38

View Full Product Details