Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18340477-18341006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGPEP1
Entrez Gene ID
54858
Gene Name
pyroglutamyl-peptidase I
Gene Aliases
PAP-I, PGI, PGP, PGP-I, PGPI, Pcp
UniGene
Hs.131776
Gene Chromosome Location
Chr.19: 18340573-18369953 on Build GRCh38
Transcripts
NM_001300927.1 NM_001308366.1 NM_017712.3 XM_006722783.3 XM_011528111.2 XM_011528112.1 XM_011528115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4566873 c.3_4GG>AA p.M1_E2>IK Chr.19: 18340684-18340685 on Build GRCh38
7402130 c.4G>C p.E2Q Chr.19: 18340685-18340685 on Build GRCh38
6744704 c.18G>T p.K6N Chr.19: 18340699-18340699 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs570498910 Chr.19: 18340625-18340625 on Build GRCh38
rs974175590 Chr.19: 18340522-18340522 on Build GRCh38
rs906149380 Chr.19: 18340769-18340769 on Build GRCh38
rs868299334 Chr.19: 18340598-18340598 on Build GRCh38
rs1012172366 Chr.19: 18340637-18340637 on Build GRCh38
rs558517831 Chr.19: 18340848-18340848 on Build GRCh38
rs768594756 Chr.19: 18340658-18340658 on Build GRCh38
rs550528452 Chr.19: 18340586-18340586 on Build GRCh38
rs774252221 Chr.19: 18340671-18340671 on Build GRCh38
rs758861288 Chr.19: 18340648-18340648 on Build GRCh38
rs953160378 Chr.19: 18340621-18340621 on Build GRCh38
rs761885894 Chr.19: 18340672-18340672 on Build GRCh38
rs916805607 Chr.19: 18340509-18340509 on Build GRCh38
rs931750897 Chr.19: 18340508-18340508 on Build GRCh38
rs79058301 Chr.19: 18340520-18340520 on Build GRCh38
rs942368410 Chr.19: 18340564-18340564 on Build GRCh38
rs540272440 Chr.19: 18340870-18340870 on Build GRCh38
rs985948226 Chr.19: 18340842-18340842 on Build GRCh38
rs575812262 Chr.19: 18340771-18340771 on Build GRCh38
rs1020650715 Chr.19: 18340597-18340597 on Build GRCh38
rs564959179 Chr.19: 18340984-18340984 on Build GRCh38
rs765620545 Chr.19: 18340720-18340720 on Build GRCh38
rs985079413 Chr.19: 18340890-18340890 on Build GRCh38
rs1041400575 Chr.19: 18340971-18340971 on Build GRCh38
rs544904714 Chr.19: 18340797-18340797 on Build GRCh38
rs947278429 Chr.19: 18340806-18340806 on Build GRCh38
rs1056551995 Chr.19: 18340942-18340942 on Build GRCh38
rs951682149 Chr.19: 18340938-18340938 on Build GRCh38
rs902996803 Chr.19: 18340824-18340824 on Build GRCh38
rs975200269 Chr.19: 18340948-18340948 on Build GRCh38
rs553180914 Chr.19: 18340646-18340646 on Build GRCh38
rs754292740 Chr.19: 18340643-18340643 on Build GRCh38
rs1002252804 Chr.19: 18340562-18340562 on Build GRCh38
rs974332725 Chr.19: 18340669-18340669 on Build GRCh38
rs4808784 Chr.19: 18340851-18340851 on Build GRCh38
rs370131707 Chr.19: 18340757-18340757 on Build GRCh38
rs979045516 Chr.19: 18340608-18340608 on Build GRCh38
rs559952224 Chr.19: 18340885-18340885 on Build GRCh38
rs947059290 Chr.19: 18340970-18340971 on Build GRCh38
rs760427293 Chr.19: 18340639-18340639 on Build GRCh38
rs1050134313 Chr.19: 18340512-18340512 on Build GRCh38
rs765932192 Chr.19: 18340641-18340641 on Build GRCh38
rs7251109 Chr.19: 18340551-18340551 on Build GRCh38
rs925730839 Chr.19: 18340504-18340504 on Build GRCh38
rs965782113 Chr.19: 18340862-18340862 on Build GRCh38
rs781277607 Chr.19: 18340654-18340654 on Build GRCh38
rs898062399 Chr.19: 18340548-18340548 on Build GRCh38
rs944382157 Chr.19: 18340636-18340636 on Build GRCh38
rs867271277 Chr.19: 18340628-18340628 on Build GRCh38
rs753441922 Chr.19: 18340707-18340707 on Build GRCh38
rs539493711 Chr.19: 18340644-18340644 on Build GRCh38
rs1009508134 Chr.19: 18340590-18340590 on Build GRCh38
rs960715323 Chr.19: 18340588-18340588 on Build GRCh38
rs938618252 Chr.19: 18340975-18340975 on Build GRCh38
rs905321038 Chr.19: 18340561-18340561 on Build GRCh38
rs772839565 Chr.19: 18340682-18340682 on Build GRCh38
rs972185943 Chr.19: 18340865-18340865 on Build GRCh38
rs1020955288 Chr.19: 18340740-18340740 on Build GRCh38
rs528988825 Chr.19: 18340943-18340943 on Build GRCh38
rs771253661 Chr.19: 18340652-18340652 on Build GRCh38
rs759136853 Chr.19: 18340715-18340715 on Build GRCh38
rs531525402 Chr.19: 18340577-18340577 on Build GRCh38
rs945469353 Chr.19: 18340946-18340946 on Build GRCh38
rs1033317509 Chr.19: 18340789-18340789 on Build GRCh38
rs1025959454 Chr.19: 18340562-18340573 on Build GRCh38
rs760095375 Chr.19: 18340684-18340684 on Build GRCh38
rs1049743684 Chr.19: 18340668-18340668 on Build GRCh38
rs781286108 Chr.19: 18340759-18340759 on Build GRCh38
rs7251555 Chr.19: 18340571-18340571 on Build GRCh38
rs753028184 Chr.19: 18340730-18340730 on Build GRCh38
rs1026341393 Chr.19: 18340937-18340938 on Build GRCh38
rs1042055780 Chr.19: 18340811-18340811 on Build GRCh38
rs985895735 Chr.19: 18340622-18340622 on Build GRCh38
rs542431818 Chr.19: 18340951-18340951 on Build GRCh38
rs778127849 Chr.19: 18340734-18340734 on Build GRCh38
rs746030271 Chr.19: 18340655-18340655 on Build GRCh38
rs771884829 Chr.19: 18340678-18340678 on Build GRCh38
rs765846256 Chr.19: 18340693-18340693 on Build GRCh38
rs1036639829 Chr.19: 18340541-18340541 on Build GRCh38
rs758951022 Chr.19: 18340733-18340733 on Build GRCh38
rs938213997 Chr.19: 18340931-18340931 on Build GRCh38
rs747036516 Chr.19: 18340650-18340650 on Build GRCh38
rs535627193 Chr.19: 18340649-18340649 on Build GRCh38
rs560242249 Chr.19: 18340527-18340527 on Build GRCh38
rs951662920 Chr.19: 18340683-18340683 on Build GRCh38
rs903870707 Chr.19: 18340971-18340971 on Build GRCh38
rs1056499448 Chr.19: 18340696-18340696 on Build GRCh38
rs925814458 Chr.19: 18340618-18340618 on Build GRCh38
rs903049479 Chr.19: 18340616-18340616 on Build GRCh38
rs558396213 Chr.19: 18340525-18340525 on Build GRCh38
rs752038433 Chr.19: 18340754-18340754 on Build GRCh38
rs749873442 Chr.19: 18340634-18340634 on Build GRCh38
rs549170362 Chr.19: 18340557-18340557 on Build GRCh38
rs192653999 Chr.19: 18340497-18340497 on Build GRCh38
rs746127218 Chr.19: 18340764-18340764 on Build GRCh38
rs926861602 Chr.19: 18340924-18340924 on Build GRCh38
rs2302946 Chr.19: 18340647-18340647 on Build GRCh38

View Full Product Details