Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18847654-18848164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UPF1
Entrez Gene ID
5976
Gene Name
UPF1, RNA helicase and ATPase
Gene Aliases
HUPF1, NORF1, RENT1, pNORF1, smg-2
UniGene
Hs.515266
Gene Chromosome Location
Chr.19: 18831926-18868232 on Build GRCh38
Transcripts
NM_001297549.1 NM_002911.3 XM_017027105.1 XM_017027106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1494196 c.372-2A>T p.? Chr.19: 18847742-18847742 on Build GRCh38
4517667 c.391_392CC>TT p.P131F Chr.19: 18847763-18847764 on Build GRCh38
993164 c.400G>A p.V134M Chr.19: 18847772-18847772 on Build GRCh38
6267438 c.461G>T p.S154I Chr.19: 18847833-18847833 on Build GRCh38
6267437 c.461G>T p.S154I Chr.19: 18847833-18847833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770734294 Chr.19: 18847992-18847992 on Build GRCh38
rs763413453 Chr.19: 18847702-18847702 on Build GRCh38
rs750848887 Chr.19: 18847876-18847876 on Build GRCh38
rs73526913 Chr.19: 18847753-18847753 on Build GRCh38
rs769471927 Chr.19: 18847857-18847857 on Build GRCh38
rs951383281 Chr.19: 18847919-18847919 on Build GRCh38
rs768109886 Chr.19: 18847861-18847861 on Build GRCh38
rs777945974 Chr.19: 18847916-18847916 on Build GRCh38
rs752056333 Chr.19: 18847716-18847716 on Build GRCh38
rs531750821 Chr.19: 18847967-18847967 on Build GRCh38
rs997157 Chr.19: 18848004-18848004 on Build GRCh38
rs906768498 Chr.19: 18847803-18847803 on Build GRCh38
rs553332952 Chr.19: 18847813-18847813 on Build GRCh38
rs746974593 Chr.19: 18847932-18847932 on Build GRCh38
rs764634848 Chr.19: 18847707-18847707 on Build GRCh38
rs745477956 Chr.19: 18847838-18847838 on Build GRCh38
rs531039736 Chr.19: 18847931-18847931 on Build GRCh38
rs762532671 Chr.19: 18848114-18848114 on Build GRCh38
rs748379903 Chr.19: 18847810-18847810 on Build GRCh38
rs753936939 Chr.19: 18847760-18847760 on Build GRCh38
rs998284 Chr.19: 18847887-18847887 on Build GRCh38
rs776507902 Chr.19: 18848077-18848077 on Build GRCh38
rs774919696 Chr.19: 18847868-18847868 on Build GRCh38
rs371953773 Chr.19: 18847870-18847870 on Build GRCh38
rs370145414 Chr.19: 18847697-18847697 on Build GRCh38
rs568176952 Chr.19: 18847733-18847733 on Build GRCh38
rs183892627 Chr.19: 18848092-18848092 on Build GRCh38
rs746983401 Chr.19: 18847791-18847791 on Build GRCh38
rs780240552 Chr.19: 18847759-18847759 on Build GRCh38
rs963464637 Chr.19: 18848018-18848018 on Build GRCh38
rs775007388 Chr.19: 18847700-18847700 on Build GRCh38
rs374584074 Chr.19: 18847771-18847771 on Build GRCh38
rs777749373 Chr.19: 18847778-18847778 on Build GRCh38
rs188036994 Chr.19: 18847884-18847884 on Build GRCh38
rs1017883856 Chr.19: 18847854-18847854 on Build GRCh38
rs551886428 Chr.19: 18848052-18848052 on Build GRCh38
rs750607861 Chr.19: 18847726-18847726 on Build GRCh38
rs767865977 Chr.19: 18847873-18847873 on Build GRCh38
rs745700469 Chr.19: 18848086-18848086 on Build GRCh38
rs368539038 Chr.19: 18847859-18847859 on Build GRCh38
rs116836107 Chr.19: 18848106-18848106 on Build GRCh38
rs998057325 Chr.19: 18847891-18847891 on Build GRCh38
rs761126334 Chr.19: 18847722-18847723 on Build GRCh38
rs763598959 Chr.19: 18848123-18848123 on Build GRCh38
rs758610458 Chr.19: 18847866-18847866 on Build GRCh38
rs192882104 Chr.19: 18848057-18848057 on Build GRCh38
rs762227342 Chr.19: 18847725-18847725 on Build GRCh38
rs935602976 Chr.19: 18848124-18848124 on Build GRCh38
rs990930716 Chr.19: 18848035-18848035 on Build GRCh38
rs985750858 Chr.19: 18848030-18848030 on Build GRCh38
rs780975568 Chr.19: 18847836-18847836 on Build GRCh38

View Full Product Details