Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 20005747-20006294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF682
Entrez Gene ID
91120
Gene Name
zinc finger protein 682
Gene Aliases
BC39498_3
UniGene
Hs.306298
Gene Chromosome Location
Chr.19: 20004418-20039468 on Build GRCh38
Transcripts
NM_001077349.1 NM_033196.2 XM_017027455.1 XM_017027456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3101998 c.1419_1420delAG p.R473fs*11 Chr.19: 20006082-20006083 on Build GRCh38
6832389 c.1494G>A p.T498T Chr.19: 20006008-20006008 on Build GRCh38
3101997 c.1455delA p.K485fs*>14 Chr.19: 20006047-20006047 on Build GRCh38
6869599 c.1465G>A p.A489T Chr.19: 20006037-20006037 on Build GRCh38
1480830 c.1355G>A p.R452H Chr.19: 20006147-20006147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771073616 Chr.19: 20006173-20006173 on Build GRCh38
rs761015990 Chr.19: 20006165-20006165 on Build GRCh38
rs77288754 Chr.19: 20005968-20005968 on Build GRCh38
rs778285984 Chr.19: 20006237-20006237 on Build GRCh38
rs766548876 Chr.19: 20006069-20006069 on Build GRCh38
rs760778744 Chr.19: 20006060-20006060 on Build GRCh38
rs202023858 Chr.19: 20006196-20006196 on Build GRCh38
rs769391025 Chr.19: 20005977-20005977 on Build GRCh38
rs772386581 Chr.19: 20006009-20006009 on Build GRCh38
rs1008676961 Chr.19: 20005774-20005774 on Build GRCh38
rs367928960 Chr.19: 20006129-20006129 on Build GRCh38
rs12327700 Chr.19: 20005832-20005832 on Build GRCh38
rs527573632 Chr.19: 20005944-20005944 on Build GRCh38
rs751505519 Chr.19: 20006103-20006103 on Build GRCh38
rs944195066 Chr.19: 20006132-20006132 on Build GRCh38
rs781027299 Chr.19: 20005962-20005962 on Build GRCh38
rs747340915 Chr.19: 20006240-20006240 on Build GRCh38
rs371187151 Chr.19: 20006008-20006008 on Build GRCh38
rs184298649 Chr.19: 20006123-20006123 on Build GRCh38
rs371737058 Chr.19: 20006148-20006148 on Build GRCh38
rs945555645 Chr.19: 20006261-20006261 on Build GRCh38
rs892663239 Chr.19: 20006090-20006090 on Build GRCh38
rs967954543 Chr.19: 20005901-20005901 on Build GRCh38
rs748722166 Chr.19: 20006229-20006229 on Build GRCh38
rs77200511 Chr.19: 20005988-20005988 on Build GRCh38
rs182845369 Chr.19: 20005953-20005953 on Build GRCh38
rs370794225 Chr.19: 20005784-20005798 on Build GRCh38
rs936762327 Chr.19: 20006235-20006235 on Build GRCh38
rs755877055 Chr.19: 20006195-20006195 on Build GRCh38
rs753850951 Chr.19: 20006072-20006072 on Build GRCh38
rs751709123 Chr.19: 20005987-20005987 on Build GRCh38
rs984615181 Chr.19: 20005870-20005870 on Build GRCh38
rs752982898 Chr.19: 20006113-20006113 on Build GRCh38
rs150453306 Chr.19: 20005831-20005831 on Build GRCh38
rs1019955264 Chr.19: 20005802-20005802 on Build GRCh38
rs542943665 Chr.19: 20006082-20006083 on Build GRCh38
rs745466116 Chr.19: 20005974-20005974 on Build GRCh38
rs1053924451 Chr.19: 20006079-20006079 on Build GRCh38
rs749214587 Chr.19: 20006046-20006052 on Build GRCh38
rs368709233 Chr.19: 20006147-20006147 on Build GRCh38
rs774483295 Chr.19: 20006180-20006180 on Build GRCh38
rs530531525 Chr.19: 20005858-20005858 on Build GRCh38
rs374716859 Chr.19: 20005960-20005960 on Build GRCh38
rs370479013 Chr.19: 20006174-20006174 on Build GRCh38
rs191905148 Chr.19: 20006097-20006097 on Build GRCh38
rs932903662 Chr.19: 20006111-20006111 on Build GRCh38
rs78187010 Chr.19: 20005967-20005967 on Build GRCh38
rs971089285 Chr.19: 20005806-20005806 on Build GRCh38
rs141272196 Chr.19: 20005925-20005925 on Build GRCh38
rs926503670 Chr.19: 20005941-20005941 on Build GRCh38
rs761807885 Chr.19: 20006184-20006184 on Build GRCh38
rs140867326 Chr.19: 20005783-20005797 on Build GRCh38
rs1009831785 Chr.19: 20005775-20005775 on Build GRCh38
rs187018165 Chr.19: 20005983-20005983 on Build GRCh38
rs754505056 Chr.19: 20006212-20006212 on Build GRCh38
rs779670117 Chr.19: 20006126-20006126 on Build GRCh38
rs760317456 Chr.19: 20006257-20006257 on Build GRCh38
rs753487516 Chr.19: 20006205-20006205 on Build GRCh38
rs754989598 Chr.19: 20006057-20006057 on Build GRCh38
rs201774459 Chr.19: 20006143-20006143 on Build GRCh38
rs940576753 Chr.19: 20005906-20005906 on Build GRCh38
rs569784520 Chr.19: 20006110-20006110 on Build GRCh38
rs771605732 Chr.19: 20006178-20006178 on Build GRCh38
rs145357593 Chr.19: 20005890-20005890 on Build GRCh38
rs779751099 Chr.19: 20005963-20005964 on Build GRCh38
rs1038383571 Chr.19: 20006031-20006031 on Build GRCh38
rs1002512013 Chr.19: 20006125-20006125 on Build GRCh38
rs771290510 Chr.19: 20006242-20006242 on Build GRCh38
rs551308675 Chr.19: 20005918-20005918 on Build GRCh38
rs560099130 Chr.19: 20005796-20005796 on Build GRCh38
rs778370819 Chr.19: 20006222-20006222 on Build GRCh38
rs74463300 Chr.19: 20005966-20005966 on Build GRCh38
rs373888671 Chr.19: 20006185-20006185 on Build GRCh38
rs369078395 Chr.19: 20006155-20006155 on Build GRCh38
rs773444580 Chr.19: 20006022-20006022 on Build GRCh38
rs375618664 Chr.19: 20006112-20006112 on Build GRCh38
rs372296532 Chr.19: 20006105-20006105 on Build GRCh38
rs17679334 Chr.19: 20006154-20006154 on Build GRCh38
rs750169359 Chr.19: 20006190-20006190 on Build GRCh38
rs758893719 Chr.19: 20006231-20006231 on Build GRCh38
rs774780429 Chr.19: 20006002-20006002 on Build GRCh38
rs746069446 Chr.19: 20006252-20006252 on Build GRCh38
rs765363888 Chr.19: 20006179-20006179 on Build GRCh38

View Full Product Details