Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 21537274-21537801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF429
Entrez Gene ID
353088
Gene Name
zinc finger protein 429
Gene Aliases
-
UniGene
Hs.572567
Gene Chromosome Location
Chr.19: 21505257-21541157 on Build GRCh38
Transcripts
NM_001001415.2 XM_017026748.1 XM_017026749.1 XM_017026750.1 XM_017026751.1 XM_017026752.1 XM_017026753.1 XM_017026754.1 XM_017026755.1 XM_017026756.1 XM_017026757.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4463786 c.1305C>T p.T435T Chr.19: 21537358-21537358 on Build GRCh38
1559524 c.1473G>C p.K491N Chr.19: 21537526-21537526 on Build GRCh38
1271378 c.1571T>G p.L524R Chr.19: 21537624-21537624 on Build GRCh38
297495 c.1510A>G p.S504G Chr.19: 21537563-21537563 on Build GRCh38
5785201 c.1384T>A p.F462I Chr.19: 21537437-21537437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759041259 Chr.19: 21537556-21537556 on Build GRCh38
rs757051580 Chr.19: 21537671-21537671 on Build GRCh38
rs1038410915 Chr.19: 21537455-21537455 on Build GRCh38
rs369566652 Chr.19: 21537562-21537562 on Build GRCh38
rs762237526 Chr.19: 21537479-21537479 on Build GRCh38
rs878915708 Chr.19: 21537629-21537629 on Build GRCh38
rs373149644 Chr.19: 21537614-21537614 on Build GRCh38
rs768947209 Chr.19: 21537477-21537477 on Build GRCh38
rs375170285 Chr.19: 21537552-21537552 on Build GRCh38
rs766927375 Chr.19: 21537594-21537594 on Build GRCh38
rs562882253 Chr.19: 21537344-21537344 on Build GRCh38
rs924137188 Chr.19: 21537413-21537413 on Build GRCh38
rs878872120 Chr.19: 21537650-21537650 on Build GRCh38
rs749036539 Chr.19: 21537354-21537354 on Build GRCh38
rs750898131 Chr.19: 21537533-21537533 on Build GRCh38
rs764664671 Chr.19: 21537749-21537749 on Build GRCh38
rs752416908 Chr.19: 21537541-21537541 on Build GRCh38
rs775015529 Chr.19: 21537756-21537756 on Build GRCh38
rs767719674 Chr.19: 21537404-21537404 on Build GRCh38
rs202239129 Chr.19: 21537421-21537421 on Build GRCh38
rs142015117 Chr.19: 21537357-21537357 on Build GRCh38
rs968330199 Chr.19: 21537384-21537384 on Build GRCh38
rs1018715501 Chr.19: 21537331-21537331 on Build GRCh38
rs371230317 Chr.19: 21537715-21537715 on Build GRCh38
rs535932722 Chr.19: 21537530-21537530 on Build GRCh38
rs775564546 Chr.19: 21537408-21537408 on Build GRCh38
rs775647778 Chr.19: 21537466-21537466 on Build GRCh38
rs773133276 Chr.19: 21537452-21537452 on Build GRCh38
rs117327054 Chr.19: 21537387-21537387 on Build GRCh38
rs771900629 Chr.19: 21537444-21537444 on Build GRCh38
rs377473432 Chr.19: 21537622-21537622 on Build GRCh38
rs199679715 Chr.19: 21537633-21537633 on Build GRCh38
rs771237128 Chr.19: 21537721-21537721 on Build GRCh38
rs368617365 Chr.19: 21537363-21537363 on Build GRCh38
rs758054433 Chr.19: 21537635-21537635 on Build GRCh38
rs974555918 Chr.19: 21537731-21537731 on Build GRCh38
rs1022372341 Chr.19: 21537368-21537368 on Build GRCh38
rs768059874 Chr.19: 21537492-21537492 on Build GRCh38
rs752291954 Chr.19: 21537632-21537632 on Build GRCh38
rs139014529 Chr.19: 21537429-21537429 on Build GRCh38
rs745393389 Chr.19: 21537316-21537316 on Build GRCh38
rs772507718 Chr.19: 21537570-21537570 on Build GRCh38
rs553815757 Chr.19: 21537454-21537454 on Build GRCh38
rs377532182 Chr.19: 21537722-21537722 on Build GRCh38
rs772025620 Chr.19: 21537373-21537373 on Build GRCh38
rs753417701 Chr.19: 21537514-21537514 on Build GRCh38
rs866964569 Chr.19: 21537625-21537626 on Build GRCh38
rs755407332 Chr.19: 21537713-21537714 on Build GRCh38
rs762544907 Chr.19: 21537760-21537760 on Build GRCh38
rs763713810 Chr.19: 21537766-21537766 on Build GRCh38
rs879108675 Chr.19: 21537639-21537639 on Build GRCh38
rs533957276 Chr.19: 21537577-21537577 on Build GRCh38
rs777507211 Chr.19: 21537668-21537668 on Build GRCh38
rs878991601 Chr.19: 21537609-21537609 on Build GRCh38
rs898879672 Chr.19: 21537353-21537353 on Build GRCh38
rs759976729 Chr.19: 21537746-21537746 on Build GRCh38
rs566306821 Chr.19: 21537559-21537559 on Build GRCh38
rs762182153 Chr.19: 21537569-21537569 on Build GRCh38
rs552572579 Chr.19: 21537417-21537417 on Build GRCh38
rs756723506 Chr.19: 21537603-21537604 on Build GRCh38
rs773715090 Chr.19: 21537574-21537574 on Build GRCh38
rs142189246 Chr.19: 21537439-21537439 on Build GRCh38
rs759051894 Chr.19: 21537621-21537621 on Build GRCh38
rs145137585 Chr.19: 21537359-21537359 on Build GRCh38
rs779948744 Chr.19: 21537560-21537560 on Build GRCh38
rs761257020 Chr.19: 21537503-21537503 on Build GRCh38
rs774207624 Chr.19: 21537358-21537358 on Build GRCh38
rs755735749 Chr.19: 21537342-21537342 on Build GRCh38
rs878934750 Chr.19: 21537553-21537553 on Build GRCh38
rs776640182 Chr.19: 21537375-21537375 on Build GRCh38
rs1047651067 Chr.19: 21537499-21537499 on Build GRCh38
rs1008656197 Chr.19: 21537309-21537309 on Build GRCh38
rs769708406 Chr.19: 21537460-21537460 on Build GRCh38
rs112131328 Chr.19: 21537661-21537661 on Build GRCh38
rs201557397 Chr.19: 21537696-21537696 on Build GRCh38
rs752006309 Chr.19: 21537419-21537419 on Build GRCh38
rs577024194 Chr.19: 21537717-21537717 on Build GRCh38
rs201618245 Chr.19: 21537718-21537718 on Build GRCh38
rs750952857 Chr.19: 21537498-21537498 on Build GRCh38
rs746886661 Chr.19: 21537557-21537557 on Build GRCh38
rs777283083 Chr.19: 21537719-21537721 on Build GRCh38
rs771622608 Chr.19: 21537630-21537630 on Build GRCh38
rs200312692 Chr.19: 21537430-21537430 on Build GRCh38
rs749367373 Chr.19: 21537561-21537561 on Build GRCh38
rs763046874 Chr.19: 21537471-21537471 on Build GRCh38
rs373699970 Chr.19: 21537478-21537478 on Build GRCh38
rs190865119 Chr.19: 21537422-21537422 on Build GRCh38
rs879058881 Chr.19: 21537564-21537564 on Build GRCh38
rs879009602 Chr.19: 21537367-21537367 on Build GRCh38
rs547915928 Chr.19: 21537544-21537544 on Build GRCh38
rs527920357 Chr.19: 21537393-21537393 on Build GRCh38
rs930348038 Chr.19: 21537655-21537655 on Build GRCh38
rs746728924 Chr.19: 21537777-21537777 on Build GRCh38
rs867943559 Chr.19: 21537624-21537625 on Build GRCh38
rs757511247 Chr.19: 21537574-21537578 on Build GRCh38
rs778530389 Chr.19: 21537442-21537442 on Build GRCh38
rs754127680 Chr.19: 21537649-21537649 on Build GRCh38
rs781155156 Chr.19: 21537695-21537695 on Build GRCh38
rs748475506 Chr.19: 21537411-21537411 on Build GRCh38
rs964777859 Chr.19: 21537333-21537333 on Build GRCh38
rs778511480 Chr.19: 21537539-21537539 on Build GRCh38
rs765738759 Chr.19: 21537512-21537512 on Build GRCh38
rs747955672 Chr.19: 21537443-21537443 on Build GRCh38
rs898841953 Chr.19: 21537646-21537646 on Build GRCh38
rs558487453 Chr.19: 21537702-21537702 on Build GRCh38
rs774710034 Chr.19: 21537568-21537568 on Build GRCh38
rs751212978 Chr.19: 21537771-21537771 on Build GRCh38

View Full Product Details