Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 29563759-29564257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VSTM2B
Entrez Gene ID
342865
Gene Name
V-set and transmembrane domain containing 2B
Gene Aliases
-
UniGene
Hs.451618
Gene Chromosome Location
Chr.19: 29525431-29564555 on Build GRCh38
Transcripts
NM_001146339.1 XM_005258856.3 XM_011526902.2 XM_011526903.2 XM_011526904.2 XM_011526905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5449241 c.830A>G p.K277R Chr.19: 29563906-29563906 on Build GRCh38
219540 c.815T>A p.L272Q Chr.19: 29563891-29563891 on Build GRCh38
439169 c.821C>T p.A274V Chr.19: 29563897-29563897 on Build GRCh38
439168 c.821C>A p.A274D Chr.19: 29563897-29563897 on Build GRCh38
1480892 c.775G>A p.G259S Chr.19: 29563851-29563851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577104343 Chr.19: 29564035-29564035 on Build GRCh38
rs989118731 Chr.19: 29563889-29563889 on Build GRCh38
rs889987507 Chr.19: 29563972-29563972 on Build GRCh38
rs1036290014 Chr.19: 29563897-29563897 on Build GRCh38
rs1048876982 Chr.19: 29563785-29563785 on Build GRCh38
rs998336221 Chr.19: 29563849-29563849 on Build GRCh38
rs185075864 Chr.19: 29563950-29563950 on Build GRCh38
rs779679626 Chr.19: 29564133-29564133 on Build GRCh38
rs34448092 Chr.19: 29564079-29564080 on Build GRCh38
rs761971447 Chr.19: 29563915-29563915 on Build GRCh38
rs142289093 Chr.19: 29563951-29563951 on Build GRCh38
rs12608461 Chr.19: 29563839-29563839 on Build GRCh38
rs369332650 Chr.19: 29563920-29563920 on Build GRCh38
rs928554350 Chr.19: 29564097-29564097 on Build GRCh38
rs971307935 Chr.19: 29564005-29564005 on Build GRCh38
rs768746781 Chr.19: 29564070-29564070 on Build GRCh38
rs1033086107 Chr.19: 29563872-29563872 on Build GRCh38
rs746859863 Chr.19: 29563850-29563850 on Build GRCh38
rs1009920384 Chr.19: 29564001-29564001 on Build GRCh38
rs1055460245 Chr.19: 29564175-29564175 on Build GRCh38
rs557741525 Chr.19: 29563982-29563982 on Build GRCh38
rs745737020 Chr.19: 29563892-29563892 on Build GRCh38
rs770590925 Chr.19: 29563854-29563854 on Build GRCh38
rs577636574 Chr.19: 29563990-29563990 on Build GRCh38
rs968266802 Chr.19: 29563823-29563823 on Build GRCh38
rs374732056 Chr.19: 29563790-29563790 on Build GRCh38
rs536481826 Chr.19: 29563931-29563931 on Build GRCh38
rs143894829 Chr.19: 29564019-29564020 on Build GRCh38
rs113762882 Chr.19: 29563838-29563838 on Build GRCh38
rs999832441 Chr.19: 29563855-29563855 on Build GRCh38
rs140018368 Chr.19: 29563907-29563907 on Build GRCh38
rs966084246 Chr.19: 29563844-29563844 on Build GRCh38
rs765728256 Chr.19: 29564156-29564156 on Build GRCh38
rs760704830 Chr.19: 29564014-29564014 on Build GRCh38
rs934237859 Chr.19: 29564231-29564231 on Build GRCh38
rs200249897 Chr.19: 29564020-29564020 on Build GRCh38
rs984591487 Chr.19: 29564055-29564055 on Build GRCh38
rs917200154 Chr.19: 29563891-29563891 on Build GRCh38
rs113111915 Chr.19: 29563966-29563966 on Build GRCh38
rs769670032 Chr.19: 29563878-29563878 on Build GRCh38
rs909870985 Chr.19: 29563927-29563927 on Build GRCh38
rs543520569 Chr.19: 29563991-29563991 on Build GRCh38
rs760099703 Chr.19: 29563953-29563953 on Build GRCh38
rs764874446 Chr.19: 29563864-29563864 on Build GRCh38
rs189533173 Chr.19: 29564016-29564016 on Build GRCh38
rs146590547 Chr.19: 29563822-29563822 on Build GRCh38
rs563274202 Chr.19: 29563992-29563992 on Build GRCh38
rs922817577 Chr.19: 29564165-29564165 on Build GRCh38
rs984615846 Chr.19: 29563875-29563875 on Build GRCh38
rs35216506 Chr.19: 29564024-29564025 on Build GRCh38
rs376915726 Chr.19: 29564120-29564120 on Build GRCh38
rs971954082 Chr.19: 29564022-29564022 on Build GRCh38
rs928262122 Chr.19: 29563903-29563903 on Build GRCh38
rs1024206186 Chr.19: 29564002-29564002 on Build GRCh38
rs757872446 Chr.19: 29563977-29563977 on Build GRCh38
rs951950336 Chr.19: 29564053-29564053 on Build GRCh38
rs953813690 Chr.19: 29563851-29563851 on Build GRCh38
rs953441213 Chr.19: 29563981-29563981 on Build GRCh38
rs775710984 Chr.19: 29563979-29563979 on Build GRCh38
rs796650457 Chr.19: 29563787-29563787 on Build GRCh38
rs997514243 Chr.19: 29563970-29563970 on Build GRCh38
rs1049893013 Chr.19: 29563967-29563967 on Build GRCh38
rs776618532 Chr.19: 29564118-29564118 on Build GRCh38
rs972726055 Chr.19: 29564154-29564154 on Build GRCh38
rs909013681 Chr.19: 29564018-29564018 on Build GRCh38
rs772475293 Chr.19: 29563919-29563919 on Build GRCh38
rs567862790 Chr.19: 29563914-29563914 on Build GRCh38
rs1011318917 Chr.19: 29563921-29563921 on Build GRCh38
rs765716574 Chr.19: 29563956-29563956 on Build GRCh38

View Full Product Details