Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40502688-40503190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPTBN4
Entrez Gene ID
57731
Gene Name
spectrin beta, non-erythrocytic 4
Gene Aliases
QV, SPNB4, SPTBN3
UniGene
Hs.32706
Gene Chromosome Location
Chr.19: 40467153-40576459 on Build GRCh38
Transcripts
NM_020971.2 XM_011527173.2 XM_017027049.1 XM_017027050.1 XM_017027051.1 XM_017027052.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1681038 c.1205C>T p.A402V Chr.19: 40502776-40502776 on Build GRCh38
5962641 c.1360C>T p.Q454* Chr.19: 40502931-40502931 on Build GRCh38
5962642 c.1360C>T p.Q454* Chr.19: 40502931-40502931 on Build GRCh38
7334773 c.1281G>A p.L427L Chr.19: 40502852-40502852 on Build GRCh38
3225706 c.1271_1273delAGG p.E425delE Chr.19: 40502842-40502844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953054307 Chr.19: 40502835-40502835 on Build GRCh38
rs1053673518 Chr.19: 40503015-40503015 on Build GRCh38
rs150218674 Chr.19: 40503146-40503146 on Build GRCh38
rs772250506 Chr.19: 40502959-40502959 on Build GRCh38
rs769834825 Chr.19: 40502837-40502837 on Build GRCh38
rs371451301 Chr.19: 40502978-40502978 on Build GRCh38
rs758575831 Chr.19: 40502940-40502943 on Build GRCh38
rs1028614347 Chr.19: 40503131-40503131 on Build GRCh38
rs192069978 Chr.19: 40502970-40502970 on Build GRCh38
rs1007193263 Chr.19: 40503151-40503151 on Build GRCh38
rs771137688 Chr.19: 40502824-40502824 on Build GRCh38
rs377417831 Chr.19: 40502740-40502740 on Build GRCh38
rs998830173 Chr.19: 40503126-40503126 on Build GRCh38
rs771209008 Chr.19: 40502849-40502849 on Build GRCh38
rs985871222 Chr.19: 40502891-40502891 on Build GRCh38
rs774474846 Chr.19: 40502944-40502944 on Build GRCh38
rs749289092 Chr.19: 40502845-40502845 on Build GRCh38
rs562924157 Chr.19: 40502947-40502947 on Build GRCh38
rs373069703 Chr.19: 40502943-40502943 on Build GRCh38
rs764543902 Chr.19: 40502883-40502883 on Build GRCh38
rs202136395 Chr.19: 40502763-40502763 on Build GRCh38
rs778441932 Chr.19: 40502780-40502780 on Build GRCh38
rs751233619 Chr.19: 40502898-40502898 on Build GRCh38
rs867399194 Chr.19: 40502744-40502744 on Build GRCh38
rs749968648 Chr.19: 40502889-40502889 on Build GRCh38
rs76311350 Chr.19: 40502971-40502971 on Build GRCh38
rs749401941 Chr.19: 40502942-40502942 on Build GRCh38
rs529442079 Chr.19: 40502756-40502756 on Build GRCh38
rs1012939662 Chr.19: 40502713-40502713 on Build GRCh38
rs753269133 Chr.19: 40502777-40502777 on Build GRCh38
rs768265091 Chr.19: 40502811-40502811 on Build GRCh38
rs901363396 Chr.19: 40503116-40503116 on Build GRCh38
rs754667625 Chr.19: 40502904-40502904 on Build GRCh38
rs757787296 Chr.19: 40502894-40502894 on Build GRCh38
rs545646185 Chr.19: 40502977-40502977 on Build GRCh38
rs934361075 Chr.19: 40503099-40503099 on Build GRCh38
rs756154554 Chr.19: 40502920-40502920 on Build GRCh38
rs934775198 Chr.19: 40503003-40503003 on Build GRCh38
rs754722828 Chr.19: 40502979-40502979 on Build GRCh38
rs978446439 Chr.19: 40503033-40503033 on Build GRCh38
rs766055917 Chr.19: 40502896-40502896 on Build GRCh38
rs767240413 Chr.19: 40502753-40502753 on Build GRCh38
rs750318017 Chr.19: 40502778-40502778 on Build GRCh38
rs370028770 Chr.19: 40502881-40502881 on Build GRCh38
rs758098441 Chr.19: 40502792-40502792 on Build GRCh38
rs770069120 Chr.19: 40502736-40502736 on Build GRCh38
rs775809176 Chr.19: 40502875-40502875 on Build GRCh38
rs111542586 Chr.19: 40503041-40503041 on Build GRCh38
rs781133594 Chr.19: 40502820-40502820 on Build GRCh38
rs773469412 Chr.19: 40502839-40502839 on Build GRCh38
rs748159055 Chr.19: 40502823-40502823 on Build GRCh38
rs1056793023 Chr.19: 40503106-40503106 on Build GRCh38
rs760534763 Chr.19: 40502919-40502919 on Build GRCh38
rs767926330 Chr.19: 40502870-40502870 on Build GRCh38
rs763287088 Chr.19: 40502742-40502742 on Build GRCh38
rs1029982884 Chr.19: 40502825-40502825 on Build GRCh38
rs112431529 Chr.19: 40502962-40502962 on Build GRCh38
rs749867676 Chr.19: 40502781-40502781 on Build GRCh38
rs768026382 Chr.19: 40502766-40502766 on Build GRCh38
rs762374654 Chr.19: 40502976-40502976 on Build GRCh38
rs910361580 Chr.19: 40502995-40502995 on Build GRCh38
rs879030239 Chr.19: 40502981-40502981 on Build GRCh38
rs760976338 Chr.19: 40502967-40502967 on Build GRCh38
rs1031846521 Chr.19: 40502752-40502752 on Build GRCh38
rs774379566 Chr.19: 40502859-40502859 on Build GRCh38
rs151234878 Chr.19: 40502809-40502809 on Build GRCh38
rs542892567 Chr.19: 40502729-40502729 on Build GRCh38
rs917519557 Chr.19: 40503144-40503144 on Build GRCh38
rs781184989 Chr.19: 40502918-40502918 on Build GRCh38
rs779521000 Chr.19: 40502808-40502808 on Build GRCh38
rs999387822 Chr.19: 40502743-40502743 on Build GRCh38
rs759950953 Chr.19: 40502869-40502869 on Build GRCh38

View Full Product Details