Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41848714-41849209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTC2
Entrez Gene ID
63946
Gene Name
DMRT like family C2
Gene Aliases
-
UniGene
Hs.350507
Gene Chromosome Location
Chr.19: 41844743-41852330 on Build GRCh38
Transcripts
NM_001040283.2 XM_011527214.1 XM_011527215.1 XM_011527216.2 XM_017027123.1 XM_017027124.1 XM_017027125.1 XM_017027126.1 XM_017027127.1 XM_017027128.1 XM_017027129.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4903081 c.694C>T p.H232Y Chr.19: 41849195-41849195 on Build GRCh38
3534530 c.688G>A p.G230R Chr.19: 41849189-41849189 on Build GRCh38
6465787 c.460C>T p.P154S Chr.19: 41848807-41848807 on Build GRCh38
5556316 c.477T>C p.H159H Chr.19: 41848824-41848824 on Build GRCh38
3534529 c.654G>A p.Q218Q Chr.19: 41849155-41849155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17853910 Chr.19: 41848898-41848898 on Build GRCh38
rs375271526 Chr.19: 41849145-41849145 on Build GRCh38
rs558894286 Chr.19: 41848920-41848920 on Build GRCh38
rs1055427732 Chr.19: 41849078-41849078 on Build GRCh38
rs967587177 Chr.19: 41848809-41848809 on Build GRCh38
rs782145630 Chr.19: 41848827-41848827 on Build GRCh38
rs782506420 Chr.19: 41848841-41848841 on Build GRCh38
rs782807415 Chr.19: 41848829-41848829 on Build GRCh38
rs370925916 Chr.19: 41848835-41848835 on Build GRCh38
rs183622708 Chr.19: 41849080-41849080 on Build GRCh38
rs782052285 Chr.19: 41848938-41848938 on Build GRCh38
rs782134710 Chr.19: 41849099-41849099 on Build GRCh38
rs371216175 Chr.19: 41849115-41849115 on Build GRCh38
rs782496035 Chr.19: 41848972-41848972 on Build GRCh38
rs782684669 Chr.19: 41848792-41848792 on Build GRCh38
rs189285681 Chr.19: 41849082-41849082 on Build GRCh38
rs782349470 Chr.19: 41849114-41849114 on Build GRCh38
rs371621401 Chr.19: 41848930-41848930 on Build GRCh38
rs1008438100 Chr.19: 41849152-41849152 on Build GRCh38
rs371867212 Chr.19: 41849122-41849122 on Build GRCh38
rs1005010048 Chr.19: 41848737-41848737 on Build GRCh38
rs782747662 Chr.19: 41849170-41849170 on Build GRCh38
rs1034591205 Chr.19: 41848765-41848765 on Build GRCh38
rs782022689 Chr.19: 41848949-41848949 on Build GRCh38
rs374220011 Chr.19: 41848887-41848887 on Build GRCh38
rs782237730 Chr.19: 41849044-41849044 on Build GRCh38
rs113061132 Chr.19: 41848749-41848749 on Build GRCh38
rs782224428 Chr.19: 41848825-41848825 on Build GRCh38
rs782205084 Chr.19: 41848785-41848785 on Build GRCh38
rs782048862 Chr.19: 41848750-41848750 on Build GRCh38
rs782543443 Chr.19: 41848786-41848786 on Build GRCh38
rs782123198 Chr.19: 41848830-41848830 on Build GRCh38
rs201302617 Chr.19: 41848990-41848990 on Build GRCh38
rs782724452 Chr.19: 41849138-41849138 on Build GRCh38
rs782072349 Chr.19: 41848838-41848838 on Build GRCh38
rs1024085506 Chr.19: 41848752-41848752 on Build GRCh38
rs940559212 Chr.19: 41848961-41848961 on Build GRCh38
rs200990664 Chr.19: 41848989-41848989 on Build GRCh38
rs782208407 Chr.19: 41848984-41848984 on Build GRCh38
rs782282221 Chr.19: 41849021-41849021 on Build GRCh38
rs782464529 Chr.19: 41849187-41849187 on Build GRCh38
rs374622487 Chr.19: 41848951-41848951 on Build GRCh38
rs769520473 Chr.19: 41848759-41848759 on Build GRCh38
rs148724244 Chr.19: 41848931-41848931 on Build GRCh38
rs781956714 Chr.19: 41848816-41848816 on Build GRCh38
rs782496359 Chr.19: 41848781-41848781 on Build GRCh38
rs151277709 Chr.19: 41848954-41848954 on Build GRCh38
rs781863181 Chr.19: 41848773-41848773 on Build GRCh38
rs145708066 Chr.19: 41848856-41848856 on Build GRCh38
rs906983346 Chr.19: 41848997-41848997 on Build GRCh38
rs907686497 Chr.19: 41848900-41848900 on Build GRCh38
rs200152667 Chr.19: 41848980-41848980 on Build GRCh38
rs145179597 Chr.19: 41848883-41848883 on Build GRCh38
rs367552578 Chr.19: 41848968-41848968 on Build GRCh38
rs782743876 Chr.19: 41848834-41848834 on Build GRCh38
rs782580801 Chr.19: 41849018-41849018 on Build GRCh38
rs369439503 Chr.19: 41848756-41848756 on Build GRCh38
rs782049925 Chr.19: 41849121-41849121 on Build GRCh38
rs782769308 Chr.19: 41848762-41848762 on Build GRCh38
rs373659217 Chr.19: 41848804-41848804 on Build GRCh38
rs782495210 Chr.19: 41848878-41848878 on Build GRCh38
rs964013106 Chr.19: 41848853-41848853 on Build GRCh38
rs781822589 Chr.19: 41848857-41848857 on Build GRCh38
rs374714026 Chr.19: 41849188-41849188 on Build GRCh38
rs781866567 Chr.19: 41849179-41849179 on Build GRCh38
rs570158158 Chr.19: 41848839-41848839 on Build GRCh38
rs199781236 Chr.19: 41848782-41848782 on Build GRCh38
rs377724016 Chr.19: 41848923-41848923 on Build GRCh38
rs957572828 Chr.19: 41848772-41848772 on Build GRCh38
rs782329023 Chr.19: 41848948-41848948 on Build GRCh38
rs782385572 Chr.19: 41848805-41848805 on Build GRCh38
rs782677269 Chr.19: 41848896-41848896 on Build GRCh38
rs535043320 Chr.19: 41849003-41849003 on Build GRCh38
rs782159590 Chr.19: 41849166-41849166 on Build GRCh38
rs782405247 Chr.19: 41848894-41848894 on Build GRCh38
rs200511054 Chr.19: 41848851-41848851 on Build GRCh38
rs782088416 Chr.19: 41848958-41848958 on Build GRCh38

View Full Product Details