Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47309658-47310083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5AR1
Entrez Gene ID
728
Gene Name
complement component 5a receptor 1
Gene Aliases
C5A, C5AR, C5R1, CD88
UniGene
Hs.2161
Gene Chromosome Location
Chr.19: 47309847-47322070 on Build GRCh38
Transcripts
NM_001736.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7220918 c.3+10G>A p.? Chr.19: 47309908-47309908 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752831998 Chr.19: 47309925-47309925 on Build GRCh38
rs995223858 Chr.19: 47309988-47309988 on Build GRCh38
rs573620022 Chr.19: 47310012-47310012 on Build GRCh38
rs768000180 Chr.19: 47309780-47309780 on Build GRCh38
rs571703605 Chr.19: 47309994-47309994 on Build GRCh38
rs953806501 Chr.19: 47309990-47309990 on Build GRCh38
rs567175577 Chr.19: 47309824-47309824 on Build GRCh38
rs1027051256 Chr.19: 47309989-47309989 on Build GRCh38
rs112996569 Chr.19: 47309873-47309873 on Build GRCh38
rs1036956696 Chr.19: 47309978-47309978 on Build GRCh38
rs1002756705 Chr.19: 47309735-47309735 on Build GRCh38
rs755062184 Chr.19: 47309858-47309858 on Build GRCh38
rs967716653 Chr.19: 47310029-47310029 on Build GRCh38
rs751215819 Chr.19: 47310015-47310015 on Build GRCh38
rs986324289 Chr.19: 47309843-47309843 on Build GRCh38
rs760264163 Chr.19: 47309966-47309966 on Build GRCh38
rs897057701 Chr.19: 47309983-47309983 on Build GRCh38
rs11670330 Chr.19: 47309908-47309908 on Build GRCh38
rs201184675 Chr.19: 47309903-47309903 on Build GRCh38
rs907060791 Chr.19: 47309965-47309965 on Build GRCh38
rs764385430 Chr.19: 47309939-47309939 on Build GRCh38
rs767688569 Chr.19: 47309860-47309860 on Build GRCh38
rs762920299 Chr.19: 47309911-47309911 on Build GRCh38
rs533745541 Chr.19: 47309996-47309996 on Build GRCh38
rs558486417 Chr.19: 47309764-47309764 on Build GRCh38
rs192858378 Chr.19: 47309811-47309811 on Build GRCh38
rs948792275 Chr.19: 47309909-47309909 on Build GRCh38
rs188839786 Chr.19: 47309776-47309776 on Build GRCh38
rs750015646 Chr.19: 47309923-47309923 on Build GRCh38
rs944828116 Chr.19: 47309863-47309863 on Build GRCh38
rs1022735428 Chr.19: 47309681-47309681 on Build GRCh38
rs1034757680 Chr.19: 47309684-47309684 on Build GRCh38
rs531451097 Chr.19: 47309707-47309707 on Build GRCh38
rs942865543 Chr.19: 47309800-47309800 on Build GRCh38
rs761464655 Chr.19: 47309864-47309865 on Build GRCh38
rs564762664 Chr.19: 47309733-47309733 on Build GRCh38
rs201013609 Chr.19: 47309798-47309798 on Build GRCh38
rs201549996 Chr.19: 47309862-47309862 on Build GRCh38
rs558018011 Chr.19: 47309991-47309991 on Build GRCh38
rs535799975 Chr.19: 47309874-47309874 on Build GRCh38
rs1041640880 Chr.19: 47309821-47309821 on Build GRCh38
rs754096914 Chr.19: 47309941-47309941 on Build GRCh38
rs1023526927 Chr.19: 47309974-47309974 on Build GRCh38
rs576111516 Chr.19: 47309848-47309848 on Build GRCh38
rs756290197 Chr.19: 47309933-47309933 on Build GRCh38
rs924927354 Chr.19: 47310029-47310030 on Build GRCh38
rs956469588 Chr.19: 47310031-47310031 on Build GRCh38
rs748383249 Chr.19: 47309879-47309879 on Build GRCh38
rs749732136 Chr.19: 47309867-47309867 on Build GRCh38
rs200157279 Chr.19: 47309876-47309876 on Build GRCh38
rs542479411 Chr.19: 47310035-47310035 on Build GRCh38
rs201922905 Chr.19: 47309874-47309874 on Build GRCh38
rs201019134 Chr.19: 47309884-47309884 on Build GRCh38
rs776284801 Chr.19: 47309887-47309887 on Build GRCh38
rs751378760 Chr.19: 47309859-47309859 on Build GRCh38
rs564969382 Chr.19: 47309736-47309736 on Build GRCh38
rs369189796 Chr.19: 47310023-47310023 on Build GRCh38
rs200043362 Chr.19: 47309866-47309866 on Build GRCh38
rs946374498 Chr.19: 47309768-47309768 on Build GRCh38
rs956471534 Chr.19: 47309687-47309687 on Build GRCh38
rs1019351425 Chr.19: 47309998-47310001 on Build GRCh38
rs765024012 Chr.19: 47309907-47309907 on Build GRCh38
rs767563852 Chr.19: 47309916-47309916 on Build GRCh38
rs183541639 Chr.19: 47310003-47310003 on Build GRCh38

View Full Product Details