Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48404734-48405227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN2D
Entrez Gene ID
2906
Gene Name
glutamate ionotropic receptor NMDA type subunit 2D
Gene Aliases
EB11, GluN2D, NMDAR2D, NR2D
UniGene
Hs.445015
Gene Chromosome Location
Chr.19: 48394875-48444937 on Build GRCh38
Transcripts
NM_000836.2 XM_011526872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4079843 c.567C>T p.A189A Chr.19: 48404835-48404835 on Build GRCh38
5662779 c.851G>T p.G284V Chr.19: 48405119-48405119 on Build GRCh38
4079845 c.715G>A p.V239I Chr.19: 48404983-48404983 on Build GRCh38
338221 c.945C>G p.D315E Chr.19: 48405213-48405213 on Build GRCh38
6675589 c.555G>A p.T185T Chr.19: 48404823-48404823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751224832 Chr.19: 48404907-48404907 on Build GRCh38
rs77992710 Chr.19: 48404991-48404991 on Build GRCh38
rs939536813 Chr.19: 48404765-48404765 on Build GRCh38
rs771302827 Chr.19: 48405186-48405186 on Build GRCh38
rs773682521 Chr.19: 48404989-48404989 on Build GRCh38
rs775352797 Chr.19: 48405112-48405112 on Build GRCh38
rs763994704 Chr.19: 48405129-48405129 on Build GRCh38
rs771300277 Chr.19: 48405098-48405098 on Build GRCh38
rs373524397 Chr.19: 48405205-48405205 on Build GRCh38
rs764741925 Chr.19: 48404774-48404774 on Build GRCh38
rs1045477025 Chr.19: 48405169-48405169 on Build GRCh38
rs778228290 Chr.19: 48405170-48405170 on Build GRCh38
rs773027827 Chr.19: 48405206-48405206 on Build GRCh38
rs756276428 Chr.19: 48405165-48405165 on Build GRCh38
rs756065208 Chr.19: 48404856-48404856 on Build GRCh38
rs769069826 Chr.19: 48404875-48404875 on Build GRCh38
rs77950614 Chr.19: 48404823-48404823 on Build GRCh38
rs565752293 Chr.19: 48405079-48405079 on Build GRCh38
rs576321942 Chr.19: 48405155-48405155 on Build GRCh38
rs111439081 Chr.19: 48404955-48404955 on Build GRCh38
rs772025904 Chr.19: 48404864-48404864 on Build GRCh38
rs778049759 Chr.19: 48405071-48405071 on Build GRCh38
rs369747884 Chr.19: 48405195-48405195 on Build GRCh38
rs752378424 Chr.19: 48405009-48405009 on Build GRCh38
rs752929902 Chr.19: 48405159-48405159 on Build GRCh38
rs939569697 Chr.19: 48405141-48405141 on Build GRCh38
rs757814136 Chr.19: 48404800-48404800 on Build GRCh38
rs373401802 Chr.19: 48404934-48404934 on Build GRCh38
rs200407469 Chr.19: 48404988-48404988 on Build GRCh38
rs781449463 Chr.19: 48405041-48405041 on Build GRCh38
rs888091926 Chr.19: 48404836-48404836 on Build GRCh38
rs768712802 Chr.19: 48404959-48404959 on Build GRCh38
rs878903342 Chr.19: 48405168-48405168 on Build GRCh38
rs753527945 Chr.19: 48405139-48405139 on Build GRCh38
rs749820198 Chr.19: 48405088-48405088 on Build GRCh38
rs754526189 Chr.19: 48404929-48404929 on Build GRCh38
rs778297666 Chr.19: 48404846-48404846 on Build GRCh38
rs555640307 Chr.19: 48405194-48405194 on Build GRCh38
rs77698773 Chr.19: 48404896-48404896 on Build GRCh38
rs755944791 Chr.19: 48404943-48404943 on Build GRCh38
rs182928265 Chr.19: 48405028-48405028 on Build GRCh38
rs763302470 Chr.19: 48404979-48404979 on Build GRCh38
rs185295623 Chr.19: 48405122-48405122 on Build GRCh38
rs770148910 Chr.19: 48405063-48405063 on Build GRCh38
rs374160642 Chr.19: 48405006-48405006 on Build GRCh38
rs35658494 Chr.19: 48404826-48404827 on Build GRCh38
rs762242145 Chr.19: 48404772-48404772 on Build GRCh38
rs760607707 Chr.19: 48405010-48405010 on Build GRCh38
rs780780422 Chr.19: 48404831-48404831 on Build GRCh38
rs753823477 Chr.19: 48405016-48405016 on Build GRCh38
rs779162967 Chr.19: 48404860-48404860 on Build GRCh38
rs766102954 Chr.19: 48404811-48404811 on Build GRCh38
rs971881000 Chr.19: 48405119-48405119 on Build GRCh38
rs1028361296 Chr.19: 48404966-48404966 on Build GRCh38
rs976585979 Chr.19: 48404952-48404952 on Build GRCh38
rs773773819 Chr.19: 48404978-48404978 on Build GRCh38
rs748558946 Chr.19: 48404973-48404973 on Build GRCh38
rs763980279 Chr.19: 48405015-48405015 on Build GRCh38
rs765801932 Chr.19: 48404903-48404903 on Build GRCh38
rs999035598 Chr.19: 48404832-48404832 on Build GRCh38
rs762332086 Chr.19: 48404884-48404884 on Build GRCh38
rs749172584 Chr.19: 48404946-48404946 on Build GRCh38
rs758239148 Chr.19: 48405013-48405013 on Build GRCh38
rs752498935 Chr.19: 48404941-48404941 on Build GRCh38
rs745987713 Chr.19: 48404862-48404862 on Build GRCh38
rs775844136 Chr.19: 48404865-48404865 on Build GRCh38
rs746376633 Chr.19: 48405191-48405191 on Build GRCh38
rs747192073 Chr.19: 48404871-48404871 on Build GRCh38
rs201492347 Chr.19: 48405153-48405153 on Build GRCh38
rs77520296 Chr.19: 48404830-48404830 on Build GRCh38
rs771716322 Chr.19: 48405193-48405193 on Build GRCh38
rs147811242 Chr.19: 48404787-48404787 on Build GRCh38
rs776924895 Chr.19: 48404878-48404878 on Build GRCh38
rs1053329857 Chr.19: 48404877-48404877 on Build GRCh38
rs563560262 Chr.19: 48404887-48404887 on Build GRCh38
rs765018291 Chr.19: 48405055-48405056 on Build GRCh38
rs777012990 Chr.19: 48404967-48404967 on Build GRCh38
rs761842024 Chr.19: 48405140-48405140 on Build GRCh38
rs201028602 Chr.19: 48404983-48404983 on Build GRCh38
rs755027413 Chr.19: 48405030-48405030 on Build GRCh38
rs765129833 Chr.19: 48405145-48405145 on Build GRCh38
rs771698080 Chr.19: 48405108-48405108 on Build GRCh38
rs531074073 Chr.19: 48404949-48404949 on Build GRCh38
rs768384656 Chr.19: 48405197-48405197 on Build GRCh38
rs776318286 Chr.19: 48405201-48405201 on Build GRCh38
rs748364628 Chr.19: 48405060-48405060 on Build GRCh38
rs190128029 Chr.19: 48405174-48405174 on Build GRCh38
rs201727209 Chr.19: 48404958-48404958 on Build GRCh38
rs377156868 Chr.19: 48405099-48405099 on Build GRCh38
rs994312324 Chr.19: 48404791-48404791 on Build GRCh38
rs746751166 Chr.19: 48405190-48405190 on Build GRCh38
rs757085096 Chr.19: 48405027-48405027 on Build GRCh38
rs759019629 Chr.19: 48405106-48405106 on Build GRCh38
rs750359083 Chr.19: 48405152-48405152 on Build GRCh38

View Full Product Details