Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55303361-55303859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRSK1
Entrez Gene ID
84446
Gene Name
BR serine/threonine kinase 1
Gene Aliases
hSAD1
UniGene
Hs.182081
Gene Chromosome Location
Chr.19: 55284101-55312562 on Build GRCh38
Transcripts
NM_032430.1 XM_005259327.3 XM_011527395.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1565005 c.1104C>A p.D368E Chr.19: 55303386-55303386 on Build GRCh38
4462313 c.1240C>T p.P414S Chr.19: 55303780-55303780 on Build GRCh38
5856801 c.1123G>A p.V375I Chr.19: 55303405-55303405 on Build GRCh38
6195822 c.1084C>G p.P362A Chr.19: 55303366-55303366 on Build GRCh38
4462233 c.1238C>T p.S413F Chr.19: 55303778-55303778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369182313 Chr.19: 55303622-55303622 on Build GRCh38
rs965104588 Chr.19: 55303536-55303536 on Build GRCh38
rs371708220 Chr.19: 55303710-55303710 on Build GRCh38
rs758425269 Chr.19: 55303712-55303712 on Build GRCh38
rs536618174 Chr.19: 55303796-55303796 on Build GRCh38
rs1551562 Chr.19: 55303513-55303513 on Build GRCh38
rs375730605 Chr.19: 55303764-55303764 on Build GRCh38
rs780991388 Chr.19: 55303723-55303723 on Build GRCh38
rs759115649 Chr.19: 55303793-55303793 on Build GRCh38
rs998433573 Chr.19: 55303563-55303563 on Build GRCh38
rs769784818 Chr.19: 55303620-55303620 on Build GRCh38
rs776766095 Chr.19: 55303759-55303759 on Build GRCh38
rs1009474954 Chr.19: 55303514-55303514 on Build GRCh38
rs781137898 Chr.19: 55303667-55303668 on Build GRCh38
rs764525325 Chr.19: 55303635-55303635 on Build GRCh38
rs763673212 Chr.19: 55303833-55303833 on Build GRCh38
rs200027140 Chr.19: 55303718-55303718 on Build GRCh38
rs774823175 Chr.19: 55303650-55303650 on Build GRCh38
rs915430218 Chr.19: 55303707-55303707 on Build GRCh38
rs45579439 Chr.19: 55303467-55303467 on Build GRCh38
rs976551855 Chr.19: 55303550-55303550 on Build GRCh38
rs866352526 Chr.19: 55303561-55303561 on Build GRCh38
rs780118374 Chr.19: 55303758-55303758 on Build GRCh38
rs17851414 Chr.19: 55303704-55303704 on Build GRCh38
rs1046521590 Chr.19: 55303769-55303769 on Build GRCh38
rs772349063 Chr.19: 55303766-55303766 on Build GRCh38
rs752386495 Chr.19: 55303801-55303801 on Build GRCh38
rs777923423 Chr.19: 55303716-55303716 on Build GRCh38
rs758373535 Chr.19: 55303443-55303443 on Build GRCh38
rs904908249 Chr.19: 55303458-55303458 on Build GRCh38
rs1048225160 Chr.19: 55303469-55303469 on Build GRCh38
rs769988526 Chr.19: 55303749-55303749 on Build GRCh38
rs1028538771 Chr.19: 55303671-55303671 on Build GRCh38
rs867316647 Chr.19: 55303670-55303670 on Build GRCh38
rs774882287 Chr.19: 55303763-55303763 on Build GRCh38
rs923357522 Chr.19: 55303554-55303554 on Build GRCh38
rs775596814 Chr.19: 55303625-55303625 on Build GRCh38
rs529205087 Chr.19: 55303394-55303394 on Build GRCh38
rs965226982 Chr.19: 55303542-55303542 on Build GRCh38
rs17851415 Chr.19: 55303791-55303791 on Build GRCh38
rs753528403 Chr.19: 55303397-55303397 on Build GRCh38
rs769989334 Chr.19: 55303764-55303765 on Build GRCh38
rs762590418 Chr.19: 55303806-55303806 on Build GRCh38
rs1020823727 Chr.19: 55303522-55303522 on Build GRCh38
rs571494926 Chr.19: 55303837-55303837 on Build GRCh38
rs1015659533 Chr.19: 55303518-55303518 on Build GRCh38
rs754489426 Chr.19: 55303404-55303404 on Build GRCh38
rs778467515 Chr.19: 55303436-55303436 on Build GRCh38
rs776757540 Chr.19: 55303617-55303617 on Build GRCh38
rs113255485 Chr.19: 55303409-55303409 on Build GRCh38
rs748120335 Chr.19: 55303438-55303438 on Build GRCh38
rs757051523 Chr.19: 55303720-55303720 on Build GRCh38
rs777715126 Chr.19: 55303445-55303445 on Build GRCh38
rs763450556 Chr.19: 55303631-55303631 on Build GRCh38
rs753486933 Chr.19: 55303705-55303705 on Build GRCh38
rs113879893 Chr.19: 55303410-55303410 on Build GRCh38
rs574981960 Chr.19: 55303473-55303473 on Build GRCh38
rs746043904 Chr.19: 55303668-55303668 on Build GRCh38
rs746105345 Chr.19: 55303730-55303730 on Build GRCh38
rs377142646 Chr.19: 55303454-55303454 on Build GRCh38
rs762234414 Chr.19: 55303683-55303683 on Build GRCh38
rs45508100 Chr.19: 55303447-55303447 on Build GRCh38
rs1032006377 Chr.19: 55303470-55303470 on Build GRCh38
rs746078555 Chr.19: 55303619-55303619 on Build GRCh38

View Full Product Details