Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56159862-56160353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF444
Entrez Gene ID
55311
Gene Name
zinc finger protein 444
Gene Aliases
EZF-2, EZF2, ZSCAN17
UniGene
Hs.24545
Gene Chromosome Location
Chr.19: 56140951-56160893 on Build GRCh38
Transcripts
NM_001253792.1 NM_018337.3 XM_005259034.2 XM_005259035.1 XM_005259036.1 XM_011527068.1 XM_011527069.1 XM_011527070.1 XM_011527071.1 XM_011527073.1 XM_017026932.1 XM_017026933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4653220 c.812C>T p.T271M Chr.19: 56160029-56160029 on Build GRCh38
230395 c.964C>T p.P322S Chr.19: 56160181-56160181 on Build GRCh38
4650525 c.893G>A p.R298H Chr.19: 56160110-56160110 on Build GRCh38
1721071 c.687_695delCCCCGGCAG p.S235_G237delSPG Chr.19: 56159904-56159912 on Build GRCh38
5959532 c.655_657delAAG p.K219delK Chr.19: 56159872-56159874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886732523 Chr.19: 56160278-56160278 on Build GRCh38
rs1007945379 Chr.19: 56160189-56160189 on Build GRCh38
rs766565433 Chr.19: 56160087-56160087 on Build GRCh38
rs762773166 Chr.19: 56160105-56160105 on Build GRCh38
rs371421706 Chr.19: 56159904-56159912 on Build GRCh38
rs370843874 Chr.19: 56160224-56160224 on Build GRCh38
rs200554083 Chr.19: 56160023-56160023 on Build GRCh38
rs776722837 Chr.19: 56160093-56160093 on Build GRCh38
rs975358348 Chr.19: 56160052-56160052 on Build GRCh38
rs767498333 Chr.19: 56160135-56160135 on Build GRCh38
rs199754530 Chr.19: 56160004-56160004 on Build GRCh38
rs758088529 Chr.19: 56160277-56160277 on Build GRCh38
rs751407962 Chr.19: 56160122-56160122 on Build GRCh38
rs756663802 Chr.19: 56160227-56160227 on Build GRCh38
rs978733634 Chr.19: 56160312-56160312 on Build GRCh38
rs145075834 Chr.19: 56159990-56159990 on Build GRCh38
rs1029603173 Chr.19: 56160217-56160217 on Build GRCh38
rs891886397 Chr.19: 56159939-56159939 on Build GRCh38
rs868037843 Chr.19: 56160025-56160025 on Build GRCh38
rs748588681 Chr.19: 56160214-56160214 on Build GRCh38
rs938720779 Chr.19: 56159929-56159929 on Build GRCh38
rs890660779 Chr.19: 56160328-56160328 on Build GRCh38
rs1030610758 Chr.19: 56159995-56159995 on Build GRCh38
rs931197230 Chr.19: 56160268-56160268 on Build GRCh38
rs1053505291 Chr.19: 56159934-56159934 on Build GRCh38
rs1012035696 Chr.19: 56159947-56159947 on Build GRCh38
rs928543324 Chr.19: 56159927-56159927 on Build GRCh38
rs991326139 Chr.19: 56160259-56160259 on Build GRCh38
rs774872693 Chr.19: 56160063-56160063 on Build GRCh38
rs1044326944 Chr.19: 56159911-56159911 on Build GRCh38
rs996350859 Chr.19: 56160211-56160211 on Build GRCh38
rs538483363 Chr.19: 56160218-56160218 on Build GRCh38
rs750000280 Chr.19: 56160262-56160262 on Build GRCh38
rs866559909 Chr.19: 56159914-56159914 on Build GRCh38
rs938569551 Chr.19: 56160124-56160124 on Build GRCh38
rs773473549 Chr.19: 56159912-56159912 on Build GRCh38
rs779115309 Chr.19: 56160210-56160210 on Build GRCh38
rs79199638 Chr.19: 56160078-56160078 on Build GRCh38
rs10407167 Chr.19: 56160323-56160323 on Build GRCh38
rs766525748 Chr.19: 56159903-56159904 on Build GRCh38
rs566678113 Chr.19: 56160183-56160183 on Build GRCh38
rs552719499 Chr.19: 56159969-56159969 on Build GRCh38
rs922022272 Chr.19: 56160319-56160319 on Build GRCh38
rs147321718 Chr.19: 56160331-56160331 on Build GRCh38
rs1026684931 Chr.19: 56160099-56160099 on Build GRCh38
rs966243956 Chr.19: 56160020-56160020 on Build GRCh38
rs981895600 Chr.19: 56160246-56160246 on Build GRCh38
rs554039855 Chr.19: 56160307-56160307 on Build GRCh38
rs760947107 Chr.19: 56159916-56159916 on Build GRCh38
rs1025351453 Chr.19: 56159950-56159950 on Build GRCh38
rs779714767 Chr.19: 56160182-56160182 on Build GRCh38
rs913262293 Chr.19: 56160145-56160145 on Build GRCh38
rs898407311 Chr.19: 56160143-56160143 on Build GRCh38
rs35920961 Chr.19: 56159998-56159999 on Build GRCh38
rs777166885 Chr.19: 56160288-56160290 on Build GRCh38
rs1045808425 Chr.19: 56160109-56160109 on Build GRCh38
rs540945373 Chr.19: 56160306-56160306 on Build GRCh38
rs752524959 Chr.19: 56159968-56159968 on Build GRCh38
rs766776396 Chr.19: 56159930-56159930 on Build GRCh38
rs772744832 Chr.19: 56160072-56160072 on Build GRCh38
rs754707088 Chr.19: 56160208-56160208 on Build GRCh38
rs955103675 Chr.19: 56160064-56160064 on Build GRCh38
rs1011750371 Chr.19: 56160330-56160330 on Build GRCh38
rs937193612 Chr.19: 56160136-56160136 on Build GRCh38
rs756711864 Chr.19: 56159982-56159982 on Build GRCh38
rs114090769 Chr.19: 56160161-56160161 on Build GRCh38
rs555942669 Chr.19: 56159912-56159913 on Build GRCh38
rs755748360 Chr.19: 56160021-56160021 on Build GRCh38
rs552871613 Chr.19: 56160230-56160230 on Build GRCh38
rs747020703 Chr.19: 56160231-56160231 on Build GRCh38
rs745426327 Chr.19: 56160002-56160002 on Build GRCh38
rs948555568 Chr.19: 56159907-56159907 on Build GRCh38
rs779584226 Chr.19: 56160037-56160037 on Build GRCh38
rs374764030 Chr.19: 56160084-56160084 on Build GRCh38
rs60359254 Chr.19: 56159919-56159927 on Build GRCh38
rs937457690 Chr.19: 56160255-56160255 on Build GRCh38
rs768929499 Chr.19: 56160054-56160054 on Build GRCh38
rs778198743 Chr.19: 56160320-56160320 on Build GRCh38
rs537440349 Chr.19: 56160249-56160249 on Build GRCh38
rs1056075536 Chr.19: 56160142-56160142 on Build GRCh38
rs536095455 Chr.19: 56160128-56160128 on Build GRCh38
rs931249506 Chr.19: 56160158-56160158 on Build GRCh38
rs60367783 Chr.19: 56160287-56160287 on Build GRCh38
rs995171042 Chr.19: 56159943-56159943 on Build GRCh38
rs776867150 Chr.19: 56160245-56160245 on Build GRCh38
rs767727906 Chr.19: 56159895-56159895 on Build GRCh38
rs749596185 Chr.19: 56160044-56160044 on Build GRCh38
rs770948744 Chr.19: 56160232-56160232 on Build GRCh38
rs554560510 Chr.19: 56160276-56160276 on Build GRCh38
rs763821520 Chr.19: 56160114-56160114 on Build GRCh38
rs748641556 Chr.19: 56160069-56160069 on Build GRCh38
rs899566911 Chr.19: 56160293-56160293 on Build GRCh38
rs979888617 Chr.19: 56160120-56160120 on Build GRCh38
rs546428125 Chr.19: 56160162-56160162 on Build GRCh38
rs201128759 Chr.19: 56159970-56159970 on Build GRCh38
rs867834411 Chr.19: 56160041-56160041 on Build GRCh38
rs867836392 Chr.19: 56159885-56159885 on Build GRCh38
rs1041141167 Chr.19: 56160288-56160288 on Build GRCh38

View Full Product Details