Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56577411-56577915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF470
Entrez Gene ID
388566
Gene Name
zinc finger protein 470
Gene Aliases
CZF-1
UniGene
Hs.204449
Gene Chromosome Location
Chr.19: 56567521-56582894 on Build GRCh38
Transcripts
NM_001001668.3 XM_017026801.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4461893 c.1225C>T p.H409Y Chr.19: 56577654-56577654 on Build GRCh38
6266020 c.1049G>T p.S350I Chr.19: 56577478-56577478 on Build GRCh38
2970988 c.1223A>T p.D408V Chr.19: 56577652-56577652 on Build GRCh38
5070258 c.1134G>C p.R378S Chr.19: 56577563-56577563 on Build GRCh38
2970992 c.1390C>T p.H464Y Chr.19: 56577819-56577819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868208018 Chr.19: 56577852-56577852 on Build GRCh38
rs372589720 Chr.19: 56577773-56577773 on Build GRCh38
rs761964508 Chr.19: 56577742-56577742 on Build GRCh38
rs760297717 Chr.19: 56577598-56577598 on Build GRCh38
rs773584915 Chr.19: 56577885-56577886 on Build GRCh38
rs780787611 Chr.19: 56577796-56577796 on Build GRCh38
rs769457764 Chr.19: 56577798-56577798 on Build GRCh38
rs957465216 Chr.19: 56577739-56577739 on Build GRCh38
rs560799616 Chr.19: 56577599-56577599 on Build GRCh38
rs201316896 Chr.19: 56577794-56577794 on Build GRCh38
rs765716853 Chr.19: 56577626-56577626 on Build GRCh38
rs767307148 Chr.19: 56577467-56577467 on Build GRCh38
rs773890131 Chr.19: 56577706-56577710 on Build GRCh38
rs777773802 Chr.19: 56577792-56577792 on Build GRCh38
rs937203939 Chr.19: 56577478-56577478 on Build GRCh38
rs765173950 Chr.19: 56577777-56577777 on Build GRCh38
rs756696301 Chr.19: 56577871-56577871 on Build GRCh38
rs1024819846 Chr.19: 56577846-56577846 on Build GRCh38
rs762985878 Chr.19: 56577633-56577633 on Build GRCh38
rs138257907 Chr.19: 56577857-56577857 on Build GRCh38
rs779977652 Chr.19: 56577598-56577600 on Build GRCh38
rs75583541 Chr.19: 56577475-56577475 on Build GRCh38
rs756449821 Chr.19: 56577474-56577474 on Build GRCh38
rs3752180 Chr.19: 56577482-56577482 on Build GRCh38
rs761504445 Chr.19: 56577744-56577745 on Build GRCh38
rs202150794 Chr.19: 56577652-56577652 on Build GRCh38
rs1029010608 Chr.19: 56577738-56577738 on Build GRCh38
rs761097712 Chr.19: 56577537-56577537 on Build GRCh38
rs768984736 Chr.19: 56577823-56577823 on Build GRCh38
rs766896534 Chr.19: 56577476-56577476 on Build GRCh38
rs376482448 Chr.19: 56577615-56577615 on Build GRCh38
rs757380097 Chr.19: 56577641-56577641 on Build GRCh38
rs763931556 Chr.19: 56577859-56577859 on Build GRCh38
rs143336519 Chr.19: 56577584-56577584 on Build GRCh38
rs141373983 Chr.19: 56577583-56577583 on Build GRCh38
rs779693482 Chr.19: 56577893-56577893 on Build GRCh38
rs773957977 Chr.19: 56577515-56577515 on Build GRCh38
rs766697736 Chr.19: 56577540-56577540 on Build GRCh38
rs767622553 Chr.19: 56577747-56577747 on Build GRCh38
rs187958416 Chr.19: 56577614-56577614 on Build GRCh38
rs750733806 Chr.19: 56577469-56577469 on Build GRCh38
rs759989506 Chr.19: 56577543-56577543 on Build GRCh38
rs974884820 Chr.19: 56577701-56577701 on Build GRCh38
rs758528878 Chr.19: 56577554-56577554 on Build GRCh38
rs375575481 Chr.19: 56577526-56577526 on Build GRCh38
rs751553748 Chr.19: 56577868-56577868 on Build GRCh38
rs200694220 Chr.19: 56577886-56577886 on Build GRCh38
rs752713151 Chr.19: 56577549-56577549 on Build GRCh38
rs759535979 Chr.19: 56577875-56577875 on Build GRCh38
rs753630888 Chr.19: 56577648-56577648 on Build GRCh38
rs771794213 Chr.19: 56577529-56577529 on Build GRCh38
rs192433985 Chr.19: 56577667-56577667 on Build GRCh38
rs1005358299 Chr.19: 56577448-56577448 on Build GRCh38
rs745675636 Chr.19: 56577501-56577501 on Build GRCh38
rs548100904 Chr.19: 56577800-56577800 on Build GRCh38
rs531878260 Chr.19: 56577708-56577708 on Build GRCh38
rs770427413 Chr.19: 56577680-56577680 on Build GRCh38
rs147638149 Chr.19: 56577531-56577531 on Build GRCh38
rs766743638 Chr.19: 56577843-56577843 on Build GRCh38
rs746484714 Chr.19: 56577793-56577793 on Build GRCh38
rs771593016 Chr.19: 56577666-56577666 on Build GRCh38
rs886800028 Chr.19: 56577592-56577592 on Build GRCh38
rs752566496 Chr.19: 56577778-56577778 on Build GRCh38
rs749666600 Chr.19: 56577810-56577810 on Build GRCh38
rs781479650 Chr.19: 56577643-56577643 on Build GRCh38
rs753265303 Chr.19: 56577632-56577632 on Build GRCh38
rs761507476 Chr.19: 56577462-56577462 on Build GRCh38
rs986311734 Chr.19: 56577581-56577581 on Build GRCh38
rs748252363 Chr.19: 56577498-56577498 on Build GRCh38
rs758231027 Chr.19: 56577788-56577788 on Build GRCh38
rs771775537 Chr.19: 56577589-56577589 on Build GRCh38
rs995144568 Chr.19: 56577578-56577578 on Build GRCh38
rs749602588 Chr.19: 56577895-56577895 on Build GRCh38
rs200592589 Chr.19: 56577591-56577591 on Build GRCh38
rs768645945 Chr.19: 56577586-56577586 on Build GRCh38
rs201323866 Chr.19: 56577635-56577635 on Build GRCh38
rs768735461 Chr.19: 56577511-56577511 on Build GRCh38
rs745564994 Chr.19: 56577797-56577797 on Build GRCh38
rs767746225 Chr.19: 56577834-56577834 on Build GRCh38
rs770179906 Chr.19: 56577699-56577699 on Build GRCh38
rs754880278 Chr.19: 56577651-56577651 on Build GRCh38
rs774981304 Chr.19: 56577826-56577826 on Build GRCh38
rs769540527 Chr.19: 56577507-56577507 on Build GRCh38
rs746563326 Chr.19: 56577676-56577676 on Build GRCh38
rs907031940 Chr.19: 56577447-56577447 on Build GRCh38
rs941307988 Chr.19: 56577771-56577771 on Build GRCh38
rs754722713 Chr.19: 56577844-56577845 on Build GRCh38
rs751682914 Chr.19: 56577638-56577638 on Build GRCh38
rs745413929 Chr.19: 56577885-56577885 on Build GRCh38
rs761797179 Chr.19: 56577831-56577831 on Build GRCh38
rs769735021 Chr.19: 56577594-56577596 on Build GRCh38
rs774226471 Chr.19: 56577714-56577714 on Build GRCh38
rs140927905 Chr.19: 56577477-56577477 on Build GRCh38
rs749277241 Chr.19: 56577615-56577615 on Build GRCh38
rs377463589 Chr.19: 56577459-56577459 on Build GRCh38
rs140590806 Chr.19: 56577839-56577839 on Build GRCh38
rs183366197 Chr.19: 56577588-56577588 on Build GRCh38
rs779016415 Chr.19: 56577497-56577497 on Build GRCh38
rs764693087 Chr.19: 56577713-56577713 on Build GRCh38
rs765767726 Chr.19: 56577547-56577547 on Build GRCh38
rs759903754 Chr.19: 56577616-56577616 on Build GRCh38
rs748136872 Chr.19: 56577663-56577663 on Build GRCh38
rs200493322 Chr.19: 56577541-56577541 on Build GRCh38
rs754790253 Chr.19: 56577774-56577774 on Build GRCh38
rs201567590 Chr.19: 56577464-56577464 on Build GRCh38
rs888322464 Chr.19: 56577444-56577444 on Build GRCh38
rs751765490 Chr.19: 56577582-56577582 on Build GRCh38
rs756542332 Chr.19: 56577822-56577822 on Build GRCh38
rs200300611 Chr.19: 56577508-56577508 on Build GRCh38
rs771643675 Chr.19: 56577760-56577761 on Build GRCh38
rs4801177 Chr.19: 56577682-56577682 on Build GRCh38
rs558252392 Chr.19: 56577504-56577504 on Build GRCh38
rs150408042 Chr.19: 56577841-56577841 on Build GRCh38
rs768146876 Chr.19: 56577620-56577623 on Build GRCh38
rs369287651 Chr.19: 56577505-56577505 on Build GRCh38
rs763401209 Chr.19: 56577709-56577709 on Build GRCh38
rs1046082606 Chr.19: 56577445-56577445 on Build GRCh38
rs746224654 Chr.19: 56577695-56577695 on Build GRCh38
rs780628210 Chr.19: 56577872-56577872 on Build GRCh38
rs369171394 Chr.19: 56577580-56577580 on Build GRCh38

View Full Product Details