Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57398739-57399235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF548
Entrez Gene ID
147694
Gene Name
zinc finger protein 548
Gene Aliases
-
UniGene
Hs.126905
Gene Chromosome Location
Chr.19: 57389850-57403497 on Build GRCh38
Transcripts
NM_001172773.1 NM_152909.3 XM_017026316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5511131 c.670T>C p.F224L Chr.19: 57398921-57398921 on Build GRCh38
2972258 c.495C>T p.F165F Chr.19: 57398746-57398746 on Build GRCh38
5464790 c.663C>T p.S221S Chr.19: 57398914-57398914 on Build GRCh38
6558436 c.865G>T p.G289* Chr.19: 57399116-57399116 on Build GRCh38
286915 c.959C>T p.P320L Chr.19: 57399210-57399210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891025667 Chr.19: 57398793-57398793 on Build GRCh38
rs769910816 Chr.19: 57399091-57399091 on Build GRCh38
rs776724937 Chr.19: 57399183-57399183 on Build GRCh38
rs956209760 Chr.19: 57398925-57398925 on Build GRCh38
rs760441279 Chr.19: 57399158-57399158 on Build GRCh38
rs969246568 Chr.19: 57399145-57399145 on Build GRCh38
rs866867902 Chr.19: 57398882-57398882 on Build GRCh38
rs746118409 Chr.19: 57398914-57398914 on Build GRCh38
rs371609505 Chr.19: 57399209-57399209 on Build GRCh38
rs756155520 Chr.19: 57399211-57399211 on Build GRCh38
rs369202044 Chr.19: 57398892-57398892 on Build GRCh38
rs772945529 Chr.19: 57399154-57399154 on Build GRCh38
rs532474115 Chr.19: 57398792-57398792 on Build GRCh38
rs752143358 Chr.19: 57399078-57399078 on Build GRCh38
rs1017847570 Chr.19: 57398860-57398860 on Build GRCh38
rs764744503 Chr.19: 57399171-57399171 on Build GRCh38
rs369371650 Chr.19: 57398816-57398816 on Build GRCh38
rs761643751 Chr.19: 57398833-57398833 on Build GRCh38
rs902120385 Chr.19: 57398907-57398907 on Build GRCh38
rs752383478 Chr.19: 57399045-57399045 on Build GRCh38
rs752295171 Chr.19: 57399172-57399172 on Build GRCh38
rs779110837 Chr.19: 57398774-57398774 on Build GRCh38
rs757660473 Chr.19: 57399208-57399208 on Build GRCh38
rs756037270 Chr.19: 57399142-57399143 on Build GRCh38
rs763979758 Chr.19: 57398819-57398819 on Build GRCh38
rs182228835 Chr.19: 57399087-57399087 on Build GRCh38
rs778480593 Chr.19: 57398916-57398916 on Build GRCh38
rs775746078 Chr.19: 57398959-57398959 on Build GRCh38
rs777005901 Chr.19: 57398815-57398815 on Build GRCh38
rs376286330 Chr.19: 57398876-57398876 on Build GRCh38
rs989412880 Chr.19: 57399136-57399136 on Build GRCh38
rs768901752 Chr.19: 57399191-57399210 on Build GRCh38
rs750352130 Chr.19: 57398850-57398850 on Build GRCh38
rs780195801 Chr.19: 57399104-57399104 on Build GRCh38
rs879251352 Chr.19: 57399176-57399176 on Build GRCh38
rs960300212 Chr.19: 57399099-57399099 on Build GRCh38
rs760569948 Chr.19: 57399001-57399001 on Build GRCh38
rs555220756 Chr.19: 57399210-57399210 on Build GRCh38
rs755376790 Chr.19: 57398765-57398765 on Build GRCh38
rs373292922 Chr.19: 57398919-57398919 on Build GRCh38
rs772306412 Chr.19: 57398977-57398977 on Build GRCh38
rs370946701 Chr.19: 57399012-57399012 on Build GRCh38
rs754480636 Chr.19: 57399028-57399028 on Build GRCh38
rs757789056 Chr.19: 57399081-57399081 on Build GRCh38
rs771137717 Chr.19: 57398936-57398936 on Build GRCh38
rs201075931 Chr.19: 57399200-57399200 on Build GRCh38
rs769956796 Chr.19: 57398956-57398956 on Build GRCh38
rs761576701 Chr.19: 57398966-57398966 on Build GRCh38
rs769910723 Chr.19: 57399022-57399023 on Build GRCh38
rs746121692 Chr.19: 57398947-57398947 on Build GRCh38
rs759027760 Chr.19: 57399161-57399161 on Build GRCh38
rs202091268 Chr.19: 57399159-57399159 on Build GRCh38
rs377513609 Chr.19: 57398984-57398984 on Build GRCh38
rs748713581 Chr.19: 57398778-57398778 on Build GRCh38
rs200185139 Chr.19: 57398853-57398853 on Build GRCh38
rs372817302 Chr.19: 57399122-57399122 on Build GRCh38
rs781705875 Chr.19: 57399086-57399086 on Build GRCh38
rs764867687 Chr.19: 57399035-57399035 on Build GRCh38
rs760514967 Chr.19: 57399015-57399015 on Build GRCh38
rs776532384 Chr.19: 57398946-57398946 on Build GRCh38
rs753337964 Chr.19: 57399022-57399022 on Build GRCh38
rs372465326 Chr.19: 57398848-57398848 on Build GRCh38
rs566751484 Chr.19: 57399199-57399199 on Build GRCh38
rs374153571 Chr.19: 57399105-57399105 on Build GRCh38
rs199925910 Chr.19: 57399138-57399138 on Build GRCh38
rs113738931 Chr.19: 57399130-57399130 on Build GRCh38
rs550331987 Chr.19: 57398840-57398840 on Build GRCh38
rs772507175 Chr.19: 57398781-57398781 on Build GRCh38
rs758845370 Chr.19: 57399046-57399046 on Build GRCh38
rs200683304 Chr.19: 57398855-57398855 on Build GRCh38
rs553311847 Chr.19: 57399174-57399174 on Build GRCh38
rs773141345 Chr.19: 57398986-57398986 on Build GRCh38
rs773964904 Chr.19: 57398826-57398826 on Build GRCh38
rs201500041 Chr.19: 57398795-57398795 on Build GRCh38
rs758831296 Chr.19: 57398906-57398906 on Build GRCh38
rs537484457 Chr.19: 57398862-57398862 on Build GRCh38
rs199971340 Chr.19: 57399056-57399056 on Build GRCh38
rs775563713 Chr.19: 57399068-57399072 on Build GRCh38
rs747153591 Chr.19: 57398794-57398794 on Build GRCh38
rs548848983 Chr.19: 57398859-57398859 on Build GRCh38

View Full Product Details