Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58127047-58127546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF329
Entrez Gene ID
79673
Gene Name
zinc finger protein 329
Gene Aliases
-
UniGene
Hs.458377
Gene Chromosome Location
Chr.19: 58126328-58155169 on Build GRCh38
Transcripts
NM_024620.3 XM_006723381.2 XM_006723382.3 XM_006723383.3 XM_006723384.3 XM_011527310.2 XM_011527311.2 XM_011527312.2 XM_011527313.2 XM_011527314.2 XM_011527315.1 XM_011527316.2 XM_017027307.1 XM_017027308.1 XM_017027309.1 XM_017027310.1 XM_017027311.1

SNP Detail

SNP ID Chromosome Location
rs899512641 Chr.19: 58127109-58127109 on Build GRCh38
rs923357341 Chr.19: 58127123-58127123 on Build GRCh38
rs1050983012 Chr.19: 58127187-58127188 on Build GRCh38
rs1035302724 Chr.19: 58127337-58127337 on Build GRCh38
rs766840527 Chr.19: 58127260-58127260 on Build GRCh38
rs76993997 Chr.19: 58127498-58127498 on Build GRCh38
rs573305625 Chr.19: 58127482-58127482 on Build GRCh38
rs746987993 Chr.19: 58127122-58127122 on Build GRCh38
rs2547258 Chr.19: 58127318-58127318 on Build GRCh38
rs80095486 Chr.19: 58127499-58127499 on Build GRCh38
rs538327301 Chr.19: 58127135-58127135 on Build GRCh38
rs553977344 Chr.19: 58127456-58127456 on Build GRCh38
rs1045192509 Chr.19: 58127499-58127499 on Build GRCh38
rs886587285 Chr.19: 58127465-58127465 on Build GRCh38
rs1026502040 Chr.19: 58127458-58127458 on Build GRCh38
rs1017170149 Chr.19: 58127244-58127244 on Build GRCh38
rs574742192 Chr.19: 58127355-58127355 on Build GRCh38
rs757987740 Chr.19: 58127522-58127522 on Build GRCh38
rs974887039 Chr.19: 58127121-58127121 on Build GRCh38
rs1039385906 Chr.19: 58127080-58127080 on Build GRCh38
rs537843766 Chr.19: 58127316-58127316 on Build GRCh38
rs769270683 Chr.19: 58127438-58127438 on Build GRCh38
rs1038250795 Chr.19: 58127485-58127485 on Build GRCh38
rs778521756 Chr.19: 58127143-58127143 on Build GRCh38
rs945332493 Chr.19: 58127288-58127288 on Build GRCh38
rs74269140 Chr.19: 58127497-58127498 on Build GRCh38
rs996138363 Chr.19: 58127520-58127520 on Build GRCh38
rs566233056 Chr.19: 58127073-58127073 on Build GRCh38
rs957167022 Chr.19: 58127338-58127338 on Build GRCh38
rs899287435 Chr.19: 58127257-58127257 on Build GRCh38
rs1044018997 Chr.19: 58127315-58127315 on Build GRCh38
rs969391669 Chr.19: 58127277-58127277 on Build GRCh38
rs913710203 Chr.19: 58127275-58127275 on Build GRCh38
rs1016918754 Chr.19: 58127497-58127497 on Build GRCh38
rs775064992 Chr.19: 58127090-58127091 on Build GRCh38
rs568787908 Chr.19: 58127278-58127278 on Build GRCh38
rs887005980 Chr.19: 58127207-58127207 on Build GRCh38
rs747820102 Chr.19: 58127389-58127389 on Build GRCh38
rs546751472 Chr.19: 58127071-58127071 on Build GRCh38
rs1013312498 Chr.19: 58127444-58127444 on Build GRCh38
rs964778588 Chr.19: 58127519-58127519 on Build GRCh38
rs775699293 Chr.19: 58127485-58127485 on Build GRCh38
rs1006500700 Chr.19: 58127493-58127493 on Build GRCh38
rs1050703508 Chr.19: 58127472-58127472 on Build GRCh38
rs556528847 Chr.19: 58127215-58127215 on Build GRCh38
rs915394944 Chr.19: 58127408-58127410 on Build GRCh38
rs879878647 Chr.19: 58127484-58127485 on Build GRCh38
rs968149834 Chr.19: 58127415-58127415 on Build GRCh38
rs1054755618 Chr.19: 58127418-58127418 on Build GRCh38
rs533977588 Chr.19: 58127424-58127424 on Build GRCh38

View Full Product Details