Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 669665-670171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM18
Entrez Gene ID
129787
Gene Name
transmembrane protein 18
Gene Aliases
-
UniGene
Hs.43899
Gene Chromosome Location
Chr.2: 667973-677439 on Build GRCh38
Transcripts
NM_152834.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7309801 c.328-1_339delGGTTATGTGGGTA p.? Chr.2: 669664-669676 on Build GRCh38
6228142 c.327+4C>T p.? Chr.2: 669753-669753 on Build GRCh38
7201081 c.227G>A p.R76K Chr.2: 669818-669818 on Build GRCh38
5782704 c.231G>A p.G77G Chr.2: 669814-669814 on Build GRCh38
7230676 c.288+5G>A p.? Chr.2: 669752-669752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890433010 Chr.2: 669798-669798 on Build GRCh38
rs1034529867 Chr.2: 669892-669892 on Build GRCh38
rs142977259 Chr.2: 669800-669800 on Build GRCh38
rs77005891 Chr.2: 669801-669801 on Build GRCh38
rs771991343 Chr.2: 669896-669896 on Build GRCh38
rs750192373 Chr.2: 669846-669846 on Build GRCh38
rs897867846 Chr.2: 670019-670019 on Build GRCh38
rs142026805 Chr.2: 670092-670092 on Build GRCh38
rs771274683 Chr.2: 669822-669822 on Build GRCh38
rs748631143 Chr.2: 669811-669811 on Build GRCh38
rs758700921 Chr.2: 669850-669851 on Build GRCh38
rs750032422 Chr.2: 669768-669768 on Build GRCh38
rs145340899 Chr.2: 669841-669841 on Build GRCh38
rs773688363 Chr.2: 669816-669816 on Build GRCh38
rs774495261 Chr.2: 669825-669825 on Build GRCh38
rs780588203 Chr.2: 669886-669886 on Build GRCh38
rs770092446 Chr.2: 669924-669924 on Build GRCh38
rs112472519 Chr.2: 669698-669698 on Build GRCh38
rs542852716 Chr.2: 669970-669970 on Build GRCh38
rs553505503 Chr.2: 669956-669956 on Build GRCh38
rs183569069 Chr.2: 669972-669972 on Build GRCh38
rs746554077 Chr.2: 669693-669693 on Build GRCh38
rs1049331531 Chr.2: 670039-670039 on Build GRCh38
rs959711342 Chr.2: 669923-669923 on Build GRCh38
rs770232573 Chr.2: 669815-669815 on Build GRCh38
rs756981253 Chr.2: 669762-669762 on Build GRCh38
rs759602383 Chr.2: 669753-669753 on Build GRCh38
rs560980133 Chr.2: 669784-669784 on Build GRCh38
rs779726251 Chr.2: 669781-669781 on Build GRCh38
rs1027520189 Chr.2: 669974-669974 on Build GRCh38
rs73908197 Chr.2: 669951-669951 on Build GRCh38
rs373260889 Chr.2: 669744-669744 on Build GRCh38
rs778713907 Chr.2: 669767-669767 on Build GRCh38
rs761458492 Chr.2: 670002-670002 on Build GRCh38
rs376441110 Chr.2: 670005-670005 on Build GRCh38
rs766756079 Chr.2: 669746-669746 on Build GRCh38
rs906168214 Chr.2: 669920-669920 on Build GRCh38
rs79223420 Chr.2: 669739-669739 on Build GRCh38
rs765995956 Chr.2: 669876-669876 on Build GRCh38
rs769196836 Chr.2: 669702-669702 on Build GRCh38
rs1019961683 Chr.2: 670115-670115 on Build GRCh38
rs77087765 Chr.2: 669940-669940 on Build GRCh38
rs73153245 Chr.2: 670125-670125 on Build GRCh38
rs191717477 Chr.2: 669968-669968 on Build GRCh38
rs186521598 Chr.2: 670131-670131 on Build GRCh38
rs142385490 Chr.2: 669847-669847 on Build GRCh38
rs150192904 Chr.2: 670065-670065 on Build GRCh38
rs756667539 Chr.2: 669894-669894 on Build GRCh38
rs764080063 Chr.2: 669761-669761 on Build GRCh38
rs571320295 Chr.2: 670036-670036 on Build GRCh38
rs146322269 Chr.2: 670099-670099 on Build GRCh38
rs777058415 Chr.2: 669732-669732 on Build GRCh38
rs576121235 Chr.2: 669977-669977 on Build GRCh38
rs13430104 Chr.2: 670102-670102 on Build GRCh38
rs925794817 Chr.2: 669988-669988 on Build GRCh38
rs754468561 Chr.2: 669877-669877 on Build GRCh38
rs375724395 Chr.2: 670147-670147 on Build GRCh38
rs749710449 Chr.2: 669821-669821 on Build GRCh38
rs761763792 Chr.2: 669836-669836 on Build GRCh38
rs907422224 Chr.2: 669909-669909 on Build GRCh38
rs751627780 Chr.2: 669752-669752 on Build GRCh38
rs747735989 Chr.2: 669889-669889 on Build GRCh38
rs142477369 Chr.2: 669826-669826 on Build GRCh38
rs530042736 Chr.2: 670145-670145 on Build GRCh38
rs141120545 Chr.2: 670012-670012 on Build GRCh38
rs969982865 Chr.2: 669842-669842 on Build GRCh38
rs767500299 Chr.2: 669832-669832 on Build GRCh38
rs931819920 Chr.2: 669990-669990 on Build GRCh38
rs138829009 Chr.2: 670091-670091 on Build GRCh38
rs749665670 Chr.2: 669899-669899 on Build GRCh38
rs749933844 Chr.2: 670062-670062 on Build GRCh38
rs963568292 Chr.2: 669959-669959 on Build GRCh38
rs758027738 Chr.2: 669776-669776 on Build GRCh38
rs558564474 Chr.2: 669933-669933 on Build GRCh38
rs1024603267 Chr.2: 669913-669913 on Build GRCh38
rs762176457 Chr.2: 669733-669733 on Build GRCh38
rs746510478 Chr.2: 669785-669785 on Build GRCh38
rs79117822 Chr.2: 669997-669997 on Build GRCh38
rs978367128 Chr.2: 670042-670042 on Build GRCh38
rs750729420 Chr.2: 669840-669841 on Build GRCh38
rs796310125 Chr.2: 669722-669722 on Build GRCh38
rs73908198 Chr.2: 669975-669975 on Build GRCh38
rs776594189 Chr.2: 669834-669834 on Build GRCh38

View Full Product Details