Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10047970-10048469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLF11
Entrez Gene ID
8462
Gene Name
Kruppel like factor 11
Gene Aliases
FKLF, FKLF1, MODY7, TIEG2, Tieg3
UniGene
Hs.12229
Gene Chromosome Location
Chr.2: 10043555-10054836 on Build GRCh38
Transcripts
NM_001177716.1 NM_001177718.1 NM_003597.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
714976 c.1126A>T p.I376F Chr.2: 10048463-10048463 on Build GRCh38
6086686 c.933G>T p.V311V Chr.2: 10048270-10048270 on Build GRCh38
82217 c.987C>T p.F329F Chr.2: 10048324-10048324 on Build GRCh38
4987394 c.994C>T p.P332S Chr.2: 10048331-10048331 on Build GRCh38
6009978 c.1058C>T p.P353L Chr.2: 10048395-10048395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769916108 Chr.2: 10048155-10048155 on Build GRCh38
rs765154301 Chr.2: 10048171-10048171 on Build GRCh38
rs144949098 Chr.2: 10048311-10048311 on Build GRCh38
rs748565756 Chr.2: 10048031-10048031 on Build GRCh38
rs966402183 Chr.2: 10048096-10048096 on Build GRCh38
rs781136269 Chr.2: 10048364-10048364 on Build GRCh38
rs753800874 Chr.2: 10048056-10048056 on Build GRCh38
rs886054723 Chr.2: 10048383-10048383 on Build GRCh38
rs751098131 Chr.2: 10048303-10048303 on Build GRCh38
rs768995860 Chr.2: 10048147-10048147 on Build GRCh38
rs374251576 Chr.2: 10048094-10048094 on Build GRCh38
rs757926432 Chr.2: 10048021-10048021 on Build GRCh38
rs781467145 Chr.2: 10048423-10048423 on Build GRCh38
rs766448123 Chr.2: 10048043-10048043 on Build GRCh38
rs763612610 Chr.2: 10048252-10048252 on Build GRCh38
rs750957935 Chr.2: 10048407-10048407 on Build GRCh38
rs750289452 Chr.2: 10048173-10048173 on Build GRCh38
rs780226216 Chr.2: 10048287-10048287 on Build GRCh38
rs1035011115 Chr.2: 10048391-10048391 on Build GRCh38
rs778387760 Chr.2: 10048277-10048277 on Build GRCh38
rs149235463 Chr.2: 10048207-10048207 on Build GRCh38
rs770836713 Chr.2: 10048307-10048307 on Build GRCh38
rs983462729 Chr.2: 10048394-10048394 on Build GRCh38
rs368577352 Chr.2: 10048292-10048292 on Build GRCh38
rs373173330 Chr.2: 10048361-10048361 on Build GRCh38
rs756696368 Chr.2: 10048325-10048325 on Build GRCh38
rs754611134 Chr.2: 10048184-10048184 on Build GRCh38
rs755222469 Chr.2: 10048421-10048421 on Build GRCh38
rs1022383826 Chr.2: 10048091-10048091 on Build GRCh38
rs780810674 Chr.2: 10048113-10048113 on Build GRCh38
rs754417822 Chr.2: 10048107-10048107 on Build GRCh38
rs371589306 Chr.2: 10048324-10048324 on Build GRCh38
rs751202965 Chr.2: 10048182-10048182 on Build GRCh38
rs886054724 Chr.2: 10048394-10048394 on Build GRCh38
rs369681560 Chr.2: 10048339-10048339 on Build GRCh38
rs200635526 Chr.2: 10048247-10048247 on Build GRCh38
rs377198797 Chr.2: 10048289-10048289 on Build GRCh38
rs772989476 Chr.2: 10048251-10048251 on Build GRCh38
rs929349988 Chr.2: 10048391-10048391 on Build GRCh38
rs964899969 Chr.2: 10048374-10048374 on Build GRCh38
rs779620355 Chr.2: 10048024-10048024 on Build GRCh38
rs754321813 Chr.2: 10048278-10048278 on Build GRCh38
rs368207952 Chr.2: 10048148-10048148 on Build GRCh38
rs1043891994 Chr.2: 10048435-10048435 on Build GRCh38
rs148922317 Chr.2: 10048008-10048008 on Build GRCh38
rs201595721 Chr.2: 10048213-10048213 on Build GRCh38
rs755666407 Chr.2: 10048275-10048275 on Build GRCh38
rs766097081 Chr.2: 10048179-10048179 on Build GRCh38
rs866704696 Chr.2: 10048216-10048216 on Build GRCh38
rs757134386 Chr.2: 10048064-10048064 on Build GRCh38
rs144431930 Chr.2: 10048290-10048290 on Build GRCh38
rs141318309 Chr.2: 10048395-10048395 on Build GRCh38
rs749614138 Chr.2: 10048136-10048136 on Build GRCh38
rs142673414 Chr.2: 10048158-10048158 on Build GRCh38
rs757722490 Chr.2: 10048283-10048283 on Build GRCh38
rs1055050235 Chr.2: 10048411-10048411 on Build GRCh38
rs1033487145 Chr.2: 10048036-10048036 on Build GRCh38
rs34336420 Chr.2: 10047996-10047996 on Build GRCh38
rs779557854 Chr.2: 10048090-10048090 on Build GRCh38
rs977679639 Chr.2: 10048110-10048110 on Build GRCh38
rs768756551 Chr.2: 10048295-10048295 on Build GRCh38
rs775364750 Chr.2: 10048386-10048386 on Build GRCh38
rs750151571 Chr.2: 10048088-10048088 on Build GRCh38
rs766503888 Chr.2: 10047995-10047995 on Build GRCh38
rs765025458 Chr.2: 10048078-10048078 on Build GRCh38
rs946559025 Chr.2: 10048427-10048427 on Build GRCh38
rs756085879 Chr.2: 10047997-10047997 on Build GRCh38
rs769770922 Chr.2: 10048248-10048248 on Build GRCh38
rs199649394 Chr.2: 10048385-10048385 on Build GRCh38
rs775612932 Chr.2: 10048215-10048215 on Build GRCh38
rs770834509 Chr.2: 10048377-10048377 on Build GRCh38
rs145504876 Chr.2: 10048143-10048143 on Build GRCh38
rs752317241 Chr.2: 10048269-10048269 on Build GRCh38
rs148123124 Chr.2: 10048119-10048119 on Build GRCh38
rs151317435 Chr.2: 10048329-10048329 on Build GRCh38
rs1030052668 Chr.2: 10048087-10048087 on Build GRCh38
rs143517559 Chr.2: 10048309-10048309 on Build GRCh38
rs372404632 Chr.2: 10048099-10048099 on Build GRCh38
rs555373433 Chr.2: 10048431-10048431 on Build GRCh38
rs759266535 Chr.2: 10048265-10048265 on Build GRCh38
rs200561467 Chr.2: 10048016-10048016 on Build GRCh38
rs145193520 Chr.2: 10048387-10048387 on Build GRCh38
rs761363367 Chr.2: 10048396-10048396 on Build GRCh38
rs774085119 Chr.2: 10048308-10048308 on Build GRCh38
rs375265808 Chr.2: 10048331-10048331 on Build GRCh38
rs758135671 Chr.2: 10048089-10048089 on Build GRCh38
rs745811457 Chr.2: 10048447-10048447 on Build GRCh38
rs781234072 Chr.2: 10048366-10048366 on Build GRCh38
rs772337993 Chr.2: 10048286-10048286 on Build GRCh38
rs760352150 Chr.2: 10048327-10048327 on Build GRCh38
rs548146556 Chr.2: 10048175-10048175 on Build GRCh38
rs766746149 Chr.2: 10048419-10048419 on Build GRCh38
rs146486664 Chr.2: 10048414-10048414 on Build GRCh38
rs267598799 Chr.2: 10048193-10048193 on Build GRCh38
rs930043820 Chr.2: 10048221-10048221 on Build GRCh38
rs779146409 Chr.2: 10048284-10048284 on Build GRCh38
rs370039403 Chr.2: 10048026-10048026 on Build GRCh38
rs572911652 Chr.2: 10048378-10048378 on Build GRCh38
rs372404152 Chr.2: 10048188-10048188 on Build GRCh38
rs777257289 Chr.2: 10048027-10048027 on Build GRCh38
rs1031608537 Chr.2: 10048316-10048316 on Build GRCh38
rs200061013 Chr.2: 10048010-10048010 on Build GRCh38
rs746495090 Chr.2: 10048025-10048025 on Build GRCh38
rs780863595 Chr.2: 10048187-10048187 on Build GRCh38
rs749713639 Chr.2: 10048201-10048201 on Build GRCh38
rs371102481 Chr.2: 10048420-10048420 on Build GRCh38
rs570616467 Chr.2: 10048285-10048285 on Build GRCh38
rs767278357 Chr.2: 10048266-10048266 on Build GRCh38
rs749197054 Chr.2: 10048305-10048305 on Build GRCh38
rs755004136 Chr.2: 10048130-10048131 on Build GRCh38
rs770367363 Chr.2: 10048128-10048128 on Build GRCh38
rs747256138 Chr.2: 10048355-10048355 on Build GRCh38
rs746577469 Chr.2: 10048106-10048106 on Build GRCh38
rs775487822 Chr.2: 10048156-10048156 on Build GRCh38
rs373828519 Chr.2: 10048034-10048034 on Build GRCh38
rs755137757 Chr.2: 10048356-10048356 on Build GRCh38
rs748230105 Chr.2: 10048426-10048426 on Build GRCh38
rs1051334448 Chr.2: 10048301-10048301 on Build GRCh38
rs774560161 Chr.2: 10048044-10048044 on Build GRCh38
rs773663627 Chr.2: 10048035-10048035 on Build GRCh38
rs778414841 Chr.2: 10048195-10048195 on Build GRCh38
rs372266786 Chr.2: 10048039-10048039 on Build GRCh38
rs745538358 Chr.2: 10048019-10048019 on Build GRCh38
rs121912645 Chr.2: 10048376-10048376 on Build GRCh38
rs1003430284 Chr.2: 10048254-10048254 on Build GRCh38
rs766954622 Chr.2: 10048264-10048264 on Build GRCh38
rs776725808 Chr.2: 10048298-10048298 on Build GRCh38
rs773720978 Chr.2: 10048130-10048130 on Build GRCh38
rs746088073 Chr.2: 10048208-10048208 on Build GRCh38
rs765703738 Chr.2: 10048406-10048406 on Build GRCh38
rs764130015 Chr.2: 10048053-10048053 on Build GRCh38
rs765760613 Chr.2: 10048335-10048335 on Build GRCh38
rs144897109 Chr.2: 10048439-10048439 on Build GRCh38
rs553083479 Chr.2: 10048332-10048332 on Build GRCh38
rs745699493 Chr.2: 10048381-10048381 on Build GRCh38
rs780743223 Chr.2: 10048365-10048365 on Build GRCh38
rs781624933 Chr.2: 10048048-10048048 on Build GRCh38
rs780354491 Chr.2: 10048341-10048341 on Build GRCh38
rs982869401 Chr.2: 10048399-10048399 on Build GRCh38
rs774416608 Chr.2: 10048144-10048144 on Build GRCh38
rs372514890 Chr.2: 10048433-10048433 on Build GRCh38
rs182304641 Chr.2: 10048257-10048257 on Build GRCh38

View Full Product Details