Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 17931661-17932166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNS3
Entrez Gene ID
3790
Gene Name
potassium voltage-gated channel modifier subfamily S member 3
Gene Aliases
KV9.3
UniGene
Hs.414489
Gene Chromosome Location
Chr.2: 17877847-17932961 on Build GRCh38
Transcripts
NM_001282428.1 NM_002252.4 XM_011532825.1 XM_017004059.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1403722 c.752G>A p.W251* Chr.2: 17931760-17931760 on Build GRCh38
3971901 c.765G>A p.L255L Chr.2: 17931773-17931773 on Build GRCh38
4766615 c.902T>G p.L301R Chr.2: 17931910-17931910 on Build GRCh38
1403719 c.704G>A p.G235E Chr.2: 17931712-17931712 on Build GRCh38
6089153 c.704G>C p.G235A Chr.2: 17931712-17931712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750486968 Chr.2: 17931767-17931767 on Build GRCh38
rs763985100 Chr.2: 17931766-17931766 on Build GRCh38
rs760585114 Chr.2: 17931885-17931885 on Build GRCh38
rs151225864 Chr.2: 17932136-17932136 on Build GRCh38
rs909473347 Chr.2: 17932032-17932032 on Build GRCh38
rs753778808 Chr.2: 17931897-17931897 on Build GRCh38
rs774736288 Chr.2: 17931969-17931969 on Build GRCh38
rs758042404 Chr.2: 17932116-17932116 on Build GRCh38
rs920954423 Chr.2: 17931707-17931707 on Build GRCh38
rs757681452 Chr.2: 17932094-17932094 on Build GRCh38
rs145204663 Chr.2: 17931691-17931691 on Build GRCh38
rs1011574710 Chr.2: 17931803-17931803 on Build GRCh38
rs775401718 Chr.2: 17931863-17931863 on Build GRCh38
rs751723451 Chr.2: 17931771-17931771 on Build GRCh38
rs937664203 Chr.2: 17932141-17932141 on Build GRCh38
rs779009471 Chr.2: 17931692-17931692 on Build GRCh38
rs780182339 Chr.2: 17931711-17931711 on Build GRCh38
rs773912262 Chr.2: 17931825-17931825 on Build GRCh38
rs371961825 Chr.2: 17931739-17931739 on Build GRCh38
rs780310813 Chr.2: 17931770-17931770 on Build GRCh38
rs142073653 Chr.2: 17931789-17931789 on Build GRCh38
rs138310384 Chr.2: 17932015-17932015 on Build GRCh38
rs1017689846 Chr.2: 17932005-17932005 on Build GRCh38
rs770286422 Chr.2: 17931822-17931822 on Build GRCh38
rs760534267 Chr.2: 17932017-17932017 on Build GRCh38
rs748591645 Chr.2: 17931725-17931725 on Build GRCh38
rs749761161 Chr.2: 17931808-17931808 on Build GRCh38
rs562414979 Chr.2: 17932040-17932040 on Build GRCh38
rs778207777 Chr.2: 17931815-17931815 on Build GRCh38
rs777976168 Chr.2: 17931882-17931897 on Build GRCh38
rs771873544 Chr.2: 17931836-17931836 on Build GRCh38
rs148665556 Chr.2: 17932105-17932105 on Build GRCh38
rs745417936 Chr.2: 17931993-17931993 on Build GRCh38
rs374189552 Chr.2: 17931931-17931931 on Build GRCh38
rs771816635 Chr.2: 17931998-17931998 on Build GRCh38
rs766302945 Chr.2: 17931921-17931921 on Build GRCh38
rs191735514 Chr.2: 17931816-17931816 on Build GRCh38
rs369862362 Chr.2: 17931939-17931939 on Build GRCh38
rs761825193 Chr.2: 17931903-17931903 on Build GRCh38
rs752619752 Chr.2: 17931689-17931689 on Build GRCh38
rs749686704 Chr.2: 17931940-17931940 on Build GRCh38
rs369263491 Chr.2: 17932079-17932079 on Build GRCh38
rs767711051 Chr.2: 17932048-17932048 on Build GRCh38
rs114910715 Chr.2: 17931722-17931722 on Build GRCh38
rs541299086 Chr.2: 17932135-17932135 on Build GRCh38
rs958132973 Chr.2: 17932026-17932026 on Build GRCh38
rs774394708 Chr.2: 17932043-17932043 on Build GRCh38
rs767353971 Chr.2: 17931745-17931745 on Build GRCh38
rs765511957 Chr.2: 17931926-17931926 on Build GRCh38
rs751674307 Chr.2: 17931922-17931922 on Build GRCh38
rs367935090 Chr.2: 17931738-17931738 on Build GRCh38
rs532095670 Chr.2: 17931786-17931786 on Build GRCh38
rs781500434 Chr.2: 17931787-17931787 on Build GRCh38
rs888223997 Chr.2: 17931869-17931869 on Build GRCh38
rs777876799 Chr.2: 17931727-17931727 on Build GRCh38
rs760640214 Chr.2: 17931754-17931754 on Build GRCh38
rs1005322359 Chr.2: 17931873-17931873 on Build GRCh38
rs201654025 Chr.2: 17931837-17931837 on Build GRCh38
rs556337820 Chr.2: 17931928-17931928 on Build GRCh38
rs1037246255 Chr.2: 17931882-17931882 on Build GRCh38
rs930979261 Chr.2: 17931709-17931710 on Build GRCh38
rs35353595 Chr.2: 17931723-17931723 on Build GRCh38
rs547884773 Chr.2: 17931701-17931701 on Build GRCh38
rs907293335 Chr.2: 17931756-17931756 on Build GRCh38
rs779601764 Chr.2: 17932100-17932100 on Build GRCh38
rs752856209 Chr.2: 17932073-17932073 on Build GRCh38
rs768387082 Chr.2: 17932142-17932142 on Build GRCh38
rs778517087 Chr.2: 17931978-17931978 on Build GRCh38
rs1054793745 Chr.2: 17931827-17931827 on Build GRCh38
rs754251858 Chr.2: 17932087-17932087 on Build GRCh38
rs140699625 Chr.2: 17932036-17932036 on Build GRCh38
rs756359466 Chr.2: 17932074-17932074 on Build GRCh38
rs143152428 Chr.2: 17932041-17932041 on Build GRCh38
rs770265810 Chr.2: 17931795-17931795 on Build GRCh38
rs375081273 Chr.2: 17931779-17931779 on Build GRCh38
rs759654395 Chr.2: 17932046-17932046 on Build GRCh38
rs759223621 Chr.2: 17931830-17931830 on Build GRCh38
rs577298726 Chr.2: 17931806-17931806 on Build GRCh38
rs771499398 Chr.2: 17931955-17931955 on Build GRCh38
rs770320711 Chr.2: 17931726-17931726 on Build GRCh38
rs762408332 Chr.2: 17931737-17931737 on Build GRCh38
rs965167461 Chr.2: 17931888-17931888 on Build GRCh38
rs966906519 Chr.2: 17932064-17932064 on Build GRCh38
rs764090269 Chr.2: 17931886-17931886 on Build GRCh38
rs758622857 Chr.2: 17931710-17931710 on Build GRCh38
rs771532058 Chr.2: 17931918-17931918 on Build GRCh38
rs753052691 Chr.2: 17931930-17931930 on Build GRCh38

View Full Product Details