Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24076079-24076574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SF3B6
Entrez Gene ID
51639
Gene Name
splicing factor 3b subunit 6
Gene Aliases
CGI-110, HSPC175, Ht006, P14, SAP14, SAP14a, SF3B14, SF3B14a
UniGene
Hs.177861
Gene Chromosome Location
Chr.2: 24067584-24076444 on Build GRCh38
Transcripts
NM_016047.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3972164 c.8T>A p.M3K Chr.2: 24076222-24076222 on Build GRCh38
1291480 c.26C>T p.A9V Chr.2: 24076204-24076204 on Build GRCh38
1406880 c.13G>T p.A5S Chr.2: 24076217-24076217 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs938716855 Chr.2: 24076360-24076360 on Build GRCh38
rs948162265 Chr.2: 24076399-24076399 on Build GRCh38
rs374464479 Chr.2: 24076139-24076139 on Build GRCh38
rs953141726 Chr.2: 24076356-24076356 on Build GRCh38
rs749282692 Chr.2: 24076276-24076276 on Build GRCh38
rs947975910 Chr.2: 24076348-24076348 on Build GRCh38
rs953288741 Chr.2: 24076467-24076467 on Build GRCh38
rs768147584 Chr.2: 24076187-24076187 on Build GRCh38
rs912832562 Chr.2: 24076180-24076180 on Build GRCh38
rs957898812 Chr.2: 24076317-24076317 on Build GRCh38
rs1030767203 Chr.2: 24076458-24076458 on Build GRCh38
rs769075521 Chr.2: 24076384-24076384 on Build GRCh38
rs923735230 Chr.2: 24076297-24076297 on Build GRCh38
rs35636238 Chr.2: 24076145-24076145 on Build GRCh38
rs541541780 Chr.2: 24076248-24076248 on Build GRCh38
rs961713217 Chr.2: 24076482-24076482 on Build GRCh38
rs931493611 Chr.2: 24076385-24076385 on Build GRCh38
rs769807017 Chr.2: 24076273-24076273 on Build GRCh38
rs886620681 Chr.2: 24076105-24076105 on Build GRCh38
rs969285641 Chr.2: 24076523-24076523 on Build GRCh38
rs906502630 Chr.2: 24076426-24076426 on Build GRCh38
rs564450783 Chr.2: 24076341-24076341 on Build GRCh38
rs371089103 Chr.2: 24076257-24076257 on Build GRCh38
rs528378956 Chr.2: 24076349-24076349 on Build GRCh38
rs972944518 Chr.2: 24076371-24076371 on Build GRCh38
rs958081631 Chr.2: 24076464-24076464 on Build GRCh38
rs116430313 Chr.2: 24076218-24076218 on Build GRCh38
rs778727087 Chr.2: 24076492-24076492 on Build GRCh38
rs1006404605 Chr.2: 24076367-24076367 on Build GRCh38
rs964700640 Chr.2: 24076162-24076162 on Build GRCh38
rs776994325 Chr.2: 24076389-24076389 on Build GRCh38
rs745667160 Chr.2: 24076252-24076252 on Build GRCh38
rs146170317 Chr.2: 24076264-24076264 on Build GRCh38
rs774882029 Chr.2: 24076149-24076149 on Build GRCh38
rs13423926 Chr.2: 24076473-24076473 on Build GRCh38
rs759943544 Chr.2: 24076174-24076174 on Build GRCh38
rs1008608365 Chr.2: 24076469-24076469 on Build GRCh38
rs755643434 Chr.2: 24076214-24076214 on Build GRCh38
rs753231728 Chr.2: 24076209-24076209 on Build GRCh38
rs916489908 Chr.2: 24076334-24076334 on Build GRCh38
rs552043274 Chr.2: 24076332-24076332 on Build GRCh38
rs879120729 Chr.2: 24076372-24076372 on Build GRCh38
rs928654611 Chr.2: 24076355-24076355 on Build GRCh38
rs1046836595 Chr.2: 24076414-24076414 on Build GRCh38
rs906485105 Chr.2: 24076329-24076329 on Build GRCh38
rs893704453 Chr.2: 24076442-24076442 on Build GRCh38
rs746962548 Chr.2: 24076271-24076271 on Build GRCh38
rs987576076 Chr.2: 24076359-24076359 on Build GRCh38
rs999192591 Chr.2: 24076456-24076456 on Build GRCh38
rs982033942 Chr.2: 24076535-24076535 on Build GRCh38
rs900742941 Chr.2: 24076392-24076392 on Build GRCh38
rs531312928 Chr.2: 24076553-24076553 on Build GRCh38
rs35100357 Chr.2: 24076318-24076318 on Build GRCh38
rs758131827 Chr.2: 24076263-24076263 on Build GRCh38
rs369320807 Chr.2: 24076436-24076436 on Build GRCh38
rs944294367 Chr.2: 24076183-24076183 on Build GRCh38
rs373453382 Chr.2: 24076235-24076235 on Build GRCh38
rs1006847574 Chr.2: 24076110-24076110 on Build GRCh38
rs773321545 Chr.2: 24076274-24076274 on Build GRCh38
rs561003609 Chr.2: 24076236-24076236 on Build GRCh38
rs895255999 Chr.2: 24076397-24076397 on Build GRCh38
rs1047813554 Chr.2: 24076363-24076363 on Build GRCh38
rs971707009 Chr.2: 24076487-24076487 on Build GRCh38
rs377564127 Chr.2: 24076250-24076250 on Build GRCh38
rs1012691714 Chr.2: 24076293-24076293 on Build GRCh38
rs1023930576 Chr.2: 24076510-24076510 on Build GRCh38
rs999413641 Chr.2: 24076427-24076427 on Build GRCh38
rs768599381 Chr.2: 24076393-24076393 on Build GRCh38
rs762029700 Chr.2: 24076390-24076390 on Build GRCh38
rs546730573 Chr.2: 24076365-24076365 on Build GRCh38
rs1029944065 Chr.2: 24076394-24076394 on Build GRCh38
rs771313669 Chr.2: 24076277-24076277 on Build GRCh38
rs551121568 Chr.2: 24076503-24076503 on Build GRCh38

View Full Product Details