Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26244656-26245156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HADHA
Entrez Gene ID
3030
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit
Gene Aliases
ECHA, GBP, HADH, LCEH, LCHAD, MTPA, TP-ALPHA
UniGene
Hs.516032
Gene Chromosome Location
Chr.2: 26190635-26244726 on Build GRCh38
Transcripts
NM_000182.4 NM_000183.2 NM_001281512.1 NM_001281513.1 XM_011532803.1 XM_017003939.1

Gene Symbol
HADHB
Entrez Gene ID
3032
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), beta subunit
Gene Aliases
ECHB, MSTP029, MTPB, TP-BETA
UniGene
Hs.515848
Gene Chromosome Location
Chr.2: 26244748-26290465 on Build GRCh38
Transcripts
NM_000182.4 NM_000183.2 NM_001281512.1 NM_001281513.1 XM_011532803.1 XM_017003939.1

SNP Detail

SNP ID Chromosome Location
rs773601932 Chr.2: 26245063-26245063 on Build GRCh38
rs950117692 Chr.2: 26244731-26244731 on Build GRCh38
rs933816749 Chr.2: 26244987-26244987 on Build GRCh38
rs542002860 Chr.2: 26244828-26244828 on Build GRCh38
rs960116242 Chr.2: 26245084-26245084 on Build GRCh38
rs886055864 Chr.2: 26244764-26244764 on Build GRCh38
rs569323176 Chr.2: 26244735-26244735 on Build GRCh38
rs922342942 Chr.2: 26244931-26244931 on Build GRCh38
rs1012719947 Chr.2: 26244988-26244988 on Build GRCh38
rs952304513 Chr.2: 26244868-26244868 on Build GRCh38
rs971480413 Chr.2: 26245000-26245000 on Build GRCh38
rs531452580 Chr.2: 26245088-26245088 on Build GRCh38
rs901047606 Chr.2: 26244976-26244976 on Build GRCh38
rs749979964 Chr.2: 26244826-26244826 on Build GRCh38
rs1054412252 Chr.2: 26244960-26244960 on Build GRCh38
rs992916753 Chr.2: 26244897-26244897 on Build GRCh38
rs902876399 Chr.2: 26244948-26244948 on Build GRCh38
rs942491812 Chr.2: 26244952-26244952 on Build GRCh38
rs886055865 Chr.2: 26244797-26244797 on Build GRCh38
rs1032863376 Chr.2: 26244695-26244695 on Build GRCh38
rs900833859 Chr.2: 26244786-26244786 on Build GRCh38
rs991599310 Chr.2: 26244724-26244724 on Build GRCh38
rs942419277 Chr.2: 26244815-26244815 on Build GRCh38
rs935686016 Chr.2: 26245137-26245137 on Build GRCh38
rs1049198683 Chr.2: 26244764-26244764 on Build GRCh38
rs749170693 Chr.2: 26244876-26244876 on Build GRCh38
rs367712851 Chr.2: 26244814-26244814 on Build GRCh38
rs1041353886 Chr.2: 26244944-26244944 on Build GRCh38
rs773668550 Chr.2: 26244940-26244940 on Build GRCh38
rs1034553504 Chr.2: 26244888-26244888 on Build GRCh38
rs919267863 Chr.2: 26244847-26244847 on Build GRCh38
rs998494698 Chr.2: 26244709-26244709 on Build GRCh38
rs558135168 Chr.2: 26244779-26244779 on Build GRCh38
rs550842864 Chr.2: 26244730-26244730 on Build GRCh38
rs547974691 Chr.2: 26245110-26245110 on Build GRCh38
rs1034850152 Chr.2: 26245078-26245078 on Build GRCh38
rs1000126643 Chr.2: 26244821-26244821 on Build GRCh38
rs938970742 Chr.2: 26244784-26244784 on Build GRCh38
rs539507145 Chr.2: 26244766-26244766 on Build GRCh38
rs960482071 Chr.2: 26244890-26244890 on Build GRCh38
rs1042667473 Chr.2: 26244811-26244812 on Build GRCh38
rs761468805 Chr.2: 26244956-26244956 on Build GRCh38
rs528080532 Chr.2: 26244974-26244974 on Build GRCh38
rs910839801 Chr.2: 26245074-26245074 on Build GRCh38
rs1000764985 Chr.2: 26244838-26244838 on Build GRCh38
rs371815532 Chr.2: 26244908-26244908 on Build GRCh38
rs564088971 Chr.2: 26244936-26244936 on Build GRCh38
rs952072020 Chr.2: 26245049-26245049 on Build GRCh38
rs889298779 Chr.2: 26244767-26244767 on Build GRCh38
rs1016296935 Chr.2: 26245055-26245055 on Build GRCh38
rs188951747 Chr.2: 26245135-26245135 on Build GRCh38
rs899148871 Chr.2: 26245068-26245068 on Build GRCh38
rs760892470 Chr.2: 26244727-26244732 on Build GRCh38
rs1016245930 Chr.2: 26244873-26244873 on Build GRCh38
rs980310311 Chr.2: 26244770-26244770 on Build GRCh38
rs574318006 Chr.2: 26244823-26244823 on Build GRCh38
rs927430831 Chr.2: 26244778-26244778 on Build GRCh38
rs532368542 Chr.2: 26244756-26244756 on Build GRCh38
rs1024425760 Chr.2: 26244834-26244834 on Build GRCh38
rs887038203 Chr.2: 26245001-26245001 on Build GRCh38
rs758062008 Chr.2: 26244882-26244882 on Build GRCh38
rs972048325 Chr.2: 26244846-26244846 on Build GRCh38
rs925673658 Chr.2: 26245122-26245122 on Build GRCh38
rs146719337 Chr.2: 26244676-26244676 on Build GRCh38
rs958749270 Chr.2: 26244714-26244714 on Build GRCh38
rs545555203 Chr.2: 26244891-26244891 on Build GRCh38
rs1024560008 Chr.2: 26244998-26244998 on Build GRCh38
rs777312410 Chr.2: 26244757-26244757 on Build GRCh38
rs1041785436 Chr.2: 26244813-26244813 on Build GRCh38
rs994004894 Chr.2: 26245026-26245026 on Build GRCh38
rs923452572 Chr.2: 26245105-26245105 on Build GRCh38
rs894303617 Chr.2: 26244966-26244966 on Build GRCh38
rs1026303994 Chr.2: 26245038-26245038 on Build GRCh38
rs886055866 Chr.2: 26245001-26245001 on Build GRCh38
rs965343951 Chr.2: 26244708-26244708 on Build GRCh38
rs575302805 Chr.2: 26244875-26244875 on Build GRCh38
rs536037279 Chr.2: 26244683-26244683 on Build GRCh38
rs72809668 Chr.2: 26244785-26244785 on Build GRCh38
rs942441747 Chr.2: 26244769-26244769 on Build GRCh38
rs964939925 Chr.2: 26245079-26245079 on Build GRCh38
rs1039421313 Chr.2: 26244923-26244923 on Build GRCh38
rs768280878 Chr.2: 26244913-26244913 on Build GRCh38
rs1041880674 Chr.2: 26245119-26245119 on Build GRCh38
rs910703873 Chr.2: 26244880-26244880 on Build GRCh38
rs3806516 Chr.2: 26244798-26244798 on Build GRCh38
rs978415074 Chr.2: 26244934-26244934 on Build GRCh38
rs137959353 Chr.2: 26245082-26245082 on Build GRCh38
rs563299915 Chr.2: 26244845-26244845 on Build GRCh38

View Full Product Details