Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27442495-27442998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTCAP3
Entrez Gene ID
200634
Gene Name
keratinocyte associated protein 3
Gene Aliases
KCP3
UniGene
Hs.59509
Gene Chromosome Location
Chr.2: 27442366-27446481 on Build GRCh38
Transcripts
NM_001168364.1 NM_001321325.1 NM_173853.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6298478 c.213+3G>T p.? Chr.2: 27442766-27442766 on Build GRCh38
2940063 c.178G>T p.V60L Chr.2: 27442728-27442728 on Build GRCh38
6390204 c.46A>G p.R16G Chr.2: 27442596-27442596 on Build GRCh38
3972202 c.111C>T p.A37A Chr.2: 27442661-27442661 on Build GRCh38
1239297 c.122G>T p.G41V Chr.2: 27442672-27442672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199618783 Chr.2: 27442795-27442795 on Build GRCh38
rs978604017 Chr.2: 27442541-27442541 on Build GRCh38
rs541928246 Chr.2: 27442519-27442519 on Build GRCh38
rs772144681 Chr.2: 27442715-27442715 on Build GRCh38
rs539437236 Chr.2: 27442805-27442805 on Build GRCh38
rs563804147 Chr.2: 27442549-27442549 on Build GRCh38
rs764536458 Chr.2: 27442762-27442762 on Build GRCh38
rs781400570 Chr.2: 27442615-27442615 on Build GRCh38
rs61732551 Chr.2: 27442800-27442800 on Build GRCh38
rs779939268 Chr.2: 27442861-27442868 on Build GRCh38
rs778479129 Chr.2: 27442606-27442606 on Build GRCh38
rs754301985 Chr.2: 27442764-27442764 on Build GRCh38
rs768820932 Chr.2: 27442787-27442787 on Build GRCh38
rs771800335 Chr.2: 27442936-27442936 on Build GRCh38
rs752594152 Chr.2: 27442592-27442592 on Build GRCh38
rs759817262 Chr.2: 27442636-27442636 on Build GRCh38
rs769577373 Chr.2: 27442902-27442902 on Build GRCh38
rs529494132 Chr.2: 27442794-27442794 on Build GRCh38
rs992755228 Chr.2: 27442517-27442517 on Build GRCh38
rs768503534 Chr.2: 27442727-27442727 on Build GRCh38
rs767602712 Chr.2: 27442665-27442679 on Build GRCh38
rs761726911 Chr.2: 27442738-27442738 on Build GRCh38
rs375274815 Chr.2: 27442784-27442784 on Build GRCh38
rs752063164 Chr.2: 27442606-27442607 on Build GRCh38
rs780073398 Chr.2: 27442783-27442783 on Build GRCh38
rs1004592347 Chr.2: 27442782-27442782 on Build GRCh38
rs763431030 Chr.2: 27442587-27442588 on Build GRCh38
rs962991411 Chr.2: 27442514-27442514 on Build GRCh38
rs746965618 Chr.2: 27442863-27442864 on Build GRCh38
rs756870946 Chr.2: 27442558-27442558 on Build GRCh38
rs545842572 Chr.2: 27442580-27442580 on Build GRCh38
rs186867851 Chr.2: 27442850-27442850 on Build GRCh38
rs752778257 Chr.2: 27442697-27442697 on Build GRCh38
rs764132012 Chr.2: 27442601-27442601 on Build GRCh38
rs1029351055 Chr.2: 27442895-27442895 on Build GRCh38
rs528795249 Chr.2: 27442698-27442698 on Build GRCh38
rs532545613 Chr.2: 27442940-27442940 on Build GRCh38
rs760505914 Chr.2: 27442600-27442600 on Build GRCh38
rs758998859 Chr.2: 27442931-27442931 on Build GRCh38
rs766100769 Chr.2: 27442678-27442678 on Build GRCh38
rs775116443 Chr.2: 27442650-27442650 on Build GRCh38
rs749995966 Chr.2: 27442677-27442677 on Build GRCh38
rs771136044 Chr.2: 27442629-27442629 on Build GRCh38
rs756782919 Chr.2: 27442605-27442605 on Build GRCh38
rs745893425 Chr.2: 27442772-27442772 on Build GRCh38
rs765211067 Chr.2: 27442745-27442745 on Build GRCh38
rs758273640 Chr.2: 27442828-27442829 on Build GRCh38
rs767316976 Chr.2: 27442814-27442814 on Build GRCh38
rs753436578 Chr.2: 27442554-27442554 on Build GRCh38
rs932674310 Chr.2: 27442570-27442570 on Build GRCh38
rs749318728 Chr.2: 27442620-27442620 on Build GRCh38
rs755164432 Chr.2: 27442878-27442878 on Build GRCh38
rs371119638 Chr.2: 27442815-27442815 on Build GRCh38
rs201863362 Chr.2: 27442802-27442802 on Build GRCh38
rs913127734 Chr.2: 27442520-27442520 on Build GRCh38
rs533810460 Chr.2: 27442959-27442959 on Build GRCh38
rs369704814 Chr.2: 27442567-27442567 on Build GRCh38
rs377479943 Chr.2: 27442591-27442591 on Build GRCh38
rs750082263 Chr.2: 27442608-27442608 on Build GRCh38
rs775295356 Chr.2: 27442937-27442937 on Build GRCh38
rs200651326 Chr.2: 27442725-27442725 on Build GRCh38
rs746725667 Chr.2: 27442726-27442726 on Build GRCh38
rs780169953 Chr.2: 27442858-27442858 on Build GRCh38
rs764723392 Chr.2: 27442674-27442674 on Build GRCh38
rs564421209 Chr.2: 27442688-27442688 on Build GRCh38
rs762813042 Chr.2: 27442971-27442971 on Build GRCh38
rs770173943 Chr.2: 27442619-27442619 on Build GRCh38
rs773788844 Chr.2: 27442806-27442806 on Build GRCh38
rs562761804 Chr.2: 27442710-27442710 on Build GRCh38
rs756510732 Chr.2: 27442691-27442691 on Build GRCh38
rs747893929 Chr.2: 27442894-27442894 on Build GRCh38
rs778118392 Chr.2: 27442752-27442753 on Build GRCh38
rs765692910 Chr.2: 27442950-27442950 on Build GRCh38
rs370582639 Chr.2: 27442958-27442958 on Build GRCh38
rs763168947 Chr.2: 27442584-27442584 on Build GRCh38
rs575937995 Chr.2: 27442741-27442742 on Build GRCh38
rs140104466 Chr.2: 27442930-27442930 on Build GRCh38
rs753245947 Chr.2: 27442604-27442604 on Build GRCh38
rs1001677984 Chr.2: 27442956-27442956 on Build GRCh38
rs1035394978 Chr.2: 27442751-27442751 on Build GRCh38
rs777608994 Chr.2: 27442904-27442904 on Build GRCh38
rs758117368 Chr.2: 27442609-27442609 on Build GRCh38
rs1039394528 Chr.2: 27442562-27442562 on Build GRCh38
rs759201593 Chr.2: 27442807-27442807 on Build GRCh38
rs749806525 Chr.2: 27442696-27442696 on Build GRCh38
rs760548117 Chr.2: 27442653-27442653 on Build GRCh38
rs759975304 Chr.2: 27442626-27442627 on Build GRCh38
rs954072195 Chr.2: 27442922-27442922 on Build GRCh38
rs562116073 Chr.2: 27442735-27442735 on Build GRCh38
rs747901419 Chr.2: 27442583-27442583 on Build GRCh38
rs1002532716 Chr.2: 27442746-27442746 on Build GRCh38
rs886732938 Chr.2: 27442826-27442826 on Build GRCh38
rs778070692 Chr.2: 27442693-27442693 on Build GRCh38
rs767502923 Chr.2: 27442529-27442529 on Build GRCh38
rs933635743 Chr.2: 27442568-27442568 on Build GRCh38
rs375999895 Chr.2: 27442673-27442673 on Build GRCh38
rs770393187 Chr.2: 27442581-27442581 on Build GRCh38
rs770820119 Chr.2: 27442916-27442916 on Build GRCh38
rs905508541 Chr.2: 27442728-27442728 on Build GRCh38
rs775803286 Chr.2: 27442720-27442720 on Build GRCh38
rs1020241017 Chr.2: 27442690-27442690 on Build GRCh38
rs200798984 Chr.2: 27442590-27442590 on Build GRCh38
rs748599228 Chr.2: 27442616-27442616 on Build GRCh38
rs761248438 Chr.2: 27442672-27442672 on Build GRCh38
rs762202480 Chr.2: 27442938-27442938 on Build GRCh38
rs749233167 Chr.2: 27442908-27442908 on Build GRCh38
rs766723934 Chr.2: 27442599-27442600 on Build GRCh38
rs565223118 Chr.2: 27442534-27442534 on Build GRCh38
rs867300887 Chr.2: 27442540-27442540 on Build GRCh38
rs370356452 Chr.2: 27442706-27442706 on Build GRCh38
rs779840704 Chr.2: 27442657-27442657 on Build GRCh38
rs779593600 Chr.2: 27442771-27442771 on Build GRCh38
rs375467712 Chr.2: 27442542-27442542 on Build GRCh38
rs758520051 Chr.2: 27442774-27442774 on Build GRCh38
rs751640885 Chr.2: 27442871-27442871 on Build GRCh38
rs765780689 Chr.2: 27442834-27442834 on Build GRCh38
rs773146825 Chr.2: 27442582-27442582 on Build GRCh38
rs772682540 Chr.2: 27442748-27442748 on Build GRCh38
rs771225635 Chr.2: 27442585-27442585 on Build GRCh38
rs370283009 Chr.2: 27442624-27442624 on Build GRCh38
rs778568876 Chr.2: 27442561-27442561 on Build GRCh38
rs751726193 Chr.2: 27442915-27442915 on Build GRCh38
rs575194923 Chr.2: 27442563-27442563 on Build GRCh38
rs750944311 Chr.2: 27442844-27442844 on Build GRCh38
rs144811599 Chr.2: 27442753-27442753 on Build GRCh38
rs754651027 Chr.2: 27442754-27442754 on Build GRCh38
rs754004983 Chr.2: 27442756-27442771 on Build GRCh38
rs762558824 Chr.2: 27442749-27442749 on Build GRCh38
rs768676208 Chr.2: 27442913-27442913 on Build GRCh38
rs372394516 Chr.2: 27442661-27442661 on Build GRCh38
rs753786461 Chr.2: 27442670-27442670 on Build GRCh38
rs781428661 Chr.2: 27442890-27442890 on Build GRCh38
rs779112199 Chr.2: 27442703-27442703 on Build GRCh38
rs757805476 Chr.2: 27442769-27442769 on Build GRCh38
rs772383785 Chr.2: 27442639-27442639 on Build GRCh38
rs200489431 Chr.2: 27442758-27442758 on Build GRCh38

View Full Product Details