Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27498639-27499141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCKR
Entrez Gene ID
2646
Gene Name
glucokinase (hexokinase 4) regulator
Gene Aliases
FGQTL5, GKRP
UniGene
Hs.89771
Gene Chromosome Location
Chr.2: 27496839-27523689 on Build GRCh38
Transcripts
NM_001486.3 XM_011532763.1 XM_017003796.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4171157 c.356T>G p.V119G Chr.2: 27498725-27498725 on Build GRCh38
4583447 c.395C>T p.P132L Chr.2: 27498764-27498764 on Build GRCh38
6467156 c.355-3C>G p.? Chr.2: 27498721-27498721 on Build GRCh38
721404 c.384G>A p.L128L Chr.2: 27498753-27498753 on Build GRCh38
6467155 c.355-4A>T p.? Chr.2: 27498720-27498720 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1024264338 Chr.2: 27499091-27499091 on Build GRCh38
rs531671010 Chr.2: 27498945-27498945 on Build GRCh38
rs774352974 Chr.2: 27498953-27498953 on Build GRCh38
rs553186246 Chr.2: 27498853-27498853 on Build GRCh38
rs768939929 Chr.2: 27498723-27498723 on Build GRCh38
rs972139687 Chr.2: 27498860-27498860 on Build GRCh38
rs900675420 Chr.2: 27499041-27499041 on Build GRCh38
rs368953418 Chr.2: 27498803-27498803 on Build GRCh38
rs813592 Chr.2: 27499104-27499104 on Build GRCh38
rs532330280 Chr.2: 27499000-27499000 on Build GRCh38
rs991471223 Chr.2: 27498721-27498721 on Build GRCh38
rs907923201 Chr.2: 27498909-27498909 on Build GRCh38
rs758922058 Chr.2: 27498819-27498819 on Build GRCh38
rs201268198 Chr.2: 27498815-27498815 on Build GRCh38
rs780716821 Chr.2: 27498824-27498824 on Build GRCh38
rs762671458 Chr.2: 27499111-27499111 on Build GRCh38
rs564926357 Chr.2: 27498989-27498989 on Build GRCh38
rs959267290 Chr.2: 27498695-27498695 on Build GRCh38
rs760656330 Chr.2: 27498995-27498995 on Build GRCh38
rs996386237 Chr.2: 27499058-27499058 on Build GRCh38
rs761658936 Chr.2: 27498736-27498736 on Build GRCh38
rs372410789 Chr.2: 27498816-27498816 on Build GRCh38
rs759053453 Chr.2: 27498770-27498770 on Build GRCh38
rs919421826 Chr.2: 27498913-27498913 on Build GRCh38
rs776995623 Chr.2: 27498732-27498732 on Build GRCh38
rs751014605 Chr.2: 27498768-27498768 on Build GRCh38
rs541974207 Chr.2: 27498903-27498903 on Build GRCh38
rs1033463558 Chr.2: 27499080-27499080 on Build GRCh38
rs971389143 Chr.2: 27499099-27499099 on Build GRCh38
rs747141244 Chr.2: 27498711-27498711 on Build GRCh38
rs766161729 Chr.2: 27499078-27499078 on Build GRCh38
rs768886978 Chr.2: 27498842-27498842 on Build GRCh38
rs150673460 Chr.2: 27498764-27498764 on Build GRCh38
rs140017894 Chr.2: 27498964-27498965 on Build GRCh38
rs183223793 Chr.2: 27498854-27498854 on Build GRCh38
rs1009741104 Chr.2: 27499026-27499026 on Build GRCh38
rs746369006 Chr.2: 27498690-27498690 on Build GRCh38
rs752372078 Chr.2: 27498793-27498793 on Build GRCh38
rs187988742 Chr.2: 27498917-27498917 on Build GRCh38
rs747016712 Chr.2: 27498826-27498826 on Build GRCh38
rs1010548025 Chr.2: 27499102-27499102 on Build GRCh38
rs903590942 Chr.2: 27499064-27499064 on Build GRCh38
rs745834393 Chr.2: 27498680-27498696 on Build GRCh38
rs769977399 Chr.2: 27499108-27499108 on Build GRCh38
rs767120253 Chr.2: 27498790-27498790 on Build GRCh38
rs769652451 Chr.2: 27498743-27498743 on Build GRCh38
rs772180243 Chr.2: 27498719-27498719 on Build GRCh38
rs780174034 Chr.2: 27498703-27498703 on Build GRCh38
rs772929772 Chr.2: 27498759-27498759 on Build GRCh38
rs1013433945 Chr.2: 27499090-27499090 on Build GRCh38
rs543556629 Chr.2: 27498966-27498966 on Build GRCh38
rs1040082445 Chr.2: 27499024-27499024 on Build GRCh38
rs766286371 Chr.2: 27498767-27498767 on Build GRCh38
rs773047068 Chr.2: 27499109-27499109 on Build GRCh38
rs748380298 Chr.2: 27499105-27499105 on Build GRCh38
rs1042101999 Chr.2: 27499039-27499039 on Build GRCh38
rs547476053 Chr.2: 27499086-27499086 on Build GRCh38
rs776639343 Chr.2: 27498994-27498994 on Build GRCh38
rs566932813 Chr.2: 27498863-27498863 on Build GRCh38
rs772492815 Chr.2: 27498697-27498697 on Build GRCh38
rs760970827 Chr.2: 27498965-27498965 on Build GRCh38
rs901480775 Chr.2: 27499025-27499025 on Build GRCh38
rs779242575 Chr.2: 27498688-27498688 on Build GRCh38

View Full Product Details