Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 36842095-36842612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRN
Entrez Gene ID
6801
Gene Name
striatin
Gene Aliases
PPP2R6A, SG2NA
UniGene
Hs.127486
Gene Chromosome Location
Chr.2: 36837698-36966607 on Build GRCh38
Transcripts
NM_003162.3 XM_005264519.4 XM_011533073.2

SNP Detail

SNP ID Chromosome Location
rs1003679871 Chr.2: 36842411-36842411 on Build GRCh38
rs966939813 Chr.2: 36842164-36842164 on Build GRCh38
rs966907017 Chr.2: 36842376-36842376 on Build GRCh38
rs781536374 Chr.2: 36842480-36842480 on Build GRCh38
rs540074000 Chr.2: 36842179-36842179 on Build GRCh38
rs966281309 Chr.2: 36842521-36842521 on Build GRCh38
rs978783574 Chr.2: 36842176-36842176 on Build GRCh38
rs555947726 Chr.2: 36842329-36842329 on Build GRCh38
rs1045521820 Chr.2: 36842238-36842238 on Build GRCh38
rs910600467 Chr.2: 36842512-36842512 on Build GRCh38
rs1004084549 Chr.2: 36842291-36842291 on Build GRCh38
rs1035590168 Chr.2: 36842443-36842443 on Build GRCh38
rs1033015905 Chr.2: 36842426-36842426 on Build GRCh38
rs1055180896 Chr.2: 36842193-36842194 on Build GRCh38
rs576912360 Chr.2: 36842242-36842242 on Build GRCh38
rs187540455 Chr.2: 36842128-36842128 on Build GRCh38
rs558376063 Chr.2: 36842191-36842191 on Build GRCh38
rs898129334 Chr.2: 36842301-36842301 on Build GRCh38
rs1022331849 Chr.2: 36842489-36842489 on Build GRCh38
rs939866507 Chr.2: 36842517-36842517 on Build GRCh38
rs574392849 Chr.2: 36842404-36842404 on Build GRCh38
rs560046087 Chr.2: 36842439-36842439 on Build GRCh38
rs1022642260 Chr.2: 36842364-36842364 on Build GRCh38
rs1040698493 Chr.2: 36842212-36842212 on Build GRCh38
rs117708697 Chr.2: 36842308-36842308 on Build GRCh38
rs986200389 Chr.2: 36842188-36842188 on Build GRCh38
rs954167318 Chr.2: 36842130-36842130 on Build GRCh38
rs199577188 Chr.2: 36842120-36842120 on Build GRCh38
rs948530077 Chr.2: 36842203-36842203 on Build GRCh38
rs988174543 Chr.2: 36842136-36842136 on Build GRCh38
rs953757841 Chr.2: 36842343-36842343 on Build GRCh38
rs541398512 Chr.2: 36842419-36842419 on Build GRCh38
rs907072135 Chr.2: 36842264-36842264 on Build GRCh38
rs545631535 Chr.2: 36842478-36842478 on Build GRCh38
rs975978232 Chr.2: 36842527-36842527 on Build GRCh38
rs1036923954 Chr.2: 36842547-36842547 on Build GRCh38
rs531355302 Chr.2: 36842532-36842532 on Build GRCh38
rs931269772 Chr.2: 36842576-36842576 on Build GRCh38
rs986985069 Chr.2: 36842137-36842137 on Build GRCh38
rs897710295 Chr.2: 36842292-36842292 on Build GRCh38
rs867939369 Chr.2: 36842182-36842182 on Build GRCh38
rs995595974 Chr.2: 36842309-36842309 on Build GRCh38
rs929219790 Chr.2: 36842305-36842305 on Build GRCh38
rs2717511 Chr.2: 36842507-36842507 on Build GRCh38
rs528658412 Chr.2: 36842234-36842234 on Build GRCh38
rs539317399 Chr.2: 36842552-36842552 on Build GRCh38
rs549720240 Chr.2: 36842482-36842482 on Build GRCh38
rs905360509 Chr.2: 36842367-36842367 on Build GRCh38
rs2717512 Chr.2: 36842177-36842177 on Build GRCh38
rs960111260 Chr.2: 36842122-36842122 on Build GRCh38
rs936670806 Chr.2: 36842183-36842183 on Build GRCh38
rs949457401 Chr.2: 36842495-36842495 on Build GRCh38
rs547220125 Chr.2: 36842254-36842254 on Build GRCh38
rs944534035 Chr.2: 36842201-36842201 on Build GRCh38
rs986231435 Chr.2: 36842511-36842511 on Build GRCh38
rs571992229 Chr.2: 36842459-36842459 on Build GRCh38
rs550256757 Chr.2: 36842568-36842568 on Build GRCh38
rs887426162 Chr.2: 36842322-36842322 on Build GRCh38
rs554518358 Chr.2: 36842163-36842163 on Build GRCh38
rs1008010814 Chr.2: 36842346-36842346 on Build GRCh38

View Full Product Details