Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37092081-37092578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPATCH11
Entrez Gene ID
253635
Gene Name
G-patch domain containing 11
Gene Aliases
CCDC75, CENP-Y, CENPY
UniGene
Hs.595250
Gene Chromosome Location
Chr.2: 37084451-37099244 on Build GRCh38
Transcripts
NM_001278505.1 NM_001322249.1 NM_174931.3 XM_017003748.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1020683 c.142A>C p.N48H Chr.2: 37092178-37092178 on Build GRCh38
3991294 c.157A>T p.M53L Chr.2: 37092193-37092193 on Build GRCh38
477378 c.438-1G>C p.? Chr.2: 37092164-37092164 on Build GRCh38
3991292 c.156A>C p.E52D Chr.2: 37092192-37092192 on Build GRCh38
3991293 c.466A>T p.M156L Chr.2: 37092193-37092193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951817647 Chr.2: 37092374-37092375 on Build GRCh38
rs905919978 Chr.2: 37092326-37092326 on Build GRCh38
rs200868969 Chr.2: 37092226-37092226 on Build GRCh38
rs755759267 Chr.2: 37092249-37092249 on Build GRCh38
rs924858604 Chr.2: 37092491-37092491 on Build GRCh38
rs1054640938 Chr.2: 37092323-37092323 on Build GRCh38
rs550309990 Chr.2: 37092294-37092294 on Build GRCh38
rs760518642 Chr.2: 37092199-37092199 on Build GRCh38
rs368200096 Chr.2: 37092285-37092285 on Build GRCh38
rs200535916 Chr.2: 37092264-37092264 on Build GRCh38
rs1028737983 Chr.2: 37092373-37092373 on Build GRCh38
rs1023085606 Chr.2: 37092483-37092484 on Build GRCh38
rs1053303251 Chr.2: 37092436-37092437 on Build GRCh38
rs186791223 Chr.2: 37092140-37092140 on Build GRCh38
rs372946346 Chr.2: 37092147-37092147 on Build GRCh38
rs764462883 Chr.2: 37092442-37092442 on Build GRCh38
rs34670386 Chr.2: 37092425-37092425 on Build GRCh38
rs934796170 Chr.2: 37092428-37092428 on Build GRCh38
rs917561003 Chr.2: 37092475-37092475 on Build GRCh38
rs1053084221 Chr.2: 37092321-37092321 on Build GRCh38
rs767679691 Chr.2: 37092398-37092399 on Build GRCh38
rs181576679 Chr.2: 37092533-37092533 on Build GRCh38
rs775240308 Chr.2: 37092196-37092196 on Build GRCh38
rs150293487 Chr.2: 37092178-37092178 on Build GRCh38
rs756383466 Chr.2: 37092156-37092156 on Build GRCh38
rs200209016 Chr.2: 37092219-37092219 on Build GRCh38
rs375363032 Chr.2: 37092170-37092170 on Build GRCh38
rs988911648 Chr.2: 37092537-37092538 on Build GRCh38
rs202239979 Chr.2: 37092353-37092356 on Build GRCh38
rs959114330 Chr.2: 37092458-37092458 on Build GRCh38
rs201245223 Chr.2: 37092227-37092227 on Build GRCh38
rs752352489 Chr.2: 37092204-37092206 on Build GRCh38
rs556576754 Chr.2: 37092158-37092158 on Build GRCh38
rs200303273 Chr.2: 37092399-37092399 on Build GRCh38
rs934906736 Chr.2: 37092496-37092496 on Build GRCh38
rs1002957660 Chr.2: 37092469-37092470 on Build GRCh38
rs767011978 Chr.2: 37092147-37092148 on Build GRCh38
rs764606777 Chr.2: 37092211-37092211 on Build GRCh38
rs1008546864 Chr.2: 37092417-37092417 on Build GRCh38
rs781656617 Chr.2: 37092145-37092145 on Build GRCh38
rs763781128 Chr.2: 37092116-37092116 on Build GRCh38
rs776083710 Chr.2: 37092205-37092205 on Build GRCh38
rs746955847 Chr.2: 37092288-37092288 on Build GRCh38
rs893191296 Chr.2: 37092338-37092338 on Build GRCh38
rs1044390331 Chr.2: 37092462-37092462 on Build GRCh38
rs996935708 Chr.2: 37092365-37092365 on Build GRCh38
rs745749278 Chr.2: 37092181-37092181 on Build GRCh38
rs139271234 Chr.2: 37092456-37092456 on Build GRCh38
rs1011539467 Chr.2: 37092324-37092325 on Build GRCh38
rs534419000 Chr.2: 37092474-37092474 on Build GRCh38
rs9678952 Chr.2: 37092393-37092393 on Build GRCh38
rs529793376 Chr.2: 37092318-37092318 on Build GRCh38
rs1023016744 Chr.2: 37092152-37092152 on Build GRCh38
rs905968659 Chr.2: 37092465-37092465 on Build GRCh38
rs1057201770 Chr.2: 37092471-37092471 on Build GRCh38
rs934792046 Chr.2: 37092322-37092322 on Build GRCh38
rs778823046 Chr.2: 37092278-37092278 on Build GRCh38
rs369210095 Chr.2: 37092128-37092128 on Build GRCh38
rs753379041 Chr.2: 37092117-37092117 on Build GRCh38
rs950341134 Chr.2: 37092315-37092315 on Build GRCh38
rs893262430 Chr.2: 37092134-37092134 on Build GRCh38
rs961219745 Chr.2: 37092259-37092259 on Build GRCh38
rs976259828 Chr.2: 37092416-37092416 on Build GRCh38
rs777250366 Chr.2: 37092298-37092299 on Build GRCh38
rs201570743 Chr.2: 37092209-37092209 on Build GRCh38
rs7568925 Chr.2: 37092523-37092523 on Build GRCh38
rs200409208 Chr.2: 37092397-37092402 on Build GRCh38
rs914807002 Chr.2: 37092443-37092444 on Build GRCh38
rs978178874 Chr.2: 37092490-37092490 on Build GRCh38
rs564857707 Chr.2: 37092265-37092265 on Build GRCh38
rs756788525 Chr.2: 37092124-37092124 on Build GRCh38
rs1044894650 Chr.2: 37092325-37092326 on Build GRCh38
rs7582985 Chr.2: 37092485-37092485 on Build GRCh38
rs35286617 Chr.2: 37092432-37092433 on Build GRCh38
rs763734220 Chr.2: 37092200-37092200 on Build GRCh38
rs745994994 Chr.2: 37092284-37092284 on Build GRCh38
rs751956124 Chr.2: 37092129-37092129 on Build GRCh38
rs778212418 Chr.2: 37092245-37092245 on Build GRCh38
rs973897851 Chr.2: 37092260-37092260 on Build GRCh38
rs964475463 Chr.2: 37092411-37092411 on Build GRCh38
rs768188612 Chr.2: 37092222-37092222 on Build GRCh38
rs903331529 Chr.2: 37092319-37092319 on Build GRCh38
rs755512195 Chr.2: 37092220-37092220 on Build GRCh38
rs1034835777 Chr.2: 37092340-37092340 on Build GRCh38
rs1015832919 Chr.2: 37092240-37092240 on Build GRCh38
rs189982802 Chr.2: 37092314-37092314 on Build GRCh38
rs772033547 Chr.2: 37092193-37092193 on Build GRCh38
rs570277160 Chr.2: 37092413-37092414 on Build GRCh38
rs147701297 Chr.2: 37092490-37092491 on Build GRCh38
rs1026193905 Chr.2: 37092367-37092370 on Build GRCh38
rs903385097 Chr.2: 37092132-37092132 on Build GRCh38
rs902074032 Chr.2: 37092112-37092112 on Build GRCh38
rs961291924 Chr.2: 37092435-37092435 on Build GRCh38
rs574837250 Chr.2: 37092169-37092169 on Build GRCh38
rs17446088 Chr.2: 37092524-37092524 on Build GRCh38

View Full Product Details