Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37352669-37353190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QPCT
Entrez Gene ID
25797
Gene Name
glutaminyl-peptide cyclotransferase
Gene Aliases
GCT, QC, sQC
UniGene
Hs.79033
Gene Chromosome Location
Chr.2: 37344610-37373322 on Build GRCh38
Transcripts
NM_012413.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5697072 c.121-3C>A p.? Chr.2: 37352786-37352786 on Build GRCh38
4094128 c.197G>T p.W66L Chr.2: 37352865-37352865 on Build GRCh38
1020774 c.236C>T p.P79L Chr.2: 37352904-37352904 on Build GRCh38
1020773 c.157C>T p.L53F Chr.2: 37352825-37352825 on Build GRCh38
4922002 c.267+1G>T p.? Chr.2: 37352936-37352936 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551802910 Chr.2: 37353012-37353012 on Build GRCh38
rs531317804 Chr.2: 37353124-37353124 on Build GRCh38
rs760504765 Chr.2: 37352902-37352902 on Build GRCh38
rs1004378835 Chr.2: 37352746-37352746 on Build GRCh38
rs1019372648 Chr.2: 37353094-37353094 on Build GRCh38
rs779013710 Chr.2: 37352890-37352890 on Build GRCh38
rs753346034 Chr.2: 37352842-37352842 on Build GRCh38
rs1033347990 Chr.2: 37352999-37352999 on Build GRCh38
rs775678218 Chr.2: 37352807-37352807 on Build GRCh38
rs111911070 Chr.2: 37352847-37352847 on Build GRCh38
rs762332056 Chr.2: 37353095-37353095 on Build GRCh38
rs1029449608 Chr.2: 37352700-37352700 on Build GRCh38
rs767568498 Chr.2: 37352743-37352744 on Build GRCh38
rs754072109 Chr.2: 37352958-37352958 on Build GRCh38
rs373503809 Chr.2: 37352780-37352780 on Build GRCh38
rs986571572 Chr.2: 37353070-37353070 on Build GRCh38
rs756236958 Chr.2: 37352859-37352859 on Build GRCh38
rs936100307 Chr.2: 37353147-37353147 on Build GRCh38
rs926066335 Chr.2: 37353092-37353092 on Build GRCh38
rs926042736 Chr.2: 37352845-37352845 on Build GRCh38
rs45582034 Chr.2: 37353019-37353019 on Build GRCh38
rs755360593 Chr.2: 37352849-37352849 on Build GRCh38
rs770836291 Chr.2: 37352811-37352811 on Build GRCh38
rs749398177 Chr.2: 37352869-37352869 on Build GRCh38
rs759448122 Chr.2: 37352817-37352817 on Build GRCh38
rs771941225 Chr.2: 37352820-37352820 on Build GRCh38
rs199791703 Chr.2: 37352763-37352763 on Build GRCh38
rs962714829 Chr.2: 37352755-37352755 on Build GRCh38
rs755669481 Chr.2: 37352747-37352747 on Build GRCh38
rs755303563 Chr.2: 37352781-37352781 on Build GRCh38
rs977605926 Chr.2: 37353042-37353042 on Build GRCh38
rs527562470 Chr.2: 37352962-37352962 on Build GRCh38
rs538179292 Chr.2: 37352815-37352815 on Build GRCh38
rs895245310 Chr.2: 37352880-37352880 on Build GRCh38
rs745811511 Chr.2: 37352978-37352978 on Build GRCh38
rs768258721 Chr.2: 37352903-37352903 on Build GRCh38
rs953892726 Chr.2: 37353067-37353067 on Build GRCh38
rs966401178 Chr.2: 37353153-37353153 on Build GRCh38
rs562549146 Chr.2: 37352851-37352851 on Build GRCh38
rs967971944 Chr.2: 37353030-37353030 on Build GRCh38
rs566909534 Chr.2: 37352806-37352806 on Build GRCh38
rs368418728 Chr.2: 37352987-37352987 on Build GRCh38
rs1017123014 Chr.2: 37352944-37352944 on Build GRCh38
rs371477578 Chr.2: 37352883-37352883 on Build GRCh38
rs912263373 Chr.2: 37352821-37352821 on Build GRCh38
rs775311582 Chr.2: 37352824-37352824 on Build GRCh38
rs1028548215 Chr.2: 37353043-37353043 on Build GRCh38
rs138792210 Chr.2: 37352931-37352931 on Build GRCh38
rs542622681 Chr.2: 37352829-37352829 on Build GRCh38
rs748323569 Chr.2: 37352796-37352796 on Build GRCh38
rs2230299 Chr.2: 37352794-37352794 on Build GRCh38
rs549801891 Chr.2: 37353166-37353166 on Build GRCh38
rs769947960 Chr.2: 37352804-37352804 on Build GRCh38
rs374001521 Chr.2: 37352975-37352975 on Build GRCh38
rs958941658 Chr.2: 37352779-37352779 on Build GRCh38
rs780525868 Chr.2: 37352773-37352773 on Build GRCh38
rs752678220 Chr.2: 37352801-37352801 on Build GRCh38
rs371567409 Chr.2: 37352816-37352816 on Build GRCh38
rs758440493 Chr.2: 37352870-37352870 on Build GRCh38
rs138559116 Chr.2: 37352949-37352949 on Build GRCh38
rs750941905 Chr.2: 37352762-37352762 on Build GRCh38
rs1024537984 Chr.2: 37353027-37353027 on Build GRCh38
rs368354793 Chr.2: 37352866-37352866 on Build GRCh38
rs779065001 Chr.2: 37352977-37352977 on Build GRCh38
rs1034635286 Chr.2: 37352769-37352769 on Build GRCh38
rs373504469 Chr.2: 37352918-37352918 on Build GRCh38
rs763468281 Chr.2: 37352754-37352754 on Build GRCh38
rs745758452 Chr.2: 37352893-37352893 on Build GRCh38
rs764678443 Chr.2: 37352924-37352924 on Build GRCh38
rs775363073 Chr.2: 37352897-37352897 on Build GRCh38
rs781496429 Chr.2: 37352850-37352850 on Build GRCh38
rs776316210 Chr.2: 37352905-37352905 on Build GRCh38
rs764267461 Chr.2: 37352954-37352954 on Build GRCh38
rs572487855 Chr.2: 37352713-37352713 on Build GRCh38
rs755136495 Chr.2: 37353091-37353091 on Build GRCh38
rs2255991 Chr.2: 37352828-37352828 on Build GRCh38
rs774611688 Chr.2: 37352998-37352998 on Build GRCh38
rs541738835 Chr.2: 37352930-37352930 on Build GRCh38
rs771992497 Chr.2: 37352896-37352896 on Build GRCh38
rs200314701 Chr.2: 37352848-37352848 on Build GRCh38
rs200573148 Chr.2: 37352904-37352904 on Build GRCh38
rs201576426 Chr.2: 37352938-37352938 on Build GRCh38
rs953907428 Chr.2: 37352728-37352728 on Build GRCh38
rs960326571 Chr.2: 37353003-37353003 on Build GRCh38

View Full Product Details