Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37372992-37373510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QPCT
Entrez Gene ID
25797
Gene Name
glutaminyl-peptide cyclotransferase
Gene Aliases
GCT, QC, sQC
UniGene
Hs.79033
Gene Chromosome Location
Chr.2: 37344610-37373322 on Build GRCh38
Transcripts
NM_012413.3

SNP Detail

SNP ID Chromosome Location
rs922056384 Chr.2: 37373376-37373376 on Build GRCh38
rs903806704 Chr.2: 37373439-37373439 on Build GRCh38
rs983058256 Chr.2: 37373254-37373254 on Build GRCh38
rs376144867 Chr.2: 37373177-37373177 on Build GRCh38
rs575626265 Chr.2: 37373139-37373139 on Build GRCh38
rs1039906085 Chr.2: 37373255-37373255 on Build GRCh38
rs1039601103 Chr.2: 37373053-37373053 on Build GRCh38
rs538916852 Chr.2: 37373062-37373062 on Build GRCh38
rs1051821229 Chr.2: 37373404-37373404 on Build GRCh38
rs147500036 Chr.2: 37373128-37373128 on Build GRCh38
rs561082255 Chr.2: 37373321-37373321 on Build GRCh38
rs983255067 Chr.2: 37373108-37373108 on Build GRCh38
rs1043178147 Chr.2: 37373223-37373223 on Build GRCh38
rs6708310 Chr.2: 37373284-37373284 on Build GRCh38
rs1030003156 Chr.2: 37373054-37373054 on Build GRCh38
rs997100561 Chr.2: 37373359-37373359 on Build GRCh38
rs903758841 Chr.2: 37373153-37373154 on Build GRCh38
rs950415538 Chr.2: 37373431-37373431 on Build GRCh38
rs573367960 Chr.2: 37373406-37373406 on Build GRCh38
rs776201233 Chr.2: 37373202-37373202 on Build GRCh38
rs974978926 Chr.2: 37373375-37373375 on Build GRCh38
rs938627741 Chr.2: 37373167-37373167 on Build GRCh38
rs946118693 Chr.2: 37373199-37373199 on Build GRCh38
rs950474258 Chr.2: 37373063-37373063 on Build GRCh38
rs891843444 Chr.2: 37373187-37373187 on Build GRCh38
rs10724 Chr.2: 37373149-37373149 on Build GRCh38
rs145985159 Chr.2: 37373240-37373241 on Build GRCh38
rs1029945755 Chr.2: 37373370-37373370 on Build GRCh38
rs927207352 Chr.2: 37373166-37373166 on Build GRCh38
rs201100507 Chr.2: 37373241-37373241 on Build GRCh38
rs1007078044 Chr.2: 37373249-37373249 on Build GRCh38
rs6544069 Chr.2: 37373444-37373444 on Build GRCh38
rs946054419 Chr.2: 37373412-37373412 on Build GRCh38
rs1051774303 Chr.2: 37373100-37373100 on Build GRCh38
rs904673253 Chr.2: 37373246-37373246 on Build GRCh38
rs11555866 Chr.2: 37373094-37373094 on Build GRCh38
rs116604774 Chr.2: 37373484-37373484 on Build GRCh38
rs942270604 Chr.2: 37373339-37373340 on Build GRCh38
rs941896288 Chr.2: 37373134-37373134 on Build GRCh38
rs1054974133 Chr.2: 37373476-37373476 on Build GRCh38
rs933428636 Chr.2: 37373399-37373399 on Build GRCh38

View Full Product Details