Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 39717597-39718136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM178A
Entrez Gene ID
130733
Gene Name
transmembrane protein 178A
Gene Aliases
TMEM178
UniGene
Hs.40808
Gene Chromosome Location
Chr.2: 39665087-39717963 on Build GRCh38
Transcripts
NM_001167959.1 NM_152390.2 XM_005264144.1 XM_005264145.1 XM_017003370.1 XM_017003371.1

SNP Detail

SNP ID Chromosome Location
rs529924607 Chr.2: 39717719-39717719 on Build GRCh38
rs1011629848 Chr.2: 39717626-39717626 on Build GRCh38
rs1018246916 Chr.2: 39717741-39717741 on Build GRCh38
rs988808563 Chr.2: 39717815-39717815 on Build GRCh38
rs1035944870 Chr.2: 39717737-39717737 on Build GRCh38
rs754668085 Chr.2: 39717873-39717873 on Build GRCh38
rs192573399 Chr.2: 39717993-39717993 on Build GRCh38
rs188720081 Chr.2: 39717894-39717894 on Build GRCh38
rs950855767 Chr.2: 39717782-39717786 on Build GRCh38
rs569959185 Chr.2: 39717675-39717675 on Build GRCh38
rs1044755510 Chr.2: 39718100-39718100 on Build GRCh38
rs762136722 Chr.2: 39717655-39717655 on Build GRCh38
rs373283330 Chr.2: 39717973-39717974 on Build GRCh38
rs779649320 Chr.2: 39717825-39717825 on Build GRCh38
rs537834796 Chr.2: 39717703-39717703 on Build GRCh38
rs746759858 Chr.2: 39717824-39717824 on Build GRCh38
rs75590688 Chr.2: 39717794-39717794 on Build GRCh38
rs976699017 Chr.2: 39717956-39717956 on Build GRCh38
rs780766008 Chr.2: 39718004-39718004 on Build GRCh38
rs943848292 Chr.2: 39717940-39717940 on Build GRCh38
rs915779439 Chr.2: 39717887-39717887 on Build GRCh38
rs968800168 Chr.2: 39717869-39717869 on Build GRCh38
rs902965514 Chr.2: 39717939-39717939 on Build GRCh38
rs76892392 Chr.2: 39717796-39717796 on Build GRCh38
rs542829479 Chr.2: 39718083-39718083 on Build GRCh38
rs999062043 Chr.2: 39717671-39717671 on Build GRCh38
rs1011928466 Chr.2: 39717664-39717664 on Build GRCh38
rs183783693 Chr.2: 39717730-39717730 on Build GRCh38
rs532477047 Chr.2: 39717633-39717633 on Build GRCh38
rs1026440342 Chr.2: 39717779-39717782 on Build GRCh38
rs982180404 Chr.2: 39718025-39718025 on Build GRCh38
rs947282833 Chr.2: 39718048-39718048 on Build GRCh38
rs950828085 Chr.2: 39718017-39718017 on Build GRCh38
rs890737617 Chr.2: 39717678-39717678 on Build GRCh38
rs556562025 Chr.2: 39717851-39717851 on Build GRCh38
rs34580359 Chr.2: 39717788-39717789 on Build GRCh38
rs200054347 Chr.2: 39717782-39717782 on Build GRCh38
rs182116543 Chr.2: 39718039-39718039 on Build GRCh38
rs191570624 Chr.2: 39717653-39717653 on Build GRCh38
rs1017861067 Chr.2: 39717982-39717982 on Build GRCh38
rs368976995 Chr.2: 39717968-39717968 on Build GRCh38
rs767159091 Chr.2: 39718112-39718112 on Build GRCh38
rs956271346 Chr.2: 39717753-39717753 on Build GRCh38
rs899412169 Chr.2: 39717994-39717994 on Build GRCh38
rs1007700224 Chr.2: 39717969-39717970 on Build GRCh38
rs747953047 Chr.2: 39718109-39718109 on Build GRCh38
rs963421114 Chr.2: 39717779-39717780 on Build GRCh38
rs141875764 Chr.2: 39717848-39717848 on Build GRCh38
rs1048647610 Chr.2: 39717628-39717628 on Build GRCh38
rs369724230 Chr.2: 39717740-39717740 on Build GRCh38
rs888082205 Chr.2: 39718043-39718043 on Build GRCh38
rs779733942 Chr.2: 39717804-39717804 on Build GRCh38
rs751115379 Chr.2: 39717868-39717868 on Build GRCh38
rs397869093 Chr.2: 39717794-39717795 on Build GRCh38
rs548366966 Chr.2: 39717785-39717785 on Build GRCh38
rs553364109 Chr.2: 39717947-39717947 on Build GRCh38
rs186728692 Chr.2: 39717623-39717623 on Build GRCh38
rs548472775 Chr.2: 39717648-39717648 on Build GRCh38
rs758993516 Chr.2: 39717735-39717735 on Build GRCh38
rs60119951 Chr.2: 39718046-39718046 on Build GRCh38
rs973774451 Chr.2: 39717779-39717781 on Build GRCh38
rs750858596 Chr.2: 39717701-39717701 on Build GRCh38

View Full Product Details