Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 42764185-42764677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OXER1
Entrez Gene ID
165140
Gene Name
oxoeicosanoid receptor 1
Gene Aliases
GPCR, GPR170, TG1019
UniGene
Hs.168439
Gene Chromosome Location
Chr.2: 42762499-42764261 on Build GRCh38
Transcripts
NM_148962.4

SNP Detail

SNP ID Chromosome Location
rs1014677301 Chr.2: 42764443-42764443 on Build GRCh38
rs962272130 Chr.2: 42764455-42764455 on Build GRCh38
rs973184237 Chr.2: 42764426-42764426 on Build GRCh38
rs150819316 Chr.2: 42764326-42764326 on Build GRCh38
rs183848298 Chr.2: 42764635-42764635 on Build GRCh38
rs765531235 Chr.2: 42764221-42764221 on Build GRCh38
rs1044821051 Chr.2: 42764409-42764409 on Build GRCh38
rs914649054 Chr.2: 42764282-42764282 on Build GRCh38
rs953502745 Chr.2: 42764278-42764278 on Build GRCh38
rs887509830 Chr.2: 42764436-42764436 on Build GRCh38
rs977592109 Chr.2: 42764647-42764647 on Build GRCh38
rs572071306 Chr.2: 42764351-42764351 on Build GRCh38
rs1006024291 Chr.2: 42764438-42764438 on Build GRCh38
rs999015851 Chr.2: 42764289-42764289 on Build GRCh38
rs759631327 Chr.2: 42764215-42764215 on Build GRCh38
rs1027386773 Chr.2: 42764427-42764427 on Build GRCh38
rs760899231 Chr.2: 42764447-42764447 on Build GRCh38
rs923257904 Chr.2: 42764277-42764277 on Build GRCh38
rs1014813044 Chr.2: 42764231-42764231 on Build GRCh38
rs372357315 Chr.2: 42764360-42764360 on Build GRCh38
rs926047726 Chr.2: 42764553-42764553 on Build GRCh38
rs934839906 Chr.2: 42764614-42764615 on Build GRCh38
rs927888706 Chr.2: 42764460-42764460 on Build GRCh38
rs781169948 Chr.2: 42764502-42764502 on Build GRCh38
rs983276343 Chr.2: 42764453-42764453 on Build GRCh38
rs192426679 Chr.2: 42764283-42764283 on Build GRCh38
rs199791354 Chr.2: 42764572-42764572 on Build GRCh38
rs1053267571 Chr.2: 42764627-42764627 on Build GRCh38
rs547304533 Chr.2: 42764223-42764223 on Build GRCh38
rs563685009 Chr.2: 42764593-42764593 on Build GRCh38
rs867827817 Chr.2: 42764465-42764465 on Build GRCh38
rs1030658591 Chr.2: 42764513-42764513 on Build GRCh38
rs578091950 Chr.2: 42764404-42764404 on Build GRCh38
rs1008586612 Chr.2: 42764350-42764350 on Build GRCh38
rs986251099 Chr.2: 42764279-42764279 on Build GRCh38
rs539369957 Chr.2: 42764266-42764266 on Build GRCh38
rs769354030 Chr.2: 42764315-42764316 on Build GRCh38
rs201353687 Chr.2: 42764214-42764214 on Build GRCh38
rs573985192 Chr.2: 42764613-42764613 on Build GRCh38
rs976498036 Chr.2: 42764272-42764272 on Build GRCh38
rs770883225 Chr.2: 42764210-42764210 on Build GRCh38
rs939241210 Chr.2: 42764466-42764466 on Build GRCh38
rs953435492 Chr.2: 42764549-42764549 on Build GRCh38
rs1043148015 Chr.2: 42764510-42764510 on Build GRCh38
rs947562471 Chr.2: 42764310-42764310 on Build GRCh38
rs10194180 Chr.2: 42764328-42764328 on Build GRCh38
rs938910306 Chr.2: 42764347-42764347 on Build GRCh38
rs541550288 Chr.2: 42764636-42764636 on Build GRCh38
rs775477725 Chr.2: 42764394-42764394 on Build GRCh38
rs767647610 Chr.2: 42764376-42764376 on Build GRCh38
rs139326225 Chr.2: 42764338-42764338 on Build GRCh38
rs543708611 Chr.2: 42764550-42764550 on Build GRCh38
rs374249711 Chr.2: 42764208-42764208 on Build GRCh38
rs1009120463 Chr.2: 42764638-42764638 on Build GRCh38
rs979530612 Chr.2: 42764495-42764515 on Build GRCh38
rs992794117 Chr.2: 42764472-42764472 on Build GRCh38
rs371620646 Chr.2: 42764225-42764225 on Build GRCh38
rs755612218 Chr.2: 42764640-42764640 on Build GRCh38
rs376669259 Chr.2: 42764372-42764372 on Build GRCh38
rs570599775 Chr.2: 42764262-42764262 on Build GRCh38
rs760758899 Chr.2: 42764209-42764209 on Build GRCh38
rs764251401 Chr.2: 42764483-42764483 on Build GRCh38
rs753696160 Chr.2: 42764592-42764592 on Build GRCh38

View Full Product Details