Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 43694822-43695346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHH2
Entrez Gene ID
130271
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H2
Gene Aliases
PLEKHH1L
UniGene
Hs.164162
Gene Chromosome Location
Chr.2: 43637300-43767987 on Build GRCh38
Transcripts
NM_172069.3 XM_017003351.1 XM_017003352.1 XM_017003353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6413481 c.452T>C p.I151T Chr.2: 43695174-43695174 on Build GRCh38
4094290 c.446G>A p.R149H Chr.2: 43695168-43695168 on Build GRCh38
5397041 c.445C>T p.R149C Chr.2: 43695167-43695167 on Build GRCh38
5994008 c.491C>G p.S164* Chr.2: 43695213-43695213 on Build GRCh38
4918620 c.434A>G p.N145S Chr.2: 43695156-43695156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557448022 Chr.2: 43694982-43694982 on Build GRCh38
rs534418099 Chr.2: 43695292-43695292 on Build GRCh38
rs773944727 Chr.2: 43695137-43695137 on Build GRCh38
rs753775265 Chr.2: 43695177-43695177 on Build GRCh38
rs758712636 Chr.2: 43694958-43694958 on Build GRCh38
rs980856797 Chr.2: 43695043-43695043 on Build GRCh38
rs186057112 Chr.2: 43695010-43695010 on Build GRCh38
rs534624759 Chr.2: 43694977-43694977 on Build GRCh38
rs747683074 Chr.2: 43695269-43695269 on Build GRCh38
rs748275230 Chr.2: 43695107-43695107 on Build GRCh38
rs763167910 Chr.2: 43695205-43695206 on Build GRCh38
rs1008080106 Chr.2: 43694935-43694935 on Build GRCh38
rs578230169 Chr.2: 43694999-43694999 on Build GRCh38
rs763985353 Chr.2: 43695232-43695236 on Build GRCh38
rs767310368 Chr.2: 43695210-43695211 on Build GRCh38
rs772801775 Chr.2: 43695163-43695163 on Build GRCh38
rs766233846 Chr.2: 43695275-43695275 on Build GRCh38
rs565609319 Chr.2: 43694964-43694964 on Build GRCh38
rs772304686 Chr.2: 43695108-43695108 on Build GRCh38
rs956680277 Chr.2: 43695315-43695315 on Build GRCh38
rs756108815 Chr.2: 43695214-43695215 on Build GRCh38
rs559813938 Chr.2: 43695106-43695106 on Build GRCh38
rs1015576074 Chr.2: 43695189-43695189 on Build GRCh38
rs757561520 Chr.2: 43695234-43695234 on Build GRCh38
rs770257186 Chr.2: 43695248-43695248 on Build GRCh38
rs1029561094 Chr.2: 43694897-43694897 on Build GRCh38
rs918151476 Chr.2: 43695041-43695041 on Build GRCh38
rs573753563 Chr.2: 43694891-43694891 on Build GRCh38
rs771350983 Chr.2: 43695121-43695121 on Build GRCh38
rs1006316131 Chr.2: 43694863-43694863 on Build GRCh38
rs757502804 Chr.2: 43694872-43694872 on Build GRCh38
rs761272172 Chr.2: 43695146-43695146 on Build GRCh38
rs925240821 Chr.2: 43695048-43695048 on Build GRCh38
rs892266265 Chr.2: 43694932-43694933 on Build GRCh38
rs778052531 Chr.2: 43695208-43695208 on Build GRCh38
rs768108023 Chr.2: 43695128-43695128 on Build GRCh38
rs146121451 Chr.2: 43695180-43695180 on Build GRCh38
rs899614438 Chr.2: 43694957-43694957 on Build GRCh38
rs746044740 Chr.2: 43694956-43694956 on Build GRCh38
rs908000190 Chr.2: 43694856-43694856 on Build GRCh38
rs992370315 Chr.2: 43695027-43695027 on Build GRCh38
rs776533319 Chr.2: 43695276-43695276 on Build GRCh38
rs780262669 Chr.2: 43694976-43694976 on Build GRCh38
rs781559751 Chr.2: 43695243-43695243 on Build GRCh38
rs202191918 Chr.2: 43695199-43695199 on Build GRCh38
rs755046870 Chr.2: 43695092-43695092 on Build GRCh38
rs747200102 Chr.2: 43695216-43695216 on Build GRCh38
rs78192773 Chr.2: 43695271-43695271 on Build GRCh38
rs955304127 Chr.2: 43694904-43694904 on Build GRCh38
rs73923833 Chr.2: 43695074-43695074 on Build GRCh38
rs777904439 Chr.2: 43695113-43695113 on Build GRCh38
rs190889990 Chr.2: 43695270-43695270 on Build GRCh38
rs1031066874 Chr.2: 43694981-43694981 on Build GRCh38
rs746336872 Chr.2: 43695245-43695245 on Build GRCh38
rs762903046 Chr.2: 43694860-43694860 on Build GRCh38
rs913843405 Chr.2: 43695097-43695097 on Build GRCh38
rs888087928 Chr.2: 43695166-43695166 on Build GRCh38
rs114160011 Chr.2: 43695042-43695042 on Build GRCh38
rs200546769 Chr.2: 43695123-43695123 on Build GRCh38
rs985010494 Chr.2: 43694854-43694854 on Build GRCh38
rs942104343 Chr.2: 43695112-43695112 on Build GRCh38
rs925377022 Chr.2: 43695281-43695281 on Build GRCh38
rs991838814 Chr.2: 43695015-43695015 on Build GRCh38
rs752472091 Chr.2: 43695212-43695213 on Build GRCh38
rs994470621 Chr.2: 43694895-43694895 on Build GRCh38
rs747385713 Chr.2: 43695118-43695118 on Build GRCh38
rs747128829 Chr.2: 43695070-43695070 on Build GRCh38
rs555502420 Chr.2: 43694920-43694920 on Build GRCh38
rs767178694 Chr.2: 43695149-43695149 on Build GRCh38
rs137890410 Chr.2: 43695088-43695088 on Build GRCh38
rs1015192815 Chr.2: 43695007-43695008 on Build GRCh38
rs1006398828 Chr.2: 43694985-43694985 on Build GRCh38
rs891003271 Chr.2: 43695179-43695179 on Build GRCh38
rs1052371454 Chr.2: 43694929-43694929 on Build GRCh38
rs773337311 Chr.2: 43695189-43695191 on Build GRCh38
rs943860175 Chr.2: 43695098-43695098 on Build GRCh38
rs533305612 Chr.2: 43695197-43695197 on Build GRCh38
rs760048923 Chr.2: 43695125-43695125 on Build GRCh38
rs758464342 Chr.2: 43695207-43695207 on Build GRCh38
rs190164545 Chr.2: 43695168-43695168 on Build GRCh38
rs186490151 Chr.2: 43695267-43695267 on Build GRCh38
rs569856373 Chr.2: 43694857-43694857 on Build GRCh38
rs940880144 Chr.2: 43694871-43694871 on Build GRCh38
rs181193760 Chr.2: 43695196-43695196 on Build GRCh38
rs1048143573 Chr.2: 43695164-43695164 on Build GRCh38
rs374128785 Chr.2: 43695167-43695167 on Build GRCh38
rs182219133 Chr.2: 43695272-43695272 on Build GRCh38
rs267599379 Chr.2: 43695194-43695194 on Build GRCh38

View Full Product Details