Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61073342-61073869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1841
Entrez Gene ID
84542
Gene Name
KIAA1841
Gene Aliases
-
UniGene
Hs.468653
Gene Chromosome Location
Chr.2: 61065870-61157942 on Build GRCh38
Transcripts
NM_001129993.1 NM_032506.2 XM_005264605.3 XM_005264606.4 XM_005264607.2 XM_011533126.2 XM_011533127.2 XM_011533128.1 XM_011533129.2 XM_011533131.2 XM_017005108.1 XM_017005109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6277160 c.343C>A p.H115N Chr.2: 61073463-61073463 on Build GRCh38
6509401 c.357C>A p.S119S Chr.2: 61073477-61073477 on Build GRCh38
3582468 c.414G>C p.M138I Chr.2: 61073534-61073534 on Build GRCh38
1615000 c.338-7A>G p.? Chr.2: 61073451-61073451 on Build GRCh38
3582467 c.414G>C p.M138I Chr.2: 61073534-61073534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372285850 Chr.2: 61073469-61073469 on Build GRCh38
rs999620048 Chr.2: 61073474-61073474 on Build GRCh38
rs1007706905 Chr.2: 61073839-61073839 on Build GRCh38
rs902470321 Chr.2: 61073785-61073785 on Build GRCh38
rs187150019 Chr.2: 61073795-61073795 on Build GRCh38
rs746598627 Chr.2: 61073570-61073570 on Build GRCh38
rs773920303 Chr.2: 61073629-61073629 on Build GRCh38
rs1013340791 Chr.2: 61073402-61073402 on Build GRCh38
rs775425270 Chr.2: 61073601-61073601 on Build GRCh38
rs770371750 Chr.2: 61073457-61073457 on Build GRCh38
rs893476206 Chr.2: 61073820-61073820 on Build GRCh38
rs892122450 Chr.2: 61073710-61073710 on Build GRCh38
rs377736645 Chr.2: 61073414-61073414 on Build GRCh38
rs770460024 Chr.2: 61073577-61073577 on Build GRCh38
rs769656330 Chr.2: 61073468-61073468 on Build GRCh38
rs954850143 Chr.2: 61073373-61073373 on Build GRCh38
rs751021643 Chr.2: 61073413-61073418 on Build GRCh38
rs144280405 Chr.2: 61073465-61073465 on Build GRCh38
rs1009857296 Chr.2: 61073752-61073752 on Build GRCh38
rs1055142728 Chr.2: 61073699-61073699 on Build GRCh38
rs142573176 Chr.2: 61073557-61073557 on Build GRCh38
rs745869228 Chr.2: 61073433-61073437 on Build GRCh38
rs780515243 Chr.2: 61073518-61073518 on Build GRCh38
rs571645694 Chr.2: 61073713-61073713 on Build GRCh38
rs368466121 Chr.2: 61073422-61073422 on Build GRCh38
rs1032463467 Chr.2: 61073806-61073806 on Build GRCh38
rs1018116382 Chr.2: 61073841-61073841 on Build GRCh38
rs758960766 Chr.2: 61073419-61073420 on Build GRCh38
rs926365121 Chr.2: 61073696-61073696 on Build GRCh38
rs553484841 Chr.2: 61073844-61073844 on Build GRCh38
rs983891265 Chr.2: 61073634-61073634 on Build GRCh38
rs1051981670 Chr.2: 61073792-61073792 on Build GRCh38
rs771600975 Chr.2: 61073443-61073443 on Build GRCh38
rs780634212 Chr.2: 61073585-61073585 on Build GRCh38
rs781760364 Chr.2: 61073432-61073432 on Build GRCh38
rs1023495568 Chr.2: 61073419-61073419 on Build GRCh38
rs1046198851 Chr.2: 61073744-61073744 on Build GRCh38
rs754119705 Chr.2: 61073527-61073527 on Build GRCh38
rs949237587 Chr.2: 61073743-61073743 on Build GRCh38
rs549526710 Chr.2: 61073650-61073650 on Build GRCh38
rs909211901 Chr.2: 61073649-61073649 on Build GRCh38
rs777303339 Chr.2: 61073451-61073451 on Build GRCh38
rs774118702 Chr.2: 61073433-61073439 on Build GRCh38
rs976220041 Chr.2: 61073611-61073611 on Build GRCh38
rs747065298 Chr.2: 61073421-61073422 on Build GRCh38
rs756527472 Chr.2: 61073514-61073514 on Build GRCh38
rs763280522 Chr.2: 61073473-61073473 on Build GRCh38
rs771998743 Chr.2: 61073522-61073524 on Build GRCh38
rs1053480619 Chr.2: 61073701-61073701 on Build GRCh38
rs140011996 Chr.2: 61073487-61073487 on Build GRCh38
rs780238706 Chr.2: 61073437-61073437 on Build GRCh38
rs1024863371 Chr.2: 61073424-61073424 on Build GRCh38
rs756504163 Chr.2: 61073435-61073435 on Build GRCh38
rs191152908 Chr.2: 61073822-61073822 on Build GRCh38
rs747858582 Chr.2: 61073439-61073439 on Build GRCh38
rs7600064 Chr.2: 61073615-61073615 on Build GRCh38
rs755029314 Chr.2: 61073422-61073423 on Build GRCh38
rs894986305 Chr.2: 61073372-61073372 on Build GRCh38
rs535178756 Chr.2: 61073721-61073721 on Build GRCh38
rs936405135 Chr.2: 61073700-61073700 on Build GRCh38
rs200687620 Chr.2: 61073408-61073408 on Build GRCh38
rs895203795 Chr.2: 61073729-61073729 on Build GRCh38
rs758135730 Chr.2: 61073545-61073545 on Build GRCh38
rs769590213 Chr.2: 61073595-61073595 on Build GRCh38
rs151251854 Chr.2: 61073509-61073509 on Build GRCh38
rs750997096 Chr.2: 61073511-61073511 on Build GRCh38
rs568119181 Chr.2: 61073655-61073655 on Build GRCh38
rs369275822 Chr.2: 61073591-61073591 on Build GRCh38
rs750898304 Chr.2: 61073433-61073433 on Build GRCh38
rs751743350 Chr.2: 61073507-61073507 on Build GRCh38
rs750001932 Chr.2: 61073627-61073627 on Build GRCh38
rs779599375 Chr.2: 61073630-61073630 on Build GRCh38
rs537808938 Chr.2: 61073618-61073618 on Build GRCh38

View Full Product Details