Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73220583-73221086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMYD5
Entrez Gene ID
10322
Gene Name
SMYD family member 5
Gene Aliases
NN8-4AG, RAI15, RRG1, ZMYND23
UniGene
Hs.631882
Gene Chromosome Location
Chr.2: 73214238-73227227 on Build GRCh38
Transcripts
NM_006062.2 XM_006711918.3 XM_017003163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2998178 c.74A>T p.Q25L Chr.2: 73220737-73220737 on Build GRCh38
6394069 c.3G>A p.M1I Chr.2: 73220666-73220666 on Build GRCh38
6362783 c.76G>T p.D26Y Chr.2: 73220739-73220739 on Build GRCh38
4493104 c.409C>T p.P137S Chr.2: 73220724-73220724 on Build GRCh38
1306981 c.417C>T p.P139P Chr.2: 73220732-73220732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982878584 Chr.2: 73220873-73220873 on Build GRCh38
rs1053931691 Chr.2: 73220999-73220999 on Build GRCh38
rs957079494 Chr.2: 73220800-73220800 on Build GRCh38
rs769658721 Chr.2: 73220738-73220738 on Build GRCh38
rs779193621 Chr.2: 73220840-73220840 on Build GRCh38
rs200859633 Chr.2: 73220611-73220611 on Build GRCh38
rs145758484 Chr.2: 73220858-73220858 on Build GRCh38
rs748979436 Chr.2: 73220747-73220747 on Build GRCh38
rs779504329 Chr.2: 73220742-73220742 on Build GRCh38
rs999035730 Chr.2: 73221030-73221030 on Build GRCh38
rs753949407 Chr.2: 73220823-73220823 on Build GRCh38
rs750826999 Chr.2: 73220666-73220666 on Build GRCh38
rs183589161 Chr.2: 73221029-73221029 on Build GRCh38
rs138495170 Chr.2: 73220861-73220861 on Build GRCh38
rs372993095 Chr.2: 73221031-73221031 on Build GRCh38
rs996844971 Chr.2: 73220953-73220953 on Build GRCh38
rs768138578 Chr.2: 73220752-73220752 on Build GRCh38
rs192977995 Chr.2: 73220904-73220904 on Build GRCh38
rs141404445 Chr.2: 73220917-73220917 on Build GRCh38
rs763662007 Chr.2: 73220644-73220644 on Build GRCh38
rs777556624 Chr.2: 73220804-73220808 on Build GRCh38
rs372537885 Chr.2: 73220655-73220655 on Build GRCh38
rs989936153 Chr.2: 73221042-73221042 on Build GRCh38
rs754948317 Chr.2: 73220709-73220709 on Build GRCh38
rs1002243078 Chr.2: 73220814-73220814 on Build GRCh38
rs568161328 Chr.2: 73220784-73220785 on Build GRCh38
rs749312875 Chr.2: 73220809-73220812 on Build GRCh38
rs964334982 Chr.2: 73220635-73220635 on Build GRCh38
rs779104791 Chr.2: 73221015-73221015 on Build GRCh38
rs539085474 Chr.2: 73220761-73220761 on Build GRCh38
rs760909723 Chr.2: 73220804-73220804 on Build GRCh38
rs773916806 Chr.2: 73220774-73220774 on Build GRCh38
rs1002126937 Chr.2: 73221062-73221062 on Build GRCh38
rs762326751 Chr.2: 73220620-73220620 on Build GRCh38
rs759861211 Chr.2: 73220828-73220828 on Build GRCh38
rs139296342 Chr.2: 73220800-73220802 on Build GRCh38
rs1030349764 Chr.2: 73220979-73220979 on Build GRCh38
rs376245554 Chr.2: 73221059-73221059 on Build GRCh38
rs533184375 Chr.2: 73220957-73220957 on Build GRCh38
rs115495075 Chr.2: 73220775-73220775 on Build GRCh38
rs948408858 Chr.2: 73220857-73220857 on Build GRCh38
rs779146983 Chr.2: 73220714-73220714 on Build GRCh38
rs887206848 Chr.2: 73220885-73220885 on Build GRCh38
rs574417933 Chr.2: 73220882-73220882 on Build GRCh38
rs560429964 Chr.2: 73221017-73221017 on Build GRCh38
rs766462014 Chr.2: 73220695-73220695 on Build GRCh38
rs2303903 Chr.2: 73220705-73220705 on Build GRCh38
rs767862890 Chr.2: 73220659-73220659 on Build GRCh38
rs957109261 Chr.2: 73221034-73221034 on Build GRCh38
rs111928263 Chr.2: 73220811-73220811 on Build GRCh38
rs745702381 Chr.2: 73220735-73220735 on Build GRCh38
rs773728844 Chr.2: 73220796-73220796 on Build GRCh38
rs781106745 Chr.2: 73220730-73220730 on Build GRCh38
rs936843752 Chr.2: 73220982-73220982 on Build GRCh38
rs2303904 Chr.2: 73220647-73220647 on Build GRCh38
rs772615030 Chr.2: 73220785-73220785 on Build GRCh38
rs369345161 Chr.2: 73220859-73220862 on Build GRCh38
rs760927097 Chr.2: 73220682-73220682 on Build GRCh38
rs899865059 Chr.2: 73220936-73220936 on Build GRCh38
rs201766809 Chr.2: 73220745-73220745 on Build GRCh38
rs757334870 Chr.2: 73220729-73220729 on Build GRCh38
rs751530227 Chr.2: 73220726-73220726 on Build GRCh38
rs545340866 Chr.2: 73220978-73220978 on Build GRCh38
rs2288632 Chr.2: 73221046-73221046 on Build GRCh38
rs2303905 Chr.2: 73220608-73220608 on Build GRCh38
rs1034181704 Chr.2: 73220815-73220815 on Build GRCh38
rs929157087 Chr.2: 73220881-73220881 on Build GRCh38

View Full Product Details