Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85578917-85579421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAMP8
Entrez Gene ID
8673
Gene Name
vesicle associated membrane protein 8
Gene Aliases
EDB, VAMP-8
UniGene
Hs.714302
Gene Chromosome Location
Chr.2: 85577491-85582033 on Build GRCh38
Transcripts
NM_003761.4 XM_017005170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1669112 c.118A>G p.N40D Chr.2: 85579123-85579123 on Build GRCh38
1023372 c.71A>C p.K24T Chr.2: 85579076-85579076 on Build GRCh38
148885 c.138C>T p.N46N Chr.2: 85579143-85579143 on Build GRCh38
1615177 c.98G>A p.R33Q Chr.2: 85579103-85579103 on Build GRCh38
6202979 c.110G>T p.R37L Chr.2: 85579115-85579115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912337396 Chr.2: 85579158-85579158 on Build GRCh38
rs753286789 Chr.2: 85579102-85579102 on Build GRCh38
rs778808980 Chr.2: 85578994-85578995 on Build GRCh38
rs547111500 Chr.2: 85579121-85579121 on Build GRCh38
rs539672042 Chr.2: 85579134-85579134 on Build GRCh38
rs774836084 Chr.2: 85579063-85579063 on Build GRCh38
rs1017850959 Chr.2: 85579358-85579358 on Build GRCh38
rs758348543 Chr.2: 85579003-85579003 on Build GRCh38
rs768755792 Chr.2: 85579046-85579046 on Build GRCh38
rs759295495 Chr.2: 85579103-85579103 on Build GRCh38
rs748551620 Chr.2: 85579133-85579133 on Build GRCh38
rs752480085 Chr.2: 85579002-85579002 on Build GRCh38
rs141492723 Chr.2: 85579126-85579126 on Build GRCh38
rs1042886872 Chr.2: 85579304-85579304 on Build GRCh38
rs757662072 Chr.2: 85578994-85578994 on Build GRCh38
rs760280599 Chr.2: 85579081-85579081 on Build GRCh38
rs772708181 Chr.2: 85579067-85579067 on Build GRCh38
rs752496672 Chr.2: 85579114-85579114 on Build GRCh38
rs765886382 Chr.2: 85579119-85579119 on Build GRCh38
rs764975250 Chr.2: 85579152-85579152 on Build GRCh38
rs1004367784 Chr.2: 85579395-85579395 on Build GRCh38
rs369422930 Chr.2: 85579040-85579040 on Build GRCh38
rs779208332 Chr.2: 85579132-85579132 on Build GRCh38
rs887446189 Chr.2: 85578946-85578946 on Build GRCh38
rs3731828 Chr.2: 85579143-85579143 on Build GRCh38
rs746524228 Chr.2: 85579204-85579205 on Build GRCh38
rs145569649 Chr.2: 85579282-85579282 on Build GRCh38
rs201135270 Chr.2: 85578998-85578998 on Build GRCh38
rs201681602 Chr.2: 85579092-85579092 on Build GRCh38
rs376281449 Chr.2: 85579149-85579149 on Build GRCh38
rs938517901 Chr.2: 85579203-85579203 on Build GRCh38
rs773686028 Chr.2: 85579136-85579136 on Build GRCh38
rs368631664 Chr.2: 85579115-85579115 on Build GRCh38
rs964990995 Chr.2: 85579369-85579369 on Build GRCh38
rs1044305477 Chr.2: 85579166-85579166 on Build GRCh38
rs143800390 Chr.2: 85579138-85579138 on Build GRCh38
rs771816128 Chr.2: 85579054-85579055 on Build GRCh38
rs1018338412 Chr.2: 85578941-85578941 on Build GRCh38
rs891638456 Chr.2: 85579230-85579230 on Build GRCh38
rs201988068 Chr.2: 85578964-85578964 on Build GRCh38
rs561356918 Chr.2: 85579039-85579039 on Build GRCh38
rs745503971 Chr.2: 85579018-85579018 on Build GRCh38
rs147504597 Chr.2: 85578974-85578974 on Build GRCh38
rs777193785 Chr.2: 85578985-85578985 on Build GRCh38
rs150845407 Chr.2: 85579128-85579128 on Build GRCh38
rs537363002 Chr.2: 85579246-85579246 on Build GRCh38
rs1009518011 Chr.2: 85579033-85579033 on Build GRCh38
rs966820699 Chr.2: 85579346-85579359 on Build GRCh38
rs1044722601 Chr.2: 85578955-85578955 on Build GRCh38
rs777623521 Chr.2: 85579034-85579034 on Build GRCh38
rs778839494 Chr.2: 85579000-85579000 on Build GRCh38
rs749965903 Chr.2: 85578971-85578971 on Build GRCh38
rs896616700 Chr.2: 85579018-85579018 on Build GRCh38
rs1001304016 Chr.2: 85579350-85579350 on Build GRCh38
rs941916026 Chr.2: 85578952-85578952 on Build GRCh38
rs771333013 Chr.2: 85579139-85579139 on Build GRCh38
rs751352802 Chr.2: 85579211-85579211 on Build GRCh38
rs773812719 Chr.2: 85579076-85579076 on Build GRCh38
rs185098390 Chr.2: 85579113-85579113 on Build GRCh38
rs947343816 Chr.2: 85579124-85579124 on Build GRCh38
rs774807701 Chr.2: 85579115-85579115 on Build GRCh38
rs749565473 Chr.2: 85579045-85579045 on Build GRCh38
rs941702590 Chr.2: 85579156-85579156 on Build GRCh38
rs927227965 Chr.2: 85579187-85579187 on Build GRCh38
rs3731827 Chr.2: 85578945-85578945 on Build GRCh38
rs555353141 Chr.2: 85579258-85579258 on Build GRCh38
rs780245621 Chr.2: 85579123-85579123 on Build GRCh38
rs140052824 Chr.2: 85579209-85579209 on Build GRCh38
rs762208919 Chr.2: 85579065-85579065 on Build GRCh38
rs775076519 Chr.2: 85579159-85579159 on Build GRCh38
rs780947233 Chr.2: 85579387-85579387 on Build GRCh38
rs903826065 Chr.2: 85579315-85579315 on Build GRCh38
rs1048666595 Chr.2: 85579307-85579307 on Build GRCh38
rs565732791 Chr.2: 85579122-85579122 on Build GRCh38
rs180708200 Chr.2: 85578995-85578995 on Build GRCh38

View Full Product Details