Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97084078-97084585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAHD2B
Entrez Gene ID
151313
Gene Name
fumarylacetoacetate hydrolase domain containing 2B
Gene Aliases
-
UniGene
Hs.567723
Gene Chromosome Location
Chr.2: 97080738-97094882 on Build GRCh38
Transcripts
NM_001320848.1 NM_199336.2 XM_011510745.2 XM_011510746.2 XM_011510747.2 XM_011510748.2 XM_011510750.2 XM_017003470.1 XM_017003471.1 XM_017003472.1 XM_017003473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1410121 c.709_711delTGC p.C237delC Chr.2: 97084252-97084254 on Build GRCh38
6092735 c.724G>T p.E242* Chr.2: 97084239-97084239 on Build GRCh38
363326 c.786G>T p.W262C Chr.2: 97084177-97084177 on Build GRCh38
722811 c.757G>A p.V253I Chr.2: 97084206-97084206 on Build GRCh38
6480698 c.736A>T p.S246C Chr.2: 97084227-97084227 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768151716 Chr.2: 97084158-97084158 on Build GRCh38
rs769493043 Chr.2: 97084250-97084250 on Build GRCh38
rs746551848 Chr.2: 97084418-97084418 on Build GRCh38
rs768371180 Chr.2: 97084326-97084326 on Build GRCh38
rs754060056 Chr.2: 97084174-97084174 on Build GRCh38
rs769317792 Chr.2: 97084328-97084328 on Build GRCh38
rs775549903 Chr.2: 97084251-97084251 on Build GRCh38
rs757040095 Chr.2: 97084261-97084261 on Build GRCh38
rs778003216 Chr.2: 97084121-97084122 on Build GRCh38
rs752426119 Chr.2: 97084295-97084295 on Build GRCh38
rs1031295197 Chr.2: 97084284-97084284 on Build GRCh38
rs754773697 Chr.2: 97084134-97084134 on Build GRCh38
rs756332304 Chr.2: 97084167-97084167 on Build GRCh38
rs534709393 Chr.2: 97084216-97084216 on Build GRCh38
rs746861856 Chr.2: 97084301-97084301 on Build GRCh38
rs745849716 Chr.2: 97084364-97084364 on Build GRCh38
rs762162971 Chr.2: 97084152-97084152 on Build GRCh38
rs745470134 Chr.2: 97084247-97084247 on Build GRCh38
rs766682338 Chr.2: 97084170-97084170 on Build GRCh38
rs558162125 Chr.2: 97084133-97084133 on Build GRCh38
rs745398719 Chr.2: 97084327-97084327 on Build GRCh38
rs745797387 Chr.2: 97084133-97084138 on Build GRCh38
rs755254004 Chr.2: 97084180-97084180 on Build GRCh38
rs778890641 Chr.2: 97084136-97084136 on Build GRCh38
rs557448239 Chr.2: 97084234-97084234 on Build GRCh38
rs372972282 Chr.2: 97084140-97084140 on Build GRCh38
rs886411293 Chr.2: 97084480-97084480 on Build GRCh38
rs757216055 Chr.2: 97084326-97084326 on Build GRCh38
rs540962995 Chr.2: 97084542-97084542 on Build GRCh38
rs541630279 Chr.2: 97084341-97084341 on Build GRCh38
rs779895937 Chr.2: 97084394-97084394 on Build GRCh38
rs143438262 Chr.2: 97084503-97084503 on Build GRCh38
rs375434642 Chr.2: 97084239-97084239 on Build GRCh38
rs955704740 Chr.2: 97084309-97084309 on Build GRCh38
rs145484440 Chr.2: 97084270-97084270 on Build GRCh38
rs933382633 Chr.2: 97084438-97084438 on Build GRCh38
rs1012725609 Chr.2: 97084156-97084156 on Build GRCh38
rs548829930 Chr.2: 97084233-97084233 on Build GRCh38
rs758226009 Chr.2: 97084297-97084297 on Build GRCh38
rs778502141 Chr.2: 97084199-97084199 on Build GRCh38
rs1048286417 Chr.2: 97084465-97084466 on Build GRCh38
rs778763923 Chr.2: 97084359-97084359 on Build GRCh38
rs758251307 Chr.2: 97084215-97084215 on Build GRCh38
rs777664545 Chr.2: 97084307-97084307 on Build GRCh38
rs772048794 Chr.2: 97084138-97084138 on Build GRCh38
rs563893801 Chr.2: 97084553-97084553 on Build GRCh38
rs774811524 Chr.2: 97084150-97084150 on Build GRCh38
rs773899048 Chr.2: 97084264-97084264 on Build GRCh38
rs751447150 Chr.2: 97084311-97084311 on Build GRCh38
rs778903553 Chr.2: 97084312-97084312 on Build GRCh38
rs866948096 Chr.2: 97084468-97084470 on Build GRCh38
rs377270443 Chr.2: 97084139-97084139 on Build GRCh38
rs764316293 Chr.2: 97084141-97084141 on Build GRCh38
rs779973879 Chr.2: 97084230-97084231 on Build GRCh38
rs535387721 Chr.2: 97084108-97084108 on Build GRCh38
rs765551519 Chr.2: 97084291-97084291 on Build GRCh38
rs772703870 Chr.2: 97084257-97084257 on Build GRCh38
rs780481770 Chr.2: 97084126-97084126 on Build GRCh38
rs898308074 Chr.2: 97084142-97084142 on Build GRCh38
rs762296956 Chr.2: 97084252-97084252 on Build GRCh38
rs546633352 Chr.2: 97084402-97084402 on Build GRCh38
rs777088352 Chr.2: 97084475-97084475 on Build GRCh38
rs777750406 Chr.2: 97084232-97084232 on Build GRCh38
rs772196498 Chr.2: 97084136-97084138 on Build GRCh38
rs759616753 Chr.2: 97084280-97084280 on Build GRCh38
rs747985950 Chr.2: 97084209-97084209 on Build GRCh38
rs753985714 Chr.2: 97084275-97084275 on Build GRCh38
rs576107978 Chr.2: 97084271-97084271 on Build GRCh38
rs58875686 Chr.2: 97084137-97084137 on Build GRCh38
rs200616594 Chr.2: 97084471-97084473 on Build GRCh38
rs772059406 Chr.2: 97084365-97084365 on Build GRCh38
rs768104120 Chr.2: 97084466-97084466 on Build GRCh38
rs561598987 Chr.2: 97084356-97084356 on Build GRCh38

View Full Product Details