Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 101288834-101289362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF149
Entrez Gene ID
284996
Gene Name
ring finger protein 149
Gene Aliases
DNAPTP2
UniGene
Hs.142074
Gene Chromosome Location
Chr.2: 101271213-101308716 on Build GRCh38
Transcripts
NM_173647.3 XM_005263920.2 XM_005263921.3 XM_011510990.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4722009 c.792T>C p.V264V Chr.2: 101289044-101289044 on Build GRCh38
1223909 c.848G>T p.R283I Chr.2: 101288988-101288988 on Build GRCh38
6767376 c.793G>T p.D265Y Chr.2: 101289043-101289043 on Build GRCh38
4642472 c.812T>C p.V271A Chr.2: 101289024-101289024 on Build GRCh38
1663935 c.830A>C p.K277T Chr.2: 101289006-101289006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555993430 Chr.2: 101288909-101288910 on Build GRCh38
rs114272320 Chr.2: 101289100-101289100 on Build GRCh38
rs577114259 Chr.2: 101288957-101288957 on Build GRCh38
rs575122360 Chr.2: 101288899-101288899 on Build GRCh38
rs561857123 Chr.2: 101289148-101289148 on Build GRCh38
rs757220386 Chr.2: 101289067-101289071 on Build GRCh38
rs1045981502 Chr.2: 101289170-101289170 on Build GRCh38
rs371323487 Chr.2: 101288992-101288992 on Build GRCh38
rs941467849 Chr.2: 101289109-101289109 on Build GRCh38
rs1045247796 Chr.2: 101289232-101289232 on Build GRCh38
rs776671773 Chr.2: 101288922-101288922 on Build GRCh38
rs781679262 Chr.2: 101289234-101289234 on Build GRCh38
rs145369067 Chr.2: 101288933-101288933 on Build GRCh38
rs374868835 Chr.2: 101288953-101288953 on Build GRCh38
rs761220792 Chr.2: 101289088-101289088 on Build GRCh38
rs113265852 Chr.2: 101288911-101288912 on Build GRCh38
rs767113060 Chr.2: 101289098-101289098 on Build GRCh38
rs754431670 Chr.2: 101288979-101288979 on Build GRCh38
rs898074080 Chr.2: 101289243-101289243 on Build GRCh38
rs999471177 Chr.2: 101288892-101288892 on Build GRCh38
rs376708004 Chr.2: 101288980-101288980 on Build GRCh38
rs28626790 Chr.2: 101289084-101289084 on Build GRCh38
rs116444630 Chr.2: 101289236-101289236 on Build GRCh38
rs747363127 Chr.2: 101289313-101289314 on Build GRCh38
rs1056806641 Chr.2: 101289167-101289167 on Build GRCh38
rs894223390 Chr.2: 101288875-101288875 on Build GRCh38
rs915922888 Chr.2: 101289144-101289144 on Build GRCh38
rs923214207 Chr.2: 101289099-101289099 on Build GRCh38
rs766761216 Chr.2: 101288971-101288971 on Build GRCh38
rs758718485 Chr.2: 101288995-101288995 on Build GRCh38
rs997501352 Chr.2: 101289275-101289275 on Build GRCh38
rs780982740 Chr.2: 101289039-101289039 on Build GRCh38
rs775297848 Chr.2: 101289072-101289072 on Build GRCh38
rs149187857 Chr.2: 101289143-101289143 on Build GRCh38
rs760123395 Chr.2: 101289101-101289101 on Build GRCh38
rs764658278 Chr.2: 101289296-101289296 on Build GRCh38
rs368561197 Chr.2: 101289085-101289085 on Build GRCh38
rs534214376 Chr.2: 101288904-101288904 on Build GRCh38
rs948450151 Chr.2: 101289217-101289217 on Build GRCh38
rs1030270162 Chr.2: 101289316-101289316 on Build GRCh38
rs747030236 Chr.2: 101289026-101289026 on Build GRCh38
rs915688762 Chr.2: 101289169-101289169 on Build GRCh38
rs775068730 Chr.2: 101288950-101288950 on Build GRCh38
rs758978804 Chr.2: 101289235-101289235 on Build GRCh38
rs778302141 Chr.2: 101288891-101288891 on Build GRCh38
rs753091852 Chr.2: 101289105-101289105 on Build GRCh38
rs773119397 Chr.2: 101289058-101289058 on Build GRCh38
rs376484956 Chr.2: 101289232-101289233 on Build GRCh38
rs1012447485 Chr.2: 101288878-101288878 on Build GRCh38
rs8179876 Chr.2: 101289233-101289233 on Build GRCh38
rs999017350 Chr.2: 101289226-101289226 on Build GRCh38
rs778192023 Chr.2: 101289007-101289007 on Build GRCh38
rs748887945 Chr.2: 101289078-101289078 on Build GRCh38
rs948766498 Chr.2: 101289163-101289163 on Build GRCh38
rs183848641 Chr.2: 101289104-101289104 on Build GRCh38
rs749202574 Chr.2: 101289060-101289060 on Build GRCh38
rs953503599 Chr.2: 101289319-101289319 on Build GRCh38
rs371951564 Chr.2: 101288964-101288964 on Build GRCh38
rs763556902 Chr.2: 101288954-101288954 on Build GRCh38
rs747443122 Chr.2: 101289030-101289030 on Build GRCh38
rs907470374 Chr.2: 101289215-101289215 on Build GRCh38
rs762203458 Chr.2: 101288868-101288869 on Build GRCh38
rs964400942 Chr.2: 101289258-101289258 on Build GRCh38
rs1045621367 Chr.2: 101288889-101288889 on Build GRCh38
rs1006853965 Chr.2: 101288915-101288915 on Build GRCh38
rs765709011 Chr.2: 101288985-101288985 on Build GRCh38
rs188689686 Chr.2: 101289238-101289238 on Build GRCh38
rs769638697 Chr.2: 101288944-101288944 on Build GRCh38
rs746001757 Chr.2: 101289127-101289127 on Build GRCh38
rs924522872 Chr.2: 101289137-101289137 on Build GRCh38
rs375456318 Chr.2: 101289228-101289229 on Build GRCh38
rs769514935 Chr.2: 101289064-101289064 on Build GRCh38

View Full Product Details