Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 101402345-101402842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFX8
Entrez Gene ID
731220
Gene Name
RFX family member 8, lacking RFX DNA binding domain
Gene Aliases
-
UniGene
Hs.662488
Gene Chromosome Location
Chr.2: 101397344-101475179 on Build GRCh38
Transcripts
NM_001145664.1 XM_011511771.2 XM_011511772.1 XM_011511773.1 XM_011511776.1 XM_011511777.1 XM_011511778.1 XM_017004851.1 XM_017004852.1 XM_017004853.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5538143 c.1033G>A p.E345K Chr.2: 101402648-101402648 on Build GRCh38
6534540 c.1331T>C p.L444P Chr.2: 101402689-101402689 on Build GRCh38
6534539 c.992T>C p.L331P Chr.2: 101402689-101402689 on Build GRCh38
6528396 c.1346A>G p.E449G Chr.2: 101402674-101402674 on Build GRCh38
6528395 c.1007A>G p.E336G Chr.2: 101402674-101402674 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048261731 Chr.2: 101402525-101402525 on Build GRCh38
rs749699476 Chr.2: 101402747-101402747 on Build GRCh38
rs780258998 Chr.2: 101402742-101402742 on Build GRCh38
rs959650095 Chr.2: 101402705-101402705 on Build GRCh38
rs763105786 Chr.2: 101402552-101402552 on Build GRCh38
rs370970266 Chr.2: 101402478-101402478 on Build GRCh38
rs1000380647 Chr.2: 101402748-101402748 on Build GRCh38
rs778750332 Chr.2: 101402634-101402634 on Build GRCh38
rs1014339867 Chr.2: 101402431-101402431 on Build GRCh38
rs2175968 Chr.2: 101402446-101402446 on Build GRCh38
rs560293444 Chr.2: 101402454-101402454 on Build GRCh38
rs1033812849 Chr.2: 101402678-101402678 on Build GRCh38
rs111429443 Chr.2: 101402476-101402476 on Build GRCh38
rs1055729235 Chr.2: 101402658-101402658 on Build GRCh38
rs79223800 Chr.2: 101402564-101402564 on Build GRCh38
rs778369560 Chr.2: 101402379-101402379 on Build GRCh38
rs1033151492 Chr.2: 101402752-101402752 on Build GRCh38
rs117260874 Chr.2: 101402621-101402621 on Build GRCh38
rs973643649 Chr.2: 101402728-101402728 on Build GRCh38
rs773922050 Chr.2: 101402792-101402792 on Build GRCh38
rs778328288 Chr.2: 101402575-101402575 on Build GRCh38
rs929409390 Chr.2: 101402512-101402512 on Build GRCh38
rs375170866 Chr.2: 101402569-101402569 on Build GRCh38
rs937298723 Chr.2: 101402645-101402645 on Build GRCh38
rs116596786 Chr.2: 101402502-101402502 on Build GRCh38
rs919133246 Chr.2: 101402536-101402537 on Build GRCh38
rs375753643 Chr.2: 101402428-101402428 on Build GRCh38
rs372128371 Chr.2: 101402560-101402560 on Build GRCh38
rs770057823 Chr.2: 101402531-101402531 on Build GRCh38
rs374769809 Chr.2: 101402704-101402704 on Build GRCh38
rs758513252 Chr.2: 101402643-101402643 on Build GRCh38
rs766122830 Chr.2: 101402602-101402602 on Build GRCh38
rs748998453 Chr.2: 101402509-101402509 on Build GRCh38
rs77323282 Chr.2: 101402563-101402563 on Build GRCh38
rs200668088 Chr.2: 101402445-101402445 on Build GRCh38
rs770145817 Chr.2: 101402737-101402737 on Build GRCh38
rs755915796 Chr.2: 101402465-101402465 on Build GRCh38
rs541417537 Chr.2: 101402443-101402443 on Build GRCh38
rs911974913 Chr.2: 101402778-101402778 on Build GRCh38
rs985833867 Chr.2: 101402495-101402495 on Build GRCh38
rs547206904 Chr.2: 101402448-101402448 on Build GRCh38
rs1038836207 Chr.2: 101402568-101402568 on Build GRCh38
rs772148011 Chr.2: 101402791-101402791 on Build GRCh38
rs1054091448 Chr.2: 101402577-101402577 on Build GRCh38
rs774610478 Chr.2: 101402764-101402764 on Build GRCh38
rs746230392 Chr.2: 101402504-101402504 on Build GRCh38
rs988520968 Chr.2: 101402442-101402442 on Build GRCh38
rs371118069 Chr.2: 101402546-101402546 on Build GRCh38
rs386648636 Chr.2: 101402446-101402448 on Build GRCh38
rs781675133 Chr.2: 101402670-101402670 on Build GRCh38
rs925627860 Chr.2: 101402455-101402455 on Build GRCh38
rs1018655999 Chr.2: 101402810-101402810 on Build GRCh38
rs778590926 Chr.2: 101402484-101402484 on Build GRCh38
rs267598800 Chr.2: 101402712-101402712 on Build GRCh38
rs930061374 Chr.2: 101402554-101402554 on Build GRCh38
rs758866541 Chr.2: 101402377-101402377 on Build GRCh38
rs11123876 Chr.2: 101402416-101402416 on Build GRCh38
rs201832728 Chr.2: 101402667-101402667 on Build GRCh38
rs971148424 Chr.2: 101402664-101402664 on Build GRCh38
rs568629571 Chr.2: 101402556-101402556 on Build GRCh38
rs965950022 Chr.2: 101402814-101402814 on Build GRCh38
rs888471411 Chr.2: 101402374-101402374 on Build GRCh38
rs750302756 Chr.2: 101402553-101402553 on Build GRCh38
rs754657455 Chr.2: 101402629-101402629 on Build GRCh38
rs375213952 Chr.2: 101402501-101402501 on Build GRCh38
rs746630582 Chr.2: 101402643-101402643 on Build GRCh38
rs746392837 Chr.2: 101402682-101402682 on Build GRCh38
rs779731788 Chr.2: 101402496-101402497 on Build GRCh38
rs752251754 Chr.2: 101402661-101402661 on Build GRCh38
rs758234287 Chr.2: 101402409-101402410 on Build GRCh38
rs779702917 Chr.2: 101402468-101402468 on Build GRCh38
rs568578609 Chr.2: 101402765-101402765 on Build GRCh38
rs750337240 Chr.2: 101402574-101402574 on Build GRCh38
rs199757001 Chr.2: 101402757-101402757 on Build GRCh38

View Full Product Details