Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 132416485-132416989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR39
Entrez Gene ID
2863
Gene Name
G protein-coupled receptor 39
Gene Aliases
-
UniGene
Hs.432395
Gene Chromosome Location
Chr.2: 132415972-132646596 on Build GRCh38
Transcripts
NM_001508.2 XM_011511021.2

SNP Detail

SNP ID Chromosome Location
rs987706867 Chr.2: 132416616-132416616 on Build GRCh38
rs370506301 Chr.2: 132416950-132416950 on Build GRCh38
rs780259344 Chr.2: 132416808-132416808 on Build GRCh38
rs1056312832 Chr.2: 132416802-132416802 on Build GRCh38
rs142166440 Chr.2: 132416833-132416833 on Build GRCh38
rs753631518 Chr.2: 132416638-132416638 on Build GRCh38
rs1029380739 Chr.2: 132416866-132416866 on Build GRCh38
rs548783317 Chr.2: 132416744-132416744 on Build GRCh38
rs751649275 Chr.2: 132416559-132416559 on Build GRCh38
rs893435489 Chr.2: 132416638-132416638 on Build GRCh38
rs932002060 Chr.2: 132416764-132416764 on Build GRCh38
rs939089575 Chr.2: 132416968-132416968 on Build GRCh38
rs555920786 Chr.2: 132416909-132416909 on Build GRCh38
rs532130097 Chr.2: 132416583-132416583 on Build GRCh38
rs112880263 Chr.2: 132416578-132416578 on Build GRCh38
rs887041221 Chr.2: 132416691-132416691 on Build GRCh38
rs1056177136 Chr.2: 132416952-132416952 on Build GRCh38
rs946468200 Chr.2: 132416653-132416653 on Build GRCh38
rs1022012915 Chr.2: 132416609-132416609 on Build GRCh38
rs79369600 Chr.2: 132416854-132416854 on Build GRCh38
rs543099404 Chr.2: 132416529-132416529 on Build GRCh38
rs915480989 Chr.2: 132416929-132416929 on Build GRCh38
rs767695785 Chr.2: 132416665-132416665 on Build GRCh38
rs943574613 Chr.2: 132416786-132416786 on Build GRCh38
rs952175465 Chr.2: 132416676-132416676 on Build GRCh38
rs1014909041 Chr.2: 132416530-132416530 on Build GRCh38
rs530293380 Chr.2: 132416719-132416719 on Build GRCh38
rs907924068 Chr.2: 132416721-132416721 on Build GRCh38
rs116602383 Chr.2: 132416948-132416948 on Build GRCh38
rs754227066 Chr.2: 132416821-132416821 on Build GRCh38
rs910631519 Chr.2: 132416517-132416517 on Build GRCh38
rs879594304 Chr.2: 132416840-132416840 on Build GRCh38
rs1004091973 Chr.2: 132416699-132416699 on Build GRCh38
rs776531765 Chr.2: 132416778-132416778 on Build GRCh38
rs761986101 Chr.2: 132416639-132416639 on Build GRCh38
rs537837580 Chr.2: 132416734-132416734 on Build GRCh38
rs576854500 Chr.2: 132416607-132416622 on Build GRCh38
rs1052070193 Chr.2: 132416632-132416632 on Build GRCh38
rs932167086 Chr.2: 132416604-132416604 on Build GRCh38
rs374260976 Chr.2: 132416795-132416795 on Build GRCh38
rs570239162 Chr.2: 132416782-132416782 on Build GRCh38
rs750606645 Chr.2: 132416736-132416736 on Build GRCh38
rs1035210688 Chr.2: 132416799-132416799 on Build GRCh38
rs755442278 Chr.2: 132416858-132416858 on Build GRCh38
rs547031182 Chr.2: 132416671-132416671 on Build GRCh38
rs567999708 Chr.2: 132416654-132416654 on Build GRCh38
rs1003363169 Chr.2: 132416672-132416672 on Build GRCh38
rs1049668948 Chr.2: 132416774-132416774 on Build GRCh38
rs963402502 Chr.2: 132416723-132416723 on Build GRCh38
rs988394696 Chr.2: 132416913-132416913 on Build GRCh38
rs368230563 Chr.2: 132416798-132416798 on Build GRCh38
rs968674377 Chr.2: 132416941-132416941 on Build GRCh38
rs1049705130 Chr.2: 132416552-132416552 on Build GRCh38
rs923285846 Chr.2: 132416760-132416760 on Build GRCh38
rs956863080 Chr.2: 132416911-132416911 on Build GRCh38
rs998096420 Chr.2: 132416525-132416525 on Build GRCh38
rs962554140 Chr.2: 132416841-132416841 on Build GRCh38
rs988320691 Chr.2: 132416780-132416780 on Build GRCh38
rs72999234 Chr.2: 132416801-132416801 on Build GRCh38
rs370977836 Chr.2: 132416790-132416790 on Build GRCh38
rs72999231 Chr.2: 132416673-132416673 on Build GRCh38
rs912133800 Chr.2: 132416617-132416617 on Build GRCh38
rs1015346924 Chr.2: 132416708-132416708 on Build GRCh38
rs922128184 Chr.2: 132416603-132416603 on Build GRCh38
rs1044815783 Chr.2: 132416664-132416664 on Build GRCh38
rs113114781 Chr.2: 132416954-132416954 on Build GRCh38
rs572326796 Chr.2: 132416872-132416872 on Build GRCh38
rs542535783 Chr.2: 132416879-132416879 on Build GRCh38
rs953703338 Chr.2: 132416745-132416745 on Build GRCh38
rs935557634 Chr.2: 132416627-132416627 on Build GRCh38
rs371443267 Chr.2: 132416759-132416759 on Build GRCh38
rs987613573 Chr.2: 132416755-132416755 on Build GRCh38
rs1010781877 Chr.2: 132416596-132416596 on Build GRCh38
rs963796678 Chr.2: 132416545-132416545 on Build GRCh38

View Full Product Details