Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 172564818-172565284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDK1
Entrez Gene ID
5163
Gene Name
pyruvate dehydrogenase kinase 1
Gene Aliases
-
UniGene
Hs.470633
Gene Chromosome Location
Chr.2: 172555373-172672693 on Build GRCh38
Transcripts
NM_001278549.1 NM_002610.4 NR_103729.1 NR_103731.1 XM_006712594.2 XM_006712595.3 XM_011511343.2 XM_011511344.2 XM_011511345.2 XM_011511347.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3961542 c.600A>G p.L200L Chr.2: 172564982-172564982 on Build GRCh38
4830791 c.634C>G p.H212D Chr.2: 172565016-172565016 on Build GRCh38
1009606 c.638G>A p.R213Q Chr.2: 172565020-172565020 on Build GRCh38
1716446 c.656T>C p.I219T Chr.2: 172565038-172565038 on Build GRCh38
5502562 c.596-1G>A p.? Chr.2: 172564977-172564977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770972654 Chr.2: 172565097-172565098 on Build GRCh38
rs781612335 Chr.2: 172565223-172565223 on Build GRCh38
rs758355812 Chr.2: 172565020-172565020 on Build GRCh38
rs909407349 Chr.2: 172565241-172565241 on Build GRCh38
rs760377304 Chr.2: 172565116-172565116 on Build GRCh38
rs774300380 Chr.2: 172565099-172565099 on Build GRCh38
rs556839669 Chr.2: 172565227-172565227 on Build GRCh38
rs1032176844 Chr.2: 172564978-172564978 on Build GRCh38
rs77896071 Chr.2: 172564900-172564900 on Build GRCh38
rs748428552 Chr.2: 172565061-172565061 on Build GRCh38
rs985396170 Chr.2: 172564861-172564861 on Build GRCh38
rs752073863 Chr.2: 172565036-172565036 on Build GRCh38
rs765512999 Chr.2: 172565120-172565120 on Build GRCh38
rs75108969 Chr.2: 172565011-172565011 on Build GRCh38
rs774212515 Chr.2: 172564970-172564970 on Build GRCh38
rs759327239 Chr.2: 172564971-172564971 on Build GRCh38
rs765375518 Chr.2: 172565016-172565016 on Build GRCh38
rs775425057 Chr.2: 172565111-172565111 on Build GRCh38
rs755860725 Chr.2: 172565070-172565070 on Build GRCh38
rs759902676 Chr.2: 172565044-172565044 on Build GRCh38
rs111983162 Chr.2: 172565017-172565017 on Build GRCh38
rs574805072 Chr.2: 172565098-172565099 on Build GRCh38
rs202231582 Chr.2: 172564994-172564994 on Build GRCh38
rs201965407 Chr.2: 172565054-172565054 on Build GRCh38
rs896457235 Chr.2: 172564898-172564898 on Build GRCh38
rs551980925 Chr.2: 172565098-172565098 on Build GRCh38
rs973782820 Chr.2: 172564968-172564968 on Build GRCh38
rs771972484 Chr.2: 172565108-172565108 on Build GRCh38
rs755417190 Chr.2: 172565053-172565053 on Build GRCh38
rs751622593 Chr.2: 172564940-172564940 on Build GRCh38
rs984957069 Chr.2: 172565221-172565221 on Build GRCh38
rs777277070 Chr.2: 172565085-172565085 on Build GRCh38
rs767257292 Chr.2: 172564988-172564988 on Build GRCh38
rs1009803240 Chr.2: 172564882-172564882 on Build GRCh38
rs766580150 Chr.2: 172564927-172564927 on Build GRCh38
rs373487872 Chr.2: 172564957-172564957 on Build GRCh38
rs910588268 Chr.2: 172564921-172564921 on Build GRCh38
rs745753140 Chr.2: 172565104-172565104 on Build GRCh38
rs566736913 Chr.2: 172564929-172564929 on Build GRCh38
rs749363874 Chr.2: 172564957-172564958 on Build GRCh38
rs909459925 Chr.2: 172564883-172564883 on Build GRCh38
rs750576998 Chr.2: 172565019-172565019 on Build GRCh38
rs184066061 Chr.2: 172565126-172565126 on Build GRCh38
rs377680754 Chr.2: 172564938-172564938 on Build GRCh38
rs766998131 Chr.2: 172565023-172565023 on Build GRCh38
rs770357642 Chr.2: 172565097-172565097 on Build GRCh38
rs375893098 Chr.2: 172564998-172564998 on Build GRCh38
rs200930840 Chr.2: 172565086-172565086 on Build GRCh38
rs754027344 Chr.2: 172564989-172564989 on Build GRCh38
rs1044242256 Chr.2: 172565145-172565145 on Build GRCh38
rs1055115584 Chr.2: 172564888-172564888 on Build GRCh38
rs760641414 Chr.2: 172565110-172565117 on Build GRCh38
rs905579462 Chr.2: 172565157-172565157 on Build GRCh38
rs150201631 Chr.2: 172564885-172564885 on Build GRCh38
rs773244911 Chr.2: 172565124-172565124 on Build GRCh38

View Full Product Details