Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197726713-197727235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOLL
Entrez Gene ID
66037
Gene Name
boule homolog, RNA binding protein
Gene Aliases
BOULE
UniGene
Hs.169797
Gene Chromosome Location
Chr.2: 197726879-197786574 on Build GRCh38
Transcripts
NM_001284358.1 NM_001284361.1 NM_001284362.1 NM_033030.5 NM_197970.2

SNP Detail

SNP ID Chromosome Location
rs184171794 Chr.2: 197726963-197726963 on Build GRCh38
rs938845381 Chr.2: 197727160-197727160 on Build GRCh38
rs1001590863 Chr.2: 197726996-197726996 on Build GRCh38
rs1035459200 Chr.2: 197726755-197726755 on Build GRCh38
rs796307800 Chr.2: 197727168-197727168 on Build GRCh38
rs954561993 Chr.2: 197726894-197726894 on Build GRCh38
rs905649267 Chr.2: 197726994-197726994 on Build GRCh38
rs896045071 Chr.2: 197727070-197727070 on Build GRCh38
rs941724924 Chr.2: 197726944-197726944 on Build GRCh38
rs746866265 Chr.2: 197727076-197727076 on Build GRCh38
rs373250286 Chr.2: 197727192-197727193 on Build GRCh38
rs942810664 Chr.2: 197727091-197727091 on Build GRCh38
rs1018772980 Chr.2: 197727136-197727136 on Build GRCh38
rs115736702 Chr.2: 197726834-197726834 on Build GRCh38
rs145963237 Chr.2: 197726826-197726826 on Build GRCh38
rs748716353 Chr.2: 197727172-197727172 on Build GRCh38
rs1021328212 Chr.2: 197726831-197726831 on Build GRCh38
rs894213616 Chr.2: 197727175-197727175 on Build GRCh38
rs968866530 Chr.2: 197727130-197727130 on Build GRCh38
rs188962496 Chr.2: 197727118-197727118 on Build GRCh38
rs758082088 Chr.2: 197727002-197727002 on Build GRCh38
rs933184032 Chr.2: 197727190-197727190 on Build GRCh38
rs565769231 Chr.2: 197726769-197726769 on Build GRCh38
rs542603072 Chr.2: 197727053-197727053 on Build GRCh38
rs1010464835 Chr.2: 197727111-197727111 on Build GRCh38
rs776101435 Chr.2: 197727164-197727164 on Build GRCh38
rs189000125 Chr.2: 197726770-197726770 on Build GRCh38
rs964189337 Chr.2: 197727140-197727140 on Build GRCh38
rs746631193 Chr.2: 197727120-197727120 on Build GRCh38
rs977127390 Chr.2: 197727153-197727153 on Build GRCh38
rs886276008 Chr.2: 197726861-197726861 on Build GRCh38
rs541124588 Chr.2: 197727151-197727151 on Build GRCh38
rs1047572004 Chr.2: 197726837-197726837 on Build GRCh38
rs538994196 Chr.2: 197726992-197726992 on Build GRCh38
rs191528754 Chr.2: 197727133-197727133 on Build GRCh38
rs777428639 Chr.2: 197726973-197726973 on Build GRCh38
rs1035392106 Chr.2: 197727049-197727049 on Build GRCh38
rs139884691 Chr.2: 197726857-197726857 on Build GRCh38
rs975383162 Chr.2: 197727095-197727095 on Build GRCh38
rs2272166 Chr.2: 197726960-197726960 on Build GRCh38
rs1002585838 Chr.2: 197726739-197726742 on Build GRCh38
rs372905447 Chr.2: 197727079-197727082 on Build GRCh38
rs1031880619 Chr.2: 197726873-197726875 on Build GRCh38
rs763773692 Chr.2: 197726860-197726861 on Build GRCh38

View Full Product Details