Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 200913155-200913691 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ORC2
Entrez Gene ID
4999
Gene Name
origin recognition complex subunit 2
Gene Aliases
ORC2L
UniGene
Hs.444870
Gene Chromosome Location
Chr.2: 200908981-200963703 on Build GRCh38
Transcripts
NM_006190.4 NR_033915.1 XM_006712555.3 XM_011511252.2 XM_011511253.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
442101 c.1612G>A p.E538K Chr.2: 200913330-200913330 on Build GRCh38
5742901 c.1589T>C p.L530P Chr.2: 200913353-200913353 on Build GRCh38
4414179 c.1582A>T p.S528C Chr.2: 200913360-200913360 on Build GRCh38
6735483 c.1575C>T p.L525L Chr.2: 200913367-200913367 on Build GRCh38
4411664 c.1598G>T p.R533L Chr.2: 200913344-200913344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751427938 Chr.2: 200913253-200913253 on Build GRCh38
rs762996489 Chr.2: 200913252-200913252 on Build GRCh38
rs900146382 Chr.2: 200913210-200913210 on Build GRCh38
rs199703940 Chr.2: 200913667-200913679 on Build GRCh38
rs760983379 Chr.2: 200913412-200913412 on Build GRCh38
rs774227899 Chr.2: 200913381-200913381 on Build GRCh38
rs913279906 Chr.2: 200913478-200913478 on Build GRCh38
rs977473503 Chr.2: 200913254-200913254 on Build GRCh38
rs992920629 Chr.2: 200913475-200913475 on Build GRCh38
rs773549106 Chr.2: 200913411-200913411 on Build GRCh38
rs779080457 Chr.2: 200913314-200913314 on Build GRCh38
rs557248708 Chr.2: 200913228-200913228 on Build GRCh38
rs377357388 Chr.2: 200913281-200913281 on Build GRCh38
rs1042213534 Chr.2: 200913484-200913484 on Build GRCh38
rs374482130 Chr.2: 200913278-200913278 on Build GRCh38
rs768616494 Chr.2: 200913272-200913272 on Build GRCh38
rs573181110 Chr.2: 200913489-200913489 on Build GRCh38
rs565939458 Chr.2: 200913433-200913433 on Build GRCh38
rs867942050 Chr.2: 200913511-200913511 on Build GRCh38
rs758874749 Chr.2: 200913451-200913452 on Build GRCh38
rs903670710 Chr.2: 200913538-200913538 on Build GRCh38
rs765721001 Chr.2: 200913449-200913449 on Build GRCh38
rs779211720 Chr.2: 200913264-200913265 on Build GRCh38
rs748632476 Chr.2: 200913504-200913504 on Build GRCh38
rs1013464598 Chr.2: 200913509-200913509 on Build GRCh38
rs780816370 Chr.2: 200913343-200913343 on Build GRCh38
rs897494656 Chr.2: 200913383-200913383 on Build GRCh38
rs775259781 Chr.2: 200913247-200913247 on Build GRCh38
rs377753630 Chr.2: 200913267-200913267 on Build GRCh38
rs1000548142 Chr.2: 200913200-200913200 on Build GRCh38
rs757626704 Chr.2: 200913250-200913251 on Build GRCh38
rs368060486 Chr.2: 200913420-200913420 on Build GRCh38
rs746350723 Chr.2: 200913364-200913366 on Build GRCh38
rs938687093 Chr.2: 200913467-200913467 on Build GRCh38
rs773307155 Chr.2: 200913316-200913316 on Build GRCh38
rs910810871 Chr.2: 200913288-200913288 on Build GRCh38
rs1033490494 Chr.2: 200913457-200913457 on Build GRCh38
rs754254328 Chr.2: 200913305-200913305 on Build GRCh38
rs374779161 Chr.2: 200913362-200913362 on Build GRCh38
rs753271221 Chr.2: 200913452-200913452 on Build GRCh38
rs1048832542 Chr.2: 200913426-200913426 on Build GRCh38
rs543285363 Chr.2: 200913327-200913327 on Build GRCh38
rs777253575 Chr.2: 200913406-200913406 on Build GRCh38
rs775087299 Chr.2: 200913348-200913348 on Build GRCh38
rs146980644 Chr.2: 200913344-200913344 on Build GRCh38
rs371450016 Chr.2: 200913439-200913439 on Build GRCh38
rs143665733 Chr.2: 200913366-200913366 on Build GRCh38
rs945208075 Chr.2: 200913479-200913479 on Build GRCh38
rs762057537 Chr.2: 200913259-200913259 on Build GRCh38
rs748528115 Chr.2: 200913323-200913323 on Build GRCh38
rs762446301 Chr.2: 200913534-200913534 on Build GRCh38
rs780080327 Chr.2: 200913213-200913213 on Build GRCh38
rs963997310 Chr.2: 200913279-200913279 on Build GRCh38
rs1038944041 Chr.2: 200913633-200913633 on Build GRCh38
rs540732693 Chr.2: 200913594-200913594 on Build GRCh38
rs758730230 Chr.2: 200913456-200913456 on Build GRCh38
rs746674632 Chr.2: 200913368-200913368 on Build GRCh38
rs16835624 Chr.2: 200913380-200913380 on Build GRCh38
rs370656830 Chr.2: 200913520-200913520 on Build GRCh38
rs370669334 Chr.2: 200913309-200913309 on Build GRCh38
rs755381940 Chr.2: 200913443-200913443 on Build GRCh38
rs761683522 Chr.2: 200913384-200913384 on Build GRCh38
rs371187163 Chr.2: 200913270-200913270 on Build GRCh38
rs538822533 Chr.2: 200913202-200913202 on Build GRCh38
rs554714404 Chr.2: 200913345-200913345 on Build GRCh38
rs1024298735 Chr.2: 200913516-200913516 on Build GRCh38

View Full Product Details