Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201269425-201269922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP8
Entrez Gene ID
841
Gene Name
caspase 8
Gene Aliases
ALPS2B, CAP4, Casp-8, FLICE, MACH, MCH5
UniGene
Hs.599762
Gene Chromosome Location
Chr.2: 201233443-201287711 on Build GRCh38
Transcripts
NM_001080124.1 NM_001080125.1 NM_001228.4 NM_033355.3 NM_033356.3 NM_033358.3 NR_111983.1 XM_005246885.1 XM_005246886.1 XM_005246887.1 XM_005246888.1 XM_005246889.1 XM_005246890.3 XM_005246891.4 XM_005246892.1 XM_005246893.2 XM_005246894.3 XM_005246895.2 XM_006712789.1 XM_006712790.3 XM_006712793.2 XM_011511969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3355740 c.401+1G>A p.? Chr.2: 201269606-201269606 on Build GRCh38
7283419 c.346C>A p.Q116K Chr.2: 201269550-201269550 on Build GRCh38
7343870 c.334G>T p.E112* Chr.2: 201269538-201269538 on Build GRCh38
7343872 c.372C>G p.F124L Chr.2: 201269576-201269576 on Build GRCh38
7343874 c.376C>T p.R126W Chr.2: 201269580-201269580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746923682 Chr.2: 201269865-201269865 on Build GRCh38
rs773675844 Chr.2: 201269523-201269523 on Build GRCh38
rs545101388 Chr.2: 201269634-201269634 on Build GRCh38
rs11897628 Chr.2: 201269598-201269598 on Build GRCh38
rs773733228 Chr.2: 201269589-201269589 on Build GRCh38
rs757197450 Chr.2: 201269466-201269466 on Build GRCh38
rs551357234 Chr.2: 201269449-201269449 on Build GRCh38
rs778565735 Chr.2: 201269572-201269572 on Build GRCh38
rs763645569 Chr.2: 201269539-201269539 on Build GRCh38
rs35655821 Chr.2: 201269695-201269695 on Build GRCh38
rs780983171 Chr.2: 201269473-201269473 on Build GRCh38
rs757151756 Chr.2: 201269549-201269549 on Build GRCh38
rs951355772 Chr.2: 201269535-201269535 on Build GRCh38
rs777185174 Chr.2: 201269459-201269459 on Build GRCh38
rs745855281 Chr.2: 201269563-201269563 on Build GRCh38
rs749554709 Chr.2: 201269495-201269495 on Build GRCh38
rs955829118 Chr.2: 201269788-201269788 on Build GRCh38
rs775667916 Chr.2: 201269491-201269491 on Build GRCh38
rs751043226 Chr.2: 201269546-201269546 on Build GRCh38
rs199830384 Chr.2: 201269613-201269613 on Build GRCh38
rs371728950 Chr.2: 201269514-201269514 on Build GRCh38
rs1038869360 Chr.2: 201269817-201269817 on Build GRCh38
rs758051022 Chr.2: 201269538-201269538 on Build GRCh38
rs949773873 Chr.2: 201269453-201269453 on Build GRCh38
rs547238272 Chr.2: 201269733-201269733 on Build GRCh38
rs372378810 Chr.2: 201269526-201269526 on Build GRCh38
rs763657503 Chr.2: 201269624-201269624 on Build GRCh38
rs947662363 Chr.2: 201269571-201269571 on Build GRCh38
rs776712453 Chr.2: 201269533-201269533 on Build GRCh38
rs751038927 Chr.2: 201269628-201269628 on Build GRCh38
rs767054828 Chr.2: 201269638-201269638 on Build GRCh38
rs951416999 Chr.2: 201269883-201269883 on Build GRCh38
rs781137365 Chr.2: 201269561-201269561 on Build GRCh38
rs887677560 Chr.2: 201269687-201269687 on Build GRCh38
rs563486849 Chr.2: 201269658-201269658 on Build GRCh38
rs574310665 Chr.2: 201269502-201269502 on Build GRCh38
rs765672355 Chr.2: 201269567-201269568 on Build GRCh38
rs796850798 Chr.2: 201269605-201269605 on Build GRCh38
rs762325737 Chr.2: 201269515-201269515 on Build GRCh38
rs748142457 Chr.2: 201269569-201269569 on Build GRCh38
rs988514241 Chr.2: 201269562-201269562 on Build GRCh38
rs1035824881 Chr.2: 201269768-201269768 on Build GRCh38
rs181730491 Chr.2: 201269670-201269670 on Build GRCh38
rs35496483 Chr.2: 201269474-201269475 on Build GRCh38
rs377216101 Chr.2: 201269519-201269519 on Build GRCh38
rs969795062 Chr.2: 201269522-201269522 on Build GRCh38
rs998239752 Chr.2: 201269856-201269856 on Build GRCh38
rs901673433 Chr.2: 201269850-201269850 on Build GRCh38
rs768834851 Chr.2: 201269468-201269469 on Build GRCh38
rs911424524 Chr.2: 201269632-201269632 on Build GRCh38
rs1004587966 Chr.2: 201269884-201269884 on Build GRCh38
rs373232880 Chr.2: 201269606-201269606 on Build GRCh38
rs368609027 Chr.2: 201269524-201269524 on Build GRCh38
rs1026624116 Chr.2: 201269482-201269482 on Build GRCh38
rs766842904 Chr.2: 201269602-201269602 on Build GRCh38
rs1002712227 Chr.2: 201269744-201269744 on Build GRCh38
rs191441520 Chr.2: 201269700-201269700 on Build GRCh38
rs1006047483 Chr.2: 201269490-201269490 on Build GRCh38
rs761301334 Chr.2: 201269601-201269601 on Build GRCh38
rs35154230 Chr.2: 201269455-201269455 on Build GRCh38
rs772577997 Chr.2: 201269587-201269587 on Build GRCh38
rs555778451 Chr.2: 201269486-201269486 on Build GRCh38
rs749952869 Chr.2: 201269643-201269643 on Build GRCh38
rs534884641 Chr.2: 201269588-201269588 on Build GRCh38
rs746960440 Chr.2: 201269462-201269462 on Build GRCh38
rs1054380210 Chr.2: 201269731-201269731 on Build GRCh38
rs748211754 Chr.2: 201269582-201269582 on Build GRCh38
rs367547356 Chr.2: 201269483-201269483 on Build GRCh38
rs756102899 Chr.2: 201269657-201269657 on Build GRCh38
rs34721127 Chr.2: 201269800-201269801 on Build GRCh38
rs768776043 Chr.2: 201269580-201269580 on Build GRCh38
rs749516269 Chr.2: 201269579-201269579 on Build GRCh38
rs774427459 Chr.2: 201269581-201269581 on Build GRCh38
rs779893658 Chr.2: 201269573-201269573 on Build GRCh38
rs762135470 Chr.2: 201269559-201269560 on Build GRCh38
rs376590303 Chr.2: 201269536-201269536 on Build GRCh38
rs559596473 Chr.2: 201269864-201269864 on Build GRCh38

View Full Product Details