Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218386550-218387054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC11A1
Entrez Gene ID
6556
Gene Name
solute carrier family 11 member 1
Gene Aliases
LSH, NRAMP, NRAMP1
UniGene
Hs.591607
Gene Chromosome Location
Chr.2: 218381766-218396894 on Build GRCh38
Transcripts
NM_000578.3 XM_005246793.3 XM_005246794.3 XM_006712709.3 XM_006712710.3 XM_006712711.3 XM_011511684.2 XM_011511685.2 XM_017004765.1 XM_017004766.1 XM_017004767.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4938424 c.425A>G p.E142G Chr.2: 218386666-218386666 on Build GRCh38
4726367 c.456A>C p.E152D Chr.2: 218386697-218386697 on Build GRCh38
6779217 c.480C>T p.F160F Chr.2: 218386721-218386721 on Build GRCh38
4091426 c.405C>T p.T135T Chr.2: 218386646-218386646 on Build GRCh38
4001503 c.400C>G p.R134G Chr.2: 218386641-218386641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922772160 Chr.2: 218386758-218386758 on Build GRCh38
rs189340934 Chr.2: 218386583-218386583 on Build GRCh38
rs761891303 Chr.2: 218386776-218386776 on Build GRCh38
rs941072595 Chr.2: 218386792-218386792 on Build GRCh38
rs555287828 Chr.2: 218386979-218386979 on Build GRCh38
rs765551371 Chr.2: 218386829-218386829 on Build GRCh38
rs375469392 Chr.2: 218386787-218386787 on Build GRCh38
rs143294002 Chr.2: 218386708-218386708 on Build GRCh38
rs11677367 Chr.2: 218387006-218387006 on Build GRCh38
rs758848111 Chr.2: 218386965-218386965 on Build GRCh38
rs765577284 Chr.2: 218386665-218386665 on Build GRCh38
rs1012016441 Chr.2: 218386664-218386664 on Build GRCh38
rs778524193 Chr.2: 218386606-218386606 on Build GRCh38
rs905717805 Chr.2: 218386910-218386910 on Build GRCh38
rs774712680 Chr.2: 218386771-218386771 on Build GRCh38
rs1024056581 Chr.2: 218386642-218386642 on Build GRCh38
rs750650152 Chr.2: 218386785-218386785 on Build GRCh38
rs901736642 Chr.2: 218386961-218386961 on Build GRCh38
rs377217125 Chr.2: 218386601-218386601 on Build GRCh38
rs770642709 Chr.2: 218386741-218386741 on Build GRCh38
rs535474612 Chr.2: 218386968-218386968 on Build GRCh38
rs763479758 Chr.2: 218386788-218386789 on Build GRCh38
rs750757237 Chr.2: 218386717-218386717 on Build GRCh38
rs916277804 Chr.2: 218386807-218386807 on Build GRCh38
rs1043535344 Chr.2: 218386846-218386846 on Build GRCh38
rs756178525 Chr.2: 218386866-218386866 on Build GRCh38
rs754830948 Chr.2: 218386713-218386713 on Build GRCh38
rs369053258 Chr.2: 218386740-218386740 on Build GRCh38
rs776019462 Chr.2: 218386685-218386685 on Build GRCh38
rs775967572 Chr.2: 218386745-218386745 on Build GRCh38
rs1024576370 Chr.2: 218386871-218386871 on Build GRCh38
rs772031291 Chr.2: 218386636-218386636 on Build GRCh38
rs759915231 Chr.2: 218386657-218386657 on Build GRCh38
rs530669474 Chr.2: 218386665-218386665 on Build GRCh38
rs151164925 Chr.2: 218386732-218386732 on Build GRCh38
rs752494281 Chr.2: 218386805-218386805 on Build GRCh38
rs754955687 Chr.2: 218386598-218386598 on Build GRCh38
rs747985621 Chr.2: 218386618-218386618 on Build GRCh38
rs1005163917 Chr.2: 218386593-218386593 on Build GRCh38
rs958106555 Chr.2: 218387009-218387009 on Build GRCh38
rs570403407 Chr.2: 218386836-218386836 on Build GRCh38
rs757505650 Chr.2: 218386703-218386703 on Build GRCh38
rs145880987 Chr.2: 218386646-218386646 on Build GRCh38
rs372158991 Chr.2: 218386765-218386765 on Build GRCh38
rs938949364 Chr.2: 218386802-218386802 on Build GRCh38
rs145071313 Chr.2: 218386686-218386686 on Build GRCh38
rs766621082 Chr.2: 218386789-218386789 on Build GRCh38
rs1056511200 Chr.2: 218386803-218386803 on Build GRCh38
rs746714183 Chr.2: 218386645-218386645 on Build GRCh38
rs775964315 Chr.2: 218386752-218386752 on Build GRCh38
rs530607695 Chr.2: 218386806-218386806 on Build GRCh38
rs201910426 Chr.2: 218386641-218386641 on Build GRCh38
rs767812103 Chr.2: 218386781-218386781 on Build GRCh38
rs568086968 Chr.2: 218386963-218386963 on Build GRCh38
rs563632925 Chr.2: 218386722-218386722 on Build GRCh38
rs1020938240 Chr.2: 218387024-218387024 on Build GRCh38
rs200748847 Chr.2: 218386751-218386751 on Build GRCh38
rs141932707 Chr.2: 218386714-218386714 on Build GRCh38
rs750537920 Chr.2: 218386710-218386710 on Build GRCh38
rs1034160155 Chr.2: 218386648-218386648 on Build GRCh38
rs780549795 Chr.2: 218387020-218387020 on Build GRCh38
rs777624950 Chr.2: 218387033-218387033 on Build GRCh38
rs762217800 Chr.2: 218386822-218386822 on Build GRCh38
rs758251194 Chr.2: 218386726-218386726 on Build GRCh38
rs928845636 Chr.2: 218386801-218386801 on Build GRCh38
rs751778521 Chr.2: 218386699-218386699 on Build GRCh38
rs962222571 Chr.2: 218386799-218386799 on Build GRCh38
rs529909528 Chr.2: 218386647-218386647 on Build GRCh38
rs755645050 Chr.2: 218387028-218387028 on Build GRCh38
rs764294063 Chr.2: 218386674-218386674 on Build GRCh38
rs764502812 Chr.2: 218386694-218386694 on Build GRCh38
rs552393447 Chr.2: 218386814-218386814 on Build GRCh38
rs897077730 Chr.2: 218386634-218386634 on Build GRCh38
rs932795237 Chr.2: 218386764-218386764 on Build GRCh38
rs894848159 Chr.2: 218386649-218386649 on Build GRCh38
rs767438872 Chr.2: 218386709-218386709 on Build GRCh38
rs749114236 Chr.2: 218386597-218386597 on Build GRCh38
rs752152218 Chr.2: 218387025-218387025 on Build GRCh38
rs1006258174 Chr.2: 218387000-218387000 on Build GRCh38
rs1036595261 Chr.2: 218386604-218386604 on Build GRCh38
rs769240006 Chr.2: 218386748-218386748 on Build GRCh38
rs972280080 Chr.2: 218386800-218386800 on Build GRCh38
rs534714777 Chr.2: 218386908-218386908 on Build GRCh38
rs546582945 Chr.2: 218386962-218386962 on Build GRCh38

View Full Product Details