Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 225652075-225652586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NYAP2
Entrez Gene ID
57624
Gene Name
neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 2
Gene Aliases
KIAA1486
UniGene
Hs.224409
Gene Chromosome Location
Chr.2: 225399710-225703653 on Build GRCh38
Transcripts
NM_020864.1 XM_005246708.3 XM_011511524.1 XM_017004564.1

SNP Detail

SNP ID Chromosome Location
rs541188206 Chr.2: 225652376-225652376 on Build GRCh38
rs765515280 Chr.2: 225652217-225652220 on Build GRCh38
rs995213077 Chr.2: 225652430-225652430 on Build GRCh38
rs771230859 Chr.2: 225652461-225652461 on Build GRCh38
rs963327938 Chr.2: 225652406-225652406 on Build GRCh38
rs960893586 Chr.2: 225652219-225652219 on Build GRCh38
rs897804025 Chr.2: 225652151-225652151 on Build GRCh38
rs944567657 Chr.2: 225652400-225652400 on Build GRCh38
rs1010430087 Chr.2: 225652180-225652180 on Build GRCh38
rs759970523 Chr.2: 225652514-225652514 on Build GRCh38
rs559879612 Chr.2: 225652470-225652470 on Build GRCh38
rs1041899005 Chr.2: 225652492-225652492 on Build GRCh38
rs1035178597 Chr.2: 225652214-225652214 on Build GRCh38
rs73074529 Chr.2: 225652262-225652262 on Build GRCh38
rs4641920 Chr.2: 225652521-225652521 on Build GRCh38
rs577439891 Chr.2: 225652457-225652457 on Build GRCh38
rs544188504 Chr.2: 225652351-225652351 on Build GRCh38
rs945672632 Chr.2: 225652274-225652274 on Build GRCh38
rs994866557 Chr.2: 225652166-225652166 on Build GRCh38
rs973704180 Chr.2: 225652410-225652410 on Build GRCh38
rs911714295 Chr.2: 225652388-225652388 on Build GRCh38
rs960841467 Chr.2: 225652488-225652488 on Build GRCh38
rs539259877 Chr.2: 225652207-225652207 on Build GRCh38
rs980129574 Chr.2: 225652542-225652542 on Build GRCh38
rs1009750494 Chr.2: 225652511-225652511 on Build GRCh38
rs931732013 Chr.2: 225652347-225652347 on Build GRCh38
rs4627553 Chr.2: 225652466-225652466 on Build GRCh38
rs553104218 Chr.2: 225652243-225652243 on Build GRCh38
rs897756262 Chr.2: 225652428-225652428 on Build GRCh38
rs1036214310 Chr.2: 225652417-225652417 on Build GRCh38
rs570683808 Chr.2: 225652206-225652206 on Build GRCh38
rs1028126181 Chr.2: 225652195-225652195 on Build GRCh38
rs186563695 Chr.2: 225652297-225652297 on Build GRCh38
rs1035126128 Chr.2: 225652473-225652473 on Build GRCh38
rs200687673 Chr.2: 225652256-225652256 on Build GRCh38
rs763726270 Chr.2: 225652379-225652379 on Build GRCh38
rs890329865 Chr.2: 225652110-225652110 on Build GRCh38
rs1049061824 Chr.2: 225652455-225652455 on Build GRCh38
rs1014824390 Chr.2: 225652129-225652129 on Build GRCh38
rs1050169667 Chr.2: 225652357-225652357 on Build GRCh38
rs563832498 Chr.2: 225652456-225652456 on Build GRCh38
rs978457941 Chr.2: 225652302-225652302 on Build GRCh38
rs1008764004 Chr.2: 225652125-225652125 on Build GRCh38
rs772550630 Chr.2: 225652240-225652240 on Build GRCh38
rs11681238 Chr.2: 225652323-225652323 on Build GRCh38
rs1039906328 Chr.2: 225652537-225652537 on Build GRCh38
rs10180401 Chr.2: 225652197-225652197 on Build GRCh38
rs111355283 Chr.2: 225652169-225652169 on Build GRCh38
rs926906029 Chr.2: 225652112-225652112 on Build GRCh38
rs1002276434 Chr.2: 225652469-225652469 on Build GRCh38
rs1029731182 Chr.2: 225652278-225652278 on Build GRCh38

View Full Product Details