Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 663426-663918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCRT2
Entrez Gene ID
85508
Gene Name
scratch family transcriptional repressor 2
Gene Aliases
ZNF898B
UniGene
Hs.355284
Gene Chromosome Location
Chr.20: 661596-676179 on Build GRCh38
Transcripts
NM_033129.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6539336 c.757T>C p.S253P Chr.20: 663838-663838 on Build GRCh38
6558488 c.747C>A p.F249L Chr.20: 663848-663848 on Build GRCh38
117589 c.720C>A p.F240L Chr.20: 663875-663875 on Build GRCh38
6497297 c.862G>C p.E288Q Chr.20: 663733-663733 on Build GRCh38
6539335 c.886G>C p.E296Q Chr.20: 663709-663709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760009226 Chr.20: 663783-663783 on Build GRCh38
rs757441060 Chr.20: 663839-663839 on Build GRCh38
rs778909173 Chr.20: 663866-663866 on Build GRCh38
rs760191714 Chr.20: 663771-663771 on Build GRCh38
rs749136861 Chr.20: 663729-663729 on Build GRCh38
rs574061608 Chr.20: 663824-663824 on Build GRCh38
rs992012184 Chr.20: 663469-663469 on Build GRCh38
rs975395309 Chr.20: 663669-663669 on Build GRCh38
rs918528845 Chr.20: 663445-663445 on Build GRCh38
rs780896155 Chr.20: 663594-663594 on Build GRCh38
rs916358332 Chr.20: 663546-663546 on Build GRCh38
rs956463485 Chr.20: 663618-663618 on Build GRCh38
rs771985405 Chr.20: 663699-663699 on Build GRCh38
rs747952310 Chr.20: 663867-663867 on Build GRCh38
rs948980861 Chr.20: 663782-663782 on Build GRCh38
rs1000949717 Chr.20: 663565-663565 on Build GRCh38
rs773042402 Chr.20: 663884-663885 on Build GRCh38
rs770363574 Chr.20: 663776-663776 on Build GRCh38
rs201413966 Chr.20: 663779-663779 on Build GRCh38
rs539854040 Chr.20: 663872-663872 on Build GRCh38
rs747139533 Chr.20: 663704-663704 on Build GRCh38
rs772838171 Chr.20: 663758-663758 on Build GRCh38
rs1006999028 Chr.20: 663646-663646 on Build GRCh38
rs747947406 Chr.20: 663748-663748 on Build GRCh38
rs117765732 Chr.20: 663449-663449 on Build GRCh38
rs748400239 Chr.20: 663583-663583 on Build GRCh38
rs6076891 Chr.20: 663659-663659 on Build GRCh38
rs566775117 Chr.20: 663554-663554 on Build GRCh38
rs1025696433 Chr.20: 663716-663716 on Build GRCh38
rs546874686 Chr.20: 663464-663464 on Build GRCh38
rs965282254 Chr.20: 663653-663653 on Build GRCh38
rs765742132 Chr.20: 663785-663785 on Build GRCh38
rs777409794 Chr.20: 663891-663891 on Build GRCh38
rs904596918 Chr.20: 663564-663564 on Build GRCh38
rs112894948 Chr.20: 663534-663534 on Build GRCh38
rs778624633 Chr.20: 663747-663747 on Build GRCh38
rs952708621 Chr.20: 663677-663677 on Build GRCh38
rs747898431 Chr.20: 663685-663694 on Build GRCh38
rs765468891 Chr.20: 663690-663690 on Build GRCh38
rs940130005 Chr.20: 663710-663710 on Build GRCh38
rs778136855 Chr.20: 663700-663700 on Build GRCh38
rs771782325 Chr.20: 663754-663754 on Build GRCh38
rs781374957 Chr.20: 663845-663845 on Build GRCh38
rs751761982 Chr.20: 663827-663827 on Build GRCh38
rs1016662746 Chr.20: 663674-663674 on Build GRCh38
rs763361664 Chr.20: 663820-663820 on Build GRCh38
rs780142197 Chr.20: 663728-663728 on Build GRCh38
rs554669181 Chr.20: 663794-663794 on Build GRCh38
rs746625438 Chr.20: 663899-663899 on Build GRCh38
rs754916635 Chr.20: 663863-663863 on Build GRCh38
rs757344816 Chr.20: 663709-663709 on Build GRCh38
rs753082465 Chr.20: 663693-663693 on Build GRCh38
rs1013148851 Chr.20: 663543-663543 on Build GRCh38
rs377277303 Chr.20: 663860-663860 on Build GRCh38
rs972383353 Chr.20: 663703-663703 on Build GRCh38
rs768521901 Chr.20: 663742-663742 on Build GRCh38
rs770079336 Chr.20: 663707-663707 on Build GRCh38
rs1053474839 Chr.20: 663892-663892 on Build GRCh38
rs908610113 Chr.20: 663689-663689 on Build GRCh38
rs575945466 Chr.20: 663612-663612 on Build GRCh38
rs575428207 Chr.20: 663616-663616 on Build GRCh38
rs998811862 Chr.20: 663619-663619 on Build GRCh38
rs112858215 Chr.20: 663558-663558 on Build GRCh38
rs1023712945 Chr.20: 663547-663547 on Build GRCh38
rs892207105 Chr.20: 663896-663896 on Build GRCh38
rs758588604 Chr.20: 663694-663694 on Build GRCh38
rs962411795 Chr.20: 663680-663680 on Build GRCh38
rs537520207 Chr.20: 663691-663691 on Build GRCh38
rs936011802 Chr.20: 663572-663572 on Build GRCh38
rs1029833394 Chr.20: 663628-663628 on Build GRCh38
rs896051534 Chr.20: 663519-663519 on Build GRCh38
rs889991399 Chr.20: 663629-663629 on Build GRCh38

View Full Product Details