Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2403439-2403942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGM6
Entrez Gene ID
343641
Gene Name
transglutaminase 6
Gene Aliases
SCA35, TG6, TGM3L, TGY, dJ734P14.3
UniGene
Hs.452039
Gene Chromosome Location
Chr.20: 2380908-2432753 on Build GRCh38
Transcripts
NM_001254734.1 NM_198994.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
443555 c.1178C>T p.A393V Chr.20: 2403665-2403665 on Build GRCh38
2887029 c.1318C>T p.L440F Chr.20: 2403805-2403805 on Build GRCh38
1533177 c.1190C>A p.A397D Chr.20: 2403677-2403677 on Build GRCh38
7393696 c.1235G>A p.R412H Chr.20: 2403722-2403722 on Build GRCh38
3544783 c.1094G>A p.G365D Chr.20: 2403581-2403581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768299438 Chr.20: 2403567-2403567 on Build GRCh38
rs546728628 Chr.20: 2403718-2403718 on Build GRCh38
rs151011799 Chr.20: 2403860-2403860 on Build GRCh38
rs931380756 Chr.20: 2403881-2403881 on Build GRCh38
rs770145628 Chr.20: 2403787-2403787 on Build GRCh38
rs758477530 Chr.20: 2403612-2403612 on Build GRCh38
rs759324511 Chr.20: 2403734-2403734 on Build GRCh38
rs774605988 Chr.20: 2403716-2403716 on Build GRCh38
rs752770191 Chr.20: 2403534-2403534 on Build GRCh38
rs745804281 Chr.20: 2403757-2403757 on Build GRCh38
rs553470142 Chr.20: 2403478-2403478 on Build GRCh38
rs200015721 Chr.20: 2403619-2403619 on Build GRCh38
rs61729227 Chr.20: 2403601-2403601 on Build GRCh38
rs920072527 Chr.20: 2403665-2403665 on Build GRCh38
rs767072224 Chr.20: 2403823-2403823 on Build GRCh38
rs200872034 Chr.20: 2403646-2403646 on Build GRCh38
rs145029765 Chr.20: 2403675-2403675 on Build GRCh38
rs777370613 Chr.20: 2403759-2403759 on Build GRCh38
rs756832151 Chr.20: 2403822-2403822 on Build GRCh38
rs765423180 Chr.20: 2403657-2403657 on Build GRCh38
rs1051520144 Chr.20: 2403898-2403898 on Build GRCh38
rs769606752 Chr.20: 2403479-2403479 on Build GRCh38
rs1025921393 Chr.20: 2403606-2403606 on Build GRCh38
rs79724097 Chr.20: 2403613-2403613 on Build GRCh38
rs781150689 Chr.20: 2403544-2403544 on Build GRCh38
rs771605676 Chr.20: 2403632-2403632 on Build GRCh38
rs775263431 Chr.20: 2403480-2403480 on Build GRCh38
rs199802654 Chr.20: 2403805-2403805 on Build GRCh38
rs906857263 Chr.20: 2403560-2403560 on Build GRCh38
rs141071237 Chr.20: 2403737-2403737 on Build GRCh38
rs150243412 Chr.20: 2403788-2403788 on Build GRCh38
rs556958028 Chr.20: 2403769-2403769 on Build GRCh38
rs998653850 Chr.20: 2403897-2403897 on Build GRCh38
rs558070858 Chr.20: 2403859-2403859 on Build GRCh38
rs747526538 Chr.20: 2403641-2403641 on Build GRCh38
rs757170552 Chr.20: 2403539-2403539 on Build GRCh38
rs545959266 Chr.20: 2403581-2403581 on Build GRCh38
rs772274519 Chr.20: 2403804-2403804 on Build GRCh38
rs1014253938 Chr.20: 2403730-2403730 on Build GRCh38
rs752383362 Chr.20: 2403746-2403746 on Build GRCh38
rs201326247 Chr.20: 2403589-2403589 on Build GRCh38
rs539728492 Chr.20: 2403855-2403855 on Build GRCh38
rs769042038 Chr.20: 2403790-2403790 on Build GRCh38
rs749044326 Chr.20: 2403621-2403621 on Build GRCh38
rs764278569 Chr.20: 2403666-2403666 on Build GRCh38
rs1043196138 Chr.20: 2403863-2403863 on Build GRCh38
rs765093828 Chr.20: 2403738-2403738 on Build GRCh38
rs533665038 Chr.20: 2403702-2403702 on Build GRCh38
rs765457146 Chr.20: 2403602-2403602 on Build GRCh38
rs751116090 Chr.20: 2403765-2403765 on Build GRCh38
rs749690053 Chr.20: 2403539-2403539 on Build GRCh38
rs920038661 Chr.20: 2403877-2403877 on Build GRCh38
rs142080899 Chr.20: 2403679-2403679 on Build GRCh38
rs868694069 Chr.20: 2403672-2403672 on Build GRCh38
rs200536588 Chr.20: 2403721-2403721 on Build GRCh38
rs373331741 Chr.20: 2403699-2403699 on Build GRCh38
rs762242728 Chr.20: 2403496-2403496 on Build GRCh38
rs746480307 Chr.20: 2403690-2403690 on Build GRCh38
rs745703185 Chr.20: 2403550-2403550 on Build GRCh38
rs749578418 Chr.20: 2403789-2403789 on Build GRCh38
rs752757164 Chr.20: 2403611-2403611 on Build GRCh38
rs769242742 Chr.20: 2403617-2403618 on Build GRCh38
rs371763464 Chr.20: 2403645-2403645 on Build GRCh38
rs757930415 Chr.20: 2403683-2403683 on Build GRCh38
rs755965073 Chr.20: 2403551-2403551 on Build GRCh38
rs115126643 Chr.20: 2403516-2403516 on Build GRCh38
rs142679146 Chr.20: 2403722-2403722 on Build GRCh38
rs148470870 Chr.20: 2403622-2403622 on Build GRCh38
rs898763439 Chr.20: 2403515-2403515 on Build GRCh38
rs748647492 Chr.20: 2403855-2403855 on Build GRCh38
rs189066202 Chr.20: 2403487-2403487 on Build GRCh38
rs200726041 Chr.20: 2403474-2403474 on Build GRCh38
rs147554438 Chr.20: 2403678-2403678 on Build GRCh38
rs754053350 Chr.20: 2403517-2403517 on Build GRCh38
rs369633361 Chr.20: 2403799-2403799 on Build GRCh38
rs752095723 Chr.20: 2403713-2403713 on Build GRCh38
rs773687683 Chr.20: 2403583-2403583 on Build GRCh38
rs138950659 Chr.20: 2403809-2403809 on Build GRCh38
rs763791368 Chr.20: 2403819-2403819 on Build GRCh38
rs552530897 Chr.20: 2403685-2403685 on Build GRCh38
rs75122556 Chr.20: 2403590-2403590 on Build GRCh38
rs201024326 Chr.20: 2403483-2403483 on Build GRCh38
rs780396011 Chr.20: 2403839-2403839 on Build GRCh38
rs749855515 Chr.20: 2403829-2403829 on Build GRCh38
rs574493508 Chr.20: 2403905-2403905 on Build GRCh38
rs930243360 Chr.20: 2403669-2403669 on Build GRCh38
rs759540566 Chr.20: 2403808-2403808 on Build GRCh38
rs552753944 Chr.20: 2403891-2403891 on Build GRCh38
rs771377547 Chr.20: 2403644-2403644 on Build GRCh38
rs774963727 Chr.20: 2403738-2403740 on Build GRCh38
rs766380908 Chr.20: 2403677-2403677 on Build GRCh38
rs776922671 Chr.20: 2403594-2403594 on Build GRCh38
rs769816743 Chr.20: 2403867-2403867 on Build GRCh38
rs760996972 Chr.20: 2403508-2403508 on Build GRCh38
rs751634905 Chr.20: 2403674-2403674 on Build GRCh38
rs747656727 Chr.20: 2403576-2403576 on Build GRCh38
rs573390589 Chr.20: 2403794-2403794 on Build GRCh38
rs762711483 Chr.20: 2403818-2403818 on Build GRCh38
rs751204014 Chr.20: 2403820-2403820 on Build GRCh38
rs756678657 Chr.20: 2403836-2403836 on Build GRCh38
rs756743789 Chr.20: 2403766-2403766 on Build GRCh38
rs116904482 Chr.20: 2403658-2403658 on Build GRCh38
rs986690561 Chr.20: 2403676-2403676 on Build GRCh38
rs771465289 Chr.20: 2403691-2403691 on Build GRCh38
rs1018670844 Chr.20: 2403753-2403753 on Build GRCh38
rs748956369 Chr.20: 2403565-2403565 on Build GRCh38
rs540825165 Chr.20: 2403889-2403889 on Build GRCh38
rs752575866 Chr.20: 2403813-2403813 on Build GRCh38
rs375737445 Chr.20: 2403510-2403510 on Build GRCh38
rs745615380 Chr.20: 2403476-2403476 on Build GRCh38
rs778598274 Chr.20: 2403629-2403629 on Build GRCh38
rs183084682 Chr.20: 2403913-2403913 on Build GRCh38
rs746393878 Chr.20: 2403771-2403771 on Build GRCh38
rs6137891 Chr.20: 2403505-2403505 on Build GRCh38
rs890392537 Chr.20: 2403899-2403899 on Build GRCh38
rs756051299 Chr.20: 2403620-2403620 on Build GRCh38
rs762613045 Chr.20: 2403482-2403482 on Build GRCh38
rs772641941 Chr.20: 2403633-2403633 on Build GRCh38
rs776229291 Chr.20: 2403552-2403552 on Build GRCh38
rs979829530 Chr.20: 2403467-2403467 on Build GRCh38
rs368495022 Chr.20: 2403504-2403504 on Build GRCh38
rs747661993 Chr.20: 2403630-2403630 on Build GRCh38
rs779923037 Chr.20: 2403558-2403558 on Build GRCh38
rs371729931 Chr.20: 2403532-2403532 on Build GRCh38
rs377161331 Chr.20: 2403843-2403843 on Build GRCh38
rs749713521 Chr.20: 2403840-2403840 on Build GRCh38
rs774694098 Chr.20: 2403791-2403791 on Build GRCh38
rs148889050 Chr.20: 2403595-2403595 on Build GRCh38
rs755125540 Chr.20: 2403524-2403524 on Build GRCh38
rs145554251 Chr.20: 2403575-2403575 on Build GRCh38
rs1055833106 Chr.20: 2403692-2403692 on Build GRCh38
rs902902939 Chr.20: 2403895-2403895 on Build GRCh38
rs775654680 Chr.20: 2403711-2403711 on Build GRCh38
rs201204911 Chr.20: 2403682-2403682 on Build GRCh38
rs1051358284 Chr.20: 2403525-2403525 on Build GRCh38
rs144338465 Chr.20: 2403703-2403703 on Build GRCh38
rs775274303 Chr.20: 2403871-2403871 on Build GRCh38

View Full Product Details