Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 66924523-66925048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIER1
Entrez Gene ID
57708
Gene Name
MIER1 transcriptional regulator
Gene Aliases
ER1, MI-ER1
UniGene
Hs.605432
Gene Chromosome Location
Chr.1: 66924895-66988619 on Build GRCh38
Transcripts
NM_001077700.2 NM_001077702.2 NM_001077703.2 NM_001146110.1 NM_001146111.1 NM_001146112.1 NM_001146113.1 NM_001278215.1 NM_020948.3 XM_011541864.2 XM_011541865.2 XM_017001921.1 XM_017001923.1 XM_017001926.1 XM_017001929.1 XM_017001932.1 NM_024763.4 NM_207014.2 XM_005271212.3 XM_011542161.2 XM_011542162.2 XM_017002353.1 XM_017002354.1 XM_017002357.1

Gene Symbol
WDR78
Entrez Gene ID
79819
Gene Name
WD repeat domain 78
Gene Aliases
DIC4
UniGene
Hs.49421
Gene Chromosome Location
Chr.1: 66812885-66924887 on Build GRCh38
Transcripts
NM_001077700.2 NM_001077702.2 NM_001077703.2 NM_001146110.1 NM_001146111.1 NM_001146112.1 NM_001146113.1 NM_001278215.1 NM_020948.3 XM_011541864.2 XM_011541865.2 XM_017001921.1 XM_017001923.1 XM_017001926.1 XM_017001929.1 XM_017001932.1 NM_024763.4 NM_207014.2 XM_005271212.3 XM_011542161.2 XM_011542162.2 XM_017002353.1 XM_017002354.1 XM_017002357.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5420856 c.1A>C p.M1L Chr.1: 66925029-66925029 on Build GRCh38
911785 c.18A>G p.S6S Chr.1: 66925046-66925046 on Build GRCh38
4241018 c.119C>T p.A40V Chr.1: 66924713-66924713 on Build GRCh38
1344030 c.89delA p.K30fs*28 Chr.1: 66924743-66924743 on Build GRCh38
6822210 c.38C>T p.A13V Chr.1: 66924794-66924794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557595505 Chr.1: 66924749-66924749 on Build GRCh38
rs940732036 Chr.1: 66924884-66924884 on Build GRCh38
rs555535237 Chr.1: 66924635-66924635 on Build GRCh38
rs758810810 Chr.1: 66924881-66924881 on Build GRCh38
rs953893842 Chr.1: 66924981-66924981 on Build GRCh38
rs753172434 Chr.1: 66924825-66924825 on Build GRCh38
rs745526269 Chr.1: 66924657-66924657 on Build GRCh38
rs761657027 Chr.1: 66924563-66924563 on Build GRCh38
rs765135400 Chr.1: 66924624-66924624 on Build GRCh38
rs543074844 Chr.1: 66924952-66924952 on Build GRCh38
rs765827222 Chr.1: 66924807-66924807 on Build GRCh38
rs895026473 Chr.1: 66924912-66924912 on Build GRCh38
rs759767089 Chr.1: 66924790-66924790 on Build GRCh38
rs752119113 Chr.1: 66924983-66924983 on Build GRCh38
rs752595113 Chr.1: 66924632-66924632 on Build GRCh38
rs147347299 Chr.1: 66924755-66924755 on Build GRCh38
rs961038083 Chr.1: 66924945-66924945 on Build GRCh38
rs781356463 Chr.1: 66924848-66924848 on Build GRCh38
rs1051720093 Chr.1: 66924690-66924690 on Build GRCh38
rs761745740 Chr.1: 66924743-66924743 on Build GRCh38
rs983818591 Chr.1: 66924663-66924663 on Build GRCh38
rs748016456 Chr.1: 66924858-66924858 on Build GRCh38
rs764977378 Chr.1: 66924876-66924876 on Build GRCh38
rs769034927 Chr.1: 66924684-66924684 on Build GRCh38
rs774142095 Chr.1: 66924797-66924797 on Build GRCh38
rs771585062 Chr.1: 66924975-66924975 on Build GRCh38
rs1035633965 Chr.1: 66924594-66924594 on Build GRCh38
rs753492390 Chr.1: 66924809-66924809 on Build GRCh38
rs778416970 Chr.1: 66924776-66924776 on Build GRCh38
rs771344086 Chr.1: 66924730-66924730 on Build GRCh38
rs755617655 Chr.1: 66924753-66924753 on Build GRCh38
rs754598952 Chr.1: 66924811-66924811 on Build GRCh38
rs764969355 Chr.1: 66924844-66924845 on Build GRCh38
rs749424771 Chr.1: 66925023-66925023 on Build GRCh38
rs751486869 Chr.1: 66924742-66924742 on Build GRCh38
rs573442932 Chr.1: 66924899-66924899 on Build GRCh38
rs902791433 Chr.1: 66924556-66924556 on Build GRCh38
rs757447164 Chr.1: 66924840-66924840 on Build GRCh38
rs376457210 Chr.1: 66924857-66924857 on Build GRCh38
rs746096241 Chr.1: 66924849-66924849 on Build GRCh38
rs529839273 Chr.1: 66924922-66924922 on Build GRCh38
rs748648195 Chr.1: 66924705-66924705 on Build GRCh38
rs184071846 Chr.1: 66924913-66924913 on Build GRCh38
rs373053790 Chr.1: 66924602-66924602 on Build GRCh38
rs977076473 Chr.1: 66924962-66924964 on Build GRCh38
rs552071959 Chr.1: 66925017-66925017 on Build GRCh38
rs991241553 Chr.1: 66924610-66924610 on Build GRCh38
rs758261154 Chr.1: 66924639-66924639 on Build GRCh38
rs763472569 Chr.1: 66924772-66924781 on Build GRCh38
rs771109477 Chr.1: 66924784-66924784 on Build GRCh38
rs772048720 Chr.1: 66924709-66924709 on Build GRCh38
rs774662919 Chr.1: 66924742-66924743 on Build GRCh38
rs747466472 Chr.1: 66924838-66924838 on Build GRCh38
rs902179089 Chr.1: 66924957-66924957 on Build GRCh38
rs776176793 Chr.1: 66924869-66924869 on Build GRCh38
rs1029148181 Chr.1: 66924985-66924985 on Build GRCh38
rs1038181675 Chr.1: 66924885-66924885 on Build GRCh38
rs757783003 Chr.1: 66924986-66924986 on Build GRCh38
rs144918004 Chr.1: 66924766-66924766 on Build GRCh38
rs1021220083 Chr.1: 66924546-66924546 on Build GRCh38
rs749184670 Chr.1: 66924962-66924962 on Build GRCh38
rs772692432 Chr.1: 66924775-66924775 on Build GRCh38
rs780717604 Chr.1: 66924650-66924650 on Build GRCh38
rs200781785 Chr.1: 66924760-66924760 on Build GRCh38
rs760228696 Chr.1: 66924803-66924803 on Build GRCh38
rs779338538 Chr.1: 66924689-66924689 on Build GRCh38
rs370390521 Chr.1: 66924772-66924772 on Build GRCh38
rs756116755 Chr.1: 66925014-66925014 on Build GRCh38
rs534984894 Chr.1: 66924664-66924664 on Build GRCh38
rs200861654 Chr.1: 66924738-66924738 on Build GRCh38
rs758964282 Chr.1: 66924874-66924874 on Build GRCh38
rs781762234 Chr.1: 66924997-66924997 on Build GRCh38
rs760255666 Chr.1: 66924752-66924752 on Build GRCh38
rs758656000 Chr.1: 66924830-66924830 on Build GRCh38
rs766952352 Chr.1: 66924745-66924745 on Build GRCh38
rs767583552 Chr.1: 66924614-66924614 on Build GRCh38
rs3008858 Chr.1: 66924733-66924733 on Build GRCh38
rs987710139 Chr.1: 66924856-66924856 on Build GRCh38
rs763904134 Chr.1: 66924740-66924740 on Build GRCh38
rs954979258 Chr.1: 66924853-66924853 on Build GRCh38
rs760537439 Chr.1: 66924864-66924864 on Build GRCh38
rs748616434 Chr.1: 66924763-66924763 on Build GRCh38
rs922244855 Chr.1: 66924631-66924631 on Build GRCh38
rs750329559 Chr.1: 66924800-66924800 on Build GRCh38
rs778470350 Chr.1: 66924831-66924831 on Build GRCh38
rs966466831 Chr.1: 66924559-66924559 on Build GRCh38
rs985288466 Chr.1: 66925012-66925012 on Build GRCh38
rs907935015 Chr.1: 66924707-66924707 on Build GRCh38
rs560719510 Chr.1: 66924915-66924915 on Build GRCh38
rs761840067 Chr.1: 66924613-66924613 on Build GRCh38
rs532150107 Chr.1: 66925006-66925006 on Build GRCh38
rs1054984455 Chr.1: 66924896-66924896 on Build GRCh38
rs771835159 Chr.1: 66924859-66924859 on Build GRCh38
rs773337180 Chr.1: 66924617-66924617 on Build GRCh38
rs533760034 Chr.1: 66924893-66924893 on Build GRCh38
rs749722266 Chr.1: 66924682-66924682 on Build GRCh38
rs768342827 Chr.1: 66924793-66924793 on Build GRCh38
rs374949940 Chr.1: 66924875-66924875 on Build GRCh38
rs763974624 Chr.1: 66924646-66924646 on Build GRCh38
rs999226591 Chr.1: 66924960-66924960 on Build GRCh38
rs765885051 Chr.1: 66924867-66924867 on Build GRCh38
rs756743616 Chr.1: 66924649-66924649 on Build GRCh38
rs954896912 Chr.1: 66925007-66925007 on Build GRCh38
rs957816553 Chr.1: 66924978-66924978 on Build GRCh38
rs764783975 Chr.1: 66924812-66924812 on Build GRCh38
rs773283184 Chr.1: 66924715-66924715 on Build GRCh38
rs776252304 Chr.1: 66924784-66924785 on Build GRCh38
rs1013496792 Chr.1: 66924914-66924914 on Build GRCh38
rs974949288 Chr.1: 66924837-66924837 on Build GRCh38
rs781041867 Chr.1: 66924561-66924561 on Build GRCh38
rs779724129 Chr.1: 66924754-66924754 on Build GRCh38
rs1886686 Chr.1: 66924785-66924785 on Build GRCh38
rs1024689822 Chr.1: 66924555-66924555 on Build GRCh38
rs771478503 Chr.1: 66924679-66924680 on Build GRCh38
rs995385810 Chr.1: 66924741-66924741 on Build GRCh38
rs1027814000 Chr.1: 66924908-66924908 on Build GRCh38
rs773157739 Chr.1: 66924861-66924861 on Build GRCh38
rs920697186 Chr.1: 66924890-66924890 on Build GRCh38
rs907861163 Chr.1: 66924863-66924863 on Build GRCh38
rs553668325 Chr.1: 66924798-66924798 on Build GRCh38
rs1015710389 Chr.1: 66924929-66924929 on Build GRCh38
rs767558804 Chr.1: 66924799-66924799 on Build GRCh38
rs747121043 Chr.1: 66924779-66924779 on Build GRCh38
rs760822543 Chr.1: 66924720-66924720 on Build GRCh38
rs201429485 Chr.1: 66924722-66924722 on Build GRCh38
rs779268125 Chr.1: 66924762-66924762 on Build GRCh38
rs752266413 Chr.1: 66924879-66924879 on Build GRCh38
rs761605468 Chr.1: 66924798-66924800 on Build GRCh38
rs764509868 Chr.1: 66924982-66924982 on Build GRCh38
rs879017521 Chr.1: 66924880-66924880 on Build GRCh38
rs751488520 Chr.1: 66924648-66924648 on Build GRCh38
rs563147879 Chr.1: 66924965-66924965 on Build GRCh38

View Full Product Details