Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 67092123-67092672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf141
Entrez Gene ID
400757
Gene Name
chromosome 1 open reading frame 141
Gene Aliases
-
UniGene
Hs.666621
Gene Chromosome Location
Chr.1: 67092176-67141646 on Build GRCh38
Transcripts
NM_001276351.1 NM_001276352.1 NR_075077.1 XM_011541465.2 XM_011541466.2 XM_011541467.1 XM_011541469.1 XM_017001276.1

SNP Detail

SNP ID Chromosome Location
rs995323562 Chr.1: 67092394-67092394 on Build GRCh38
rs753820878 Chr.1: 67092503-67092503 on Build GRCh38
rs41302808 Chr.1: 67092259-67092259 on Build GRCh38
rs899265011 Chr.1: 67092390-67092390 on Build GRCh38
rs928225174 Chr.1: 67092601-67092601 on Build GRCh38
rs958042385 Chr.1: 67092558-67092558 on Build GRCh38
rs921045007 Chr.1: 67092147-67092147 on Build GRCh38
rs74486993 Chr.1: 67092574-67092574 on Build GRCh38
rs6672126 Chr.1: 67092380-67092380 on Build GRCh38
rs1003499867 Chr.1: 67092185-67092185 on Build GRCh38
rs958211701 Chr.1: 67092159-67092159 on Build GRCh38
rs965255843 Chr.1: 67092414-67092414 on Build GRCh38
rs996942245 Chr.1: 67092497-67092497 on Build GRCh38
rs757749994 Chr.1: 67092458-67092458 on Build GRCh38
rs914125566 Chr.1: 67092200-67092200 on Build GRCh38
rs562341047 Chr.1: 67092364-67092364 on Build GRCh38
rs6672194 Chr.1: 67092283-67092283 on Build GRCh38
rs989631206 Chr.1: 67092571-67092571 on Build GRCh38
rs11806560 Chr.1: 67092592-67092592 on Build GRCh38
rs976690360 Chr.1: 67092424-67092424 on Build GRCh38
rs990071040 Chr.1: 67092163-67092163 on Build GRCh38
rs866611746 Chr.1: 67092371-67092371 on Build GRCh38
rs968555177 Chr.1: 67092322-67092322 on Build GRCh38
rs867127882 Chr.1: 67092578-67092578 on Build GRCh38
rs549369903 Chr.1: 67092645-67092645 on Build GRCh38
rs187179949 Chr.1: 67092237-67092237 on Build GRCh38
rs6687611 Chr.1: 67092486-67092486 on Build GRCh38
rs540835144 Chr.1: 67092552-67092552 on Build GRCh38
rs927965131 Chr.1: 67092341-67092341 on Build GRCh38
rs1028052042 Chr.1: 67092529-67092529 on Build GRCh38
rs918287663 Chr.1: 67092457-67092457 on Build GRCh38
rs1052697804 Chr.1: 67092616-67092616 on Build GRCh38
rs773006587 Chr.1: 67092408-67092408 on Build GRCh38
rs575197212 Chr.1: 67092427-67092427 on Build GRCh38
rs11803348 Chr.1: 67092461-67092461 on Build GRCh38
rs78778857 Chr.1: 67092561-67092561 on Build GRCh38
rs796928012 Chr.1: 67092468-67092468 on Build GRCh38
rs938003407 Chr.1: 67092595-67092595 on Build GRCh38
rs948241218 Chr.1: 67092580-67092580 on Build GRCh38
rs544344325 Chr.1: 67092453-67092453 on Build GRCh38
rs892492410 Chr.1: 67092641-67092641 on Build GRCh38
rs939631368 Chr.1: 67092611-67092611 on Build GRCh38
rs367779051 Chr.1: 67092164-67092164 on Build GRCh38
rs965026974 Chr.1: 67092488-67092488 on Build GRCh38
rs765916439 Chr.1: 67092196-67092196 on Build GRCh38

View Full Product Details