Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 69980199-69980718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC7
Entrez Gene ID
57554
Gene Name
leucine rich repeat containing 7
Gene Aliases
DENSIN
UniGene
Hs.479658
Gene Chromosome Location
Chr.1: 69567130-70152426 on Build GRCh38
Transcripts
NM_020794.2 XM_017001885.1 XM_017001886.1 XM_017001887.1 XM_017001888.1 XM_017001889.1 XM_017001890.1 XM_017001891.1 XM_017001892.1 XM_017001893.1 XM_017001894.1 XM_017001895.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5702337 c.662_663TG>GT p.V221G Chr.1: 69980443-69980444 on Build GRCh38
5479244 c.664T>C p.L222L Chr.1: 69980445-69980445 on Build GRCh38
3492095 c.613C>T p.Q205* Chr.1: 69980394-69980394 on Build GRCh38
426649 c.666A>G p.L222L Chr.1: 69980447-69980447 on Build GRCh38
1256854 c.646A>G p.N216D Chr.1: 69980427-69980427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002150690 Chr.1: 69980549-69980549 on Build GRCh38
rs764432719 Chr.1: 69980348-69980348 on Build GRCh38
rs994637911 Chr.1: 69980626-69980626 on Build GRCh38
rs777168970 Chr.1: 69980450-69980450 on Build GRCh38
rs767682677 Chr.1: 69980494-69980494 on Build GRCh38
rs940884172 Chr.1: 69980615-69980615 on Build GRCh38
rs780652273 Chr.1: 69980244-69980244 on Build GRCh38
rs922389381 Chr.1: 69980657-69980657 on Build GRCh38
rs889747519 Chr.1: 69980231-69980231 on Build GRCh38
rs112804003 Chr.1: 69980576-69980576 on Build GRCh38
rs777524400 Chr.1: 69980374-69980374 on Build GRCh38
rs776127143 Chr.1: 69980473-69980473 on Build GRCh38
rs943587557 Chr.1: 69980319-69980319 on Build GRCh38
rs906465045 Chr.1: 69980539-69980539 on Build GRCh38
rs899032224 Chr.1: 69980620-69980620 on Build GRCh38
rs12046022 Chr.1: 69980499-69980499 on Build GRCh38
rs755974555 Chr.1: 69980353-69980353 on Build GRCh38
rs779266349 Chr.1: 69980648-69980648 on Build GRCh38
rs145148783 Chr.1: 69980300-69980300 on Build GRCh38
rs754758861 Chr.1: 69980498-69980498 on Build GRCh38
rs966555334 Chr.1: 69980330-69980330 on Build GRCh38
rs527978643 Chr.1: 69980690-69980690 on Build GRCh38
rs765799194 Chr.1: 69980329-69980329 on Build GRCh38
rs779799696 Chr.1: 69980364-69980364 on Build GRCh38
rs868859917 Chr.1: 69980375-69980375 on Build GRCh38
rs28654288 Chr.1: 69980409-69980409 on Build GRCh38
rs950837420 Chr.1: 69980667-69980667 on Build GRCh38
rs762882678 Chr.1: 69980343-69980343 on Build GRCh38
rs770437026 Chr.1: 69980469-69980469 on Build GRCh38
rs764048134 Chr.1: 69980477-69980477 on Build GRCh38
rs751854713 Chr.1: 69980478-69980478 on Build GRCh38
rs763159387 Chr.1: 69980474-69980474 on Build GRCh38
rs778769007 Chr.1: 69980366-69980366 on Build GRCh38
rs926144661 Chr.1: 69980352-69980352 on Build GRCh38
rs960985147 Chr.1: 69980471-69980471 on Build GRCh38
rs749612704 Chr.1: 69980304-69980304 on Build GRCh38
rs945066189 Chr.1: 69980528-69980528 on Build GRCh38
rs866811213 Chr.1: 69980363-69980363 on Build GRCh38
rs746621683 Chr.1: 69980396-69980396 on Build GRCh38
rs377699321 Chr.1: 69980537-69980537 on Build GRCh38
rs551907912 Chr.1: 69980334-69980334 on Build GRCh38
rs770659764 Chr.1: 69980411-69980411 on Build GRCh38
rs1055030139 Chr.1: 69980573-69980573 on Build GRCh38
rs754935557 Chr.1: 69980365-69980365 on Build GRCh38
rs753630237 Chr.1: 69980495-69980495 on Build GRCh38
rs751724425 Chr.1: 69980349-69980349 on Build GRCh38
rs562936932 Chr.1: 69980292-69980292 on Build GRCh38
rs761960801 Chr.1: 69980489-69980489 on Build GRCh38
rs1026400925 Chr.1: 69980658-69980658 on Build GRCh38
rs543066406 Chr.1: 69980285-69980285 on Build GRCh38
rs752548614 Chr.1: 69980503-69980503 on Build GRCh38
rs71733859 Chr.1: 69980613-69980614 on Build GRCh38
rs373319851 Chr.1: 69980376-69980376 on Build GRCh38
rs772131034 Chr.1: 69980602-69980603 on Build GRCh38
rs759914566 Chr.1: 69980459-69980459 on Build GRCh38
rs373806375 Chr.1: 69980370-69980370 on Build GRCh38

View Full Product Details