Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74568293-74568808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH3
Entrez Gene ID
127254
Gene Name
glutamate rich 3
Gene Aliases
C1orf173
UniGene
Hs.531182
Gene Chromosome Location
Chr.1: 74568111-74674343 on Build GRCh38
Transcripts
NM_001002912.4 XM_017000274.1 XM_017000276.1 XM_017000277.1 XM_017000278.1 XM_017000279.1 XM_017000280.1 XM_017000281.1 XM_017000282.1 XM_017000283.1 XM_017000284.1 XM_017000285.1

SNP Detail

SNP ID Chromosome Location
rs528000992 Chr.1: 74568685-74568685 on Build GRCh38
rs758592883 Chr.1: 74568426-74568426 on Build GRCh38
rs573325102 Chr.1: 74568709-74568709 on Build GRCh38
rs185405674 Chr.1: 74568635-74568635 on Build GRCh38
rs1004863093 Chr.1: 74568755-74568755 on Build GRCh38
rs923587995 Chr.1: 74568728-74568728 on Build GRCh38
rs557430246 Chr.1: 74568552-74568552 on Build GRCh38
rs553611416 Chr.1: 74568611-74568611 on Build GRCh38
rs1014376977 Chr.1: 74568679-74568679 on Build GRCh38
rs963163391 Chr.1: 74568771-74568771 on Build GRCh38
rs1056627093 Chr.1: 74568750-74568750 on Build GRCh38
rs559163816 Chr.1: 74568774-74568774 on Build GRCh38
rs1005569966 Chr.1: 74568445-74568445 on Build GRCh38
rs986439808 Chr.1: 74568577-74568577 on Build GRCh38
rs902282391 Chr.1: 74568352-74568352 on Build GRCh38
rs894578274 Chr.1: 74568617-74568617 on Build GRCh38
rs985606560 Chr.1: 74568622-74568622 on Build GRCh38
rs540845513 Chr.1: 74568725-74568725 on Build GRCh38
rs571739938 Chr.1: 74568394-74568394 on Build GRCh38
rs368057451 Chr.1: 74568502-74568502 on Build GRCh38
rs911481056 Chr.1: 74568632-74568632 on Build GRCh38
rs755744552 Chr.1: 74568571-74568571 on Build GRCh38
rs1043349151 Chr.1: 74568343-74568343 on Build GRCh38
rs955736805 Chr.1: 74568605-74568605 on Build GRCh38
rs28506970 Chr.1: 74568775-74568775 on Build GRCh38
rs1016933804 Chr.1: 74568548-74568548 on Build GRCh38
rs543294063 Chr.1: 74568585-74568585 on Build GRCh38
rs921013242 Chr.1: 74568513-74568513 on Build GRCh38
rs371619148 Chr.1: 74568759-74568759 on Build GRCh38
rs755124817 Chr.1: 74568382-74568382 on Build GRCh38
rs765493161 Chr.1: 74568392-74568392 on Build GRCh38
rs1029864557 Chr.1: 74568602-74568602 on Build GRCh38
rs1056321296 Chr.1: 74568367-74568367 on Build GRCh38
rs935118998 Chr.1: 74568737-74568737 on Build GRCh38
rs972810542 Chr.1: 74568597-74568597 on Build GRCh38
rs747361636 Chr.1: 74568776-74568776 on Build GRCh38
rs570725865 Chr.1: 74568353-74568353 on Build GRCh38
rs187778141 Chr.1: 74568341-74568341 on Build GRCh38
rs1012050003 Chr.1: 74568402-74568402 on Build GRCh38
rs576003540 Chr.1: 74568559-74568559 on Build GRCh38
rs896199748 Chr.1: 74568758-74568758 on Build GRCh38
rs913594731 Chr.1: 74568579-74568579 on Build GRCh38
rs1043684846 Chr.1: 74568596-74568596 on Build GRCh38
rs192720029 Chr.1: 74568616-74568616 on Build GRCh38
rs955017037 Chr.1: 74568532-74568532 on Build GRCh38
rs1017193371 Chr.1: 74568765-74568765 on Build GRCh38
rs887154330 Chr.1: 74568731-74568731 on Build GRCh38
rs1044877297 Chr.1: 74568780-74568780 on Build GRCh38
rs539684356 Chr.1: 74568408-74568408 on Build GRCh38
rs778165106 Chr.1: 74568551-74568551 on Build GRCh38
rs954548576 Chr.1: 74568564-74568564 on Build GRCh38
rs975097383 Chr.1: 74568512-74568512 on Build GRCh38
rs979093780 Chr.1: 74568710-74568710 on Build GRCh38

View Full Product Details